log in or register
Price Range:
Leasing Available
Financing is avaliable for many of our products.
See Details
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
Home  >  Life Science > Electrophoresis Products > Electrophoresis Power Supplies > Programmable Power Supplies for Electrophoresis
Programmable Power Supplies for Electrophoresis
Programmable Power Supplies for Electrophoresis
  • Safe and reliable power for electrophoresis equipment – full overload protection
These power suppliers are simple to operate – just connect to your electrophoresis cell and select the desired voltages. All models feature a two-line LED display that indicates voltage, current, power, and run time. Built-in timers ensure repeatable, controlled separations – set time from 0 to 99 hours, 59 minutes or 0 to 99.99 kilovolt-hours. Each unit can store up to nine sets of frequently used parameters, each with nine steps, in nonvolatile memory. Select constant current, constant voltage, or constant power – automatic crossover ensures stability of operating parameters. Program runs to count down by time or volt-hours. Three or four outputs let you control multiple separations at once.
What's included: 6-ft cord with U.S. standard plug (European plug on 230 VAC models).
 Product Detail
Sort
 Availability
 Price
Representative photo only Programmable Power Supply 6000 VDC 150 mA 115V
Programmable Power Supply, 6000 VDC/150 mA, 115V
Dimensions " W 12-1/4,Dimensions " H 5-7/8,Dimensions " D 10-1/4
EW-28403-88
Call 1-888-358-4705 for availability.
$3,490.00 USD/EACH
Qty:
EW-28403-60 Programmable Power Supply, 400 VDC/300 mA, 115V
Programmable Power Supply, 400 VDC/300 mA, 115V
Dimensions " W 9-1/2,Dimensions " H 5-1/8,Dimensions " D 7-7/8
EW-28403-60
Call 1-888-358-4705 for availability.
$595.00 USD/EACH
Qty:
Representative photo only Programmable Power Supply 1200 VDC 500 mA 115V
Programmable Power Supply, 1200 VDC/500 mA, 115V
Dimensions " W 12-1/4,Dimensions " H 5-7/8,Dimensions " D 10-1/4
EW-28403-68
Call 1-888-358-4705 for availability.
$2,150.00 USD/EACH
Qty:
Representative photo only Programmable Power Supply 300 VDC 1000 mA 230V
Programmable Power Supply, 300 VDC/1000 mA, 230V
Dimensions " W 12-1/4,Dimensions " H 5-7/8,Dimensions " D 10-1/4
EW-28403-74
Call 1-888-358-4705 for availability.
$1,450.00 USD/EACH
Qty:
Representative photo only Programmable Power Supply 600 VDC 1000 mA 230V
Programmable Power Supply, 600 VDC/1000 mA, 230V
Dimensions " W 12-1/4,Dimensions " H 5-7/8,Dimensions " D 10-1/4
EW-28403-86
Call 1-888-358-4705 for availability.
$2,060.00 USD/EACH
Qty:
Representative photo only Programmable Power Supply 300 VDC 1000 mA 115V
Programmable Power Supply, 300 VDC/1000 mA, 115V
Dimensions " W 12-1/4,Dimensions " H 5-7/8,Dimensions " D 10-1/4
EW-28403-72
Call 1-888-358-4705 for availability.
$1,450.00 USD/EACH
Qty:
Representative photo only Programmable Power Supply 1200 VDC 500 mA 230V
Programmable Power Supply, 1200 VDC/500 mA, 230V
Dimensions " W 12-1/4,Dimensions " H 5-7/8,Dimensions " D 10-1/4
EW-28403-70
Call 1-888-358-4705 for availability.
$2,150.00 USD/EACH
Qty:
Representative photo only Programmable Power Supply 600 VDC 500 mA 115V
Programmable Power Supply, 600 VDC/500 mA, 115V
Dimensions " W 12-1/4,Dimensions " H 5-7/8,Dimensions " D 10-1/4
EW-28403-92
Call 1-888-358-4705 for availability.
$1,770.00 USD/EACH
Qty:
Representative photo only Programmable Power Supply 600 VDC 500 mA 230V
Programmable Power Supply, 600 VDC/500 mA, 230V
Dimensions " W 12-1/4,Dimensions " H 5-7/8,Dimensions " D 10-1/4
EW-28403-94
Call 1-888-358-4705 for availability.
$1,770.00 USD/EACH
Qty:
Representative photo only Programmable Power Supply 400 VDC 300 mA 230V
Programmable Power Supply, 400 VDC/300 mA, 230V
Dimensions " W 9-1/2,Dimensions " H 5-1/8,Dimensions " D 7-7/8
EW-28403-62
Call 1-888-358-4705 for availability.
$595.00 USD/EACH
Qty:
Representative photo only Programmable Power Supply 300 VDC 2000 mA 230V
Programmable Power Supply, 300 VDC/2000 mA, 230V
Dimensions " W 12-1/4,Dimensions " H 5-7/8,Dimensions " D 10-1/4
EW-28403-66
Call 1-888-358-4705 for availability.
$1,990.00 USD/EACH
Qty:
Representative photo only Programmable Power Supply 3000 VDC 300 mA 230V
Programmable Power Supply, 3000 VDC/300 mA, 230V
Dimensions " W 12-1/4,Dimensions " H 5-7/8,Dimensions " D 10-1/4
EW-28403-82
Call 1-888-358-4705 for availability.
$2,940.00 USD/EACH
Qty:
Representative photo only Programmable Power Supply 3000 VDC 150 mA 230V
Programmable Power Supply, 3000 VDC/150 mA, 230V
Dimensions " W 12-1/4,Dimensions " H 5-7/8,Dimensions " D 10-1/4
EW-28403-78
Call 1-888-358-4705 for availability.
$2,210.00 USD/EACH
Qty:
Representative photo only Programmable Power Supply 600 VDC 1000 mA 115V
Programmable Power Supply, 600 VDC/1000 mA, 115V
Dimensions " W 12-1/4,Dimensions " H 5-7/8,Dimensions " D 10-1/4
EW-28403-84
Call 1-888-358-4705 for availability.
$2,060.00 USD/EACH
Qty:
Representative photo only Programmable Power Supply 6000 VDC 150 mA 230V
Programmable Power Supply, 6000 VDC/150 mA, 230V
Dimensions " W 12-1/4,Dimensions " H 5-7/8,Dimensions " D 10-1/4
EW-28403-90
Call 1-888-358-4705 for availability.
$3,490.00 USD/EACH
Qty:
18 results (showing 1 - 15 )
Per Page 
Home  >  Life Science > Electrophoresis Products > Electrophoresis Power Supplies > Programmable Power Supplies for Electrophoresis
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title