Subtotal = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
Category:
Brand:
Remel (74)
Product Type:
Agar (103)
Accessory (102)
Availability:
In Stock (249)
Capacity (mL):
0 - 20 (46)
21 - 40 (12)
41 - 60 (26)
1.5 (10)
6.3 (1)
Capacity (g):
Material:
Glass (21)
Membrane MWCO (Daltons):
Diameter (mm):
5 to 10 (47)
Max rpm:
15,200 (34)
6000 (20)
13,300 (20)
14,800 (12)
15,000 (11)
2200 (8)
3400 (7)
13,500 (7)
14,000 (6)
17,000 (4)
14,500 (3)
3000 (2)
13,400 (2)
10,000 (2)
3100 (2)
13300 (2)
18,000 (2)
13,200 (1)
3900 (1)
7000 (1)
Max RCF (x g):
25,314 (29)
16,300 (18)
2000 (14)
17,000 (10)
20,800 (8)
21,000 (8)
21,380 (3)
1792 (2)
15,000 (2)
12,100 (2)
8500 (2)
1740 (2)
16,110 (1)
15,500 (1)
4500 (1)
8400 (1)
2200 (1)
19,150 (1)
2800 (1)
1680 (1)
CE Compliance:
Yes (25)
Dimensions (OD (mm)):
17 (15)
29 (9)
16 (6)
11 (5)
28.8 (5)
15.8 (5)
25.4 (4)
69.5 (4)
28 (4)
13 (4)
61.5 (3)
25.7 (3)
28.7 (3)
16.1 (3)
31.8 (2)
61.8 (2)
60 (2)
16.9 (2)
25.5 (2)
15.9 (2)
No. of wells:
10 (12)
14 (11)
12 (3)
20 (3)
15 (3)
30 (2)
29 (2)
10, 14 (2)
24 (1)
15, 20 (1)
Dimensions (L (mm)):
115 (9)
120 (8)
118.9 (4)
119 (3)
139 (3)
160 (2)
133.4 (2)
46 (2)
94.5 (2)
107.7 (2)
89.1 (2)
118 (2)
119.4 (2)
135 (2)
151 (2)
158 (2)
40 (1)
127.7 (1)
169.8 (1)
184.5 (1)
Well shape:
Round (2)
2,600 pages
of answers

FREE
2013/14 Cole-Parmer
Sourcebook
Choose from over 250 pages of products for life science, including tissue culture, electrophoresis, and electroporation. Choose from a number of well known brands including Corning, Thermo Scientific, UVP, EMD Chemicals, Nalge NUNC, and Eppendorf.
Request your copy today.

Leasing Available
Financing is avaliable for many of our products.

See Details
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334

More Unique Lab & Industrial Supplies

 

Proteomics

Proteomics

Cole-Parmer offers a wide range of products for use in the proteomics laboratory. For over 50 years we have been providing solutions to all facets of science. Besides our well known line of Masterflex pumps and tubing, we also provide proteomic products that are sure to fit your needs. Products include vertical electrophoresis systems, dialysis membranes, solid-phase extraction colums, and centrifugal filters. Along with our wide range of products, we also offer expert assistance in finding the right product. For more information or help choosing the right product for your need, contact our Application Specialists.

Proteomics refers to the study of an organism's complete protein profile, including their functions, structures, and modifications over time. The proteome of an organism can differ over time due to varying conditions, unlike the organisms genome that remains constant. These varying protein levels and the modifications that take place within cells are of interest to the proteomic researcher to help identify potential new drugs for the treatment of diseases or other conditions.

Products for Proteomics


  • Sample Preparation Products
    Choose from a wide variety of proteomic sample preparation products including homogenizers, blenders, incubators, centrifuges, and more
  • Separation Products
    Products for proteomic studies include electrophoresis systems, power supplies, multi-well plates, and solid-phase extraction columns
  • Dialysis Products
    Choose from a variety of dialysis products for proteomics including membranes, dialyzers, spin columns, cassettes, and tubes
  • Safety Products
    Safety products for proteomics include gloves, safety glasses and goggles, glove boxes, and biosafety equipment to product you and your samples
  • Additional Laboratory Products
    Choose from a variety of laboratory products and equipment for proteomics including hot plates, microscopes, spectrophotometers, and syringe pumps

Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
 Product Detail
Sort
 Availability
 Price
Representative photo only 15 mL conical bottom disposable plastic centrifuge tubes screw cap rim seal PPCO 500 bag

15 mL conical bottom disposable plastic centrifuge tubes, screw cap/rim seal, PPCO, 500/bag

Product Type Conical-bottom Tubes,Color clear,Dimensions OD (mm) 17
EW-06344-22
In stock.
$124.00 USD/CASE

Qty:

EW-17306-01 Rotor sold separately

Compact Variable-Speed Universal Centrifuges; 115V; 50/60Hz

Product Type Centrifuge,Capacity Dependent upon rotor (see accessories),Max rpm 6000
EW-17306-01
In stock.
$1,850.00 USD/EACH

Qty:

Representative photo only 50 mL conical bottom disposable plastic centrifuge tubes flat top rim seall PPCO 500 bag

50 mL conical bottom disposable plastic centrifuge tubes, flat top/rim seall, PPCO, 500/bag

Product Type Conical-bottom Tubes,Color clear,Dimensions OD (mm) 29
EW-06344-27
In stock.
$164.00 USD/CASE

Qty:

EW-14201-52 Representative Image. Check individual details.

Prepared agar plate, PDA (potato dextrose agar), 10/pk

Product Type Agar,Description PDA (Potato Dextrose Agar), for cultivation of yeasts and mold and for the inducement of sporulation,Qty/pk 10
EW-14201-52
Usually ships in 7 days.
$16.50 USD/PACKAGE

Qty:

EW-14207-70 Representative image only.  Check individual details.

Thermo Scientific Contact plate sterile tryptic soy agar with lecithin, polysorbate 80, 10/PK

Product Type Prepared Media Plates,Description Irradiated. For cultivation of bacteria, yeast, and fungi.,Media Contact plate sterile tryptic soy agar with lecithin, polysorbate 80,Qty/pk 10
EW-14207-70
Usually ships in 4 days.
$42.75 USD/PACKAGE

Qty:

Representative photo only Fixed speed Centrifuge with 60 minute Timer 115V

Fixed-speed Centrifuge with 60-minute Timer, 115V

Product Type Centrifuge,Max rpm 3400,Power VAC 115
EW-17250-10
In stock.
$450.00 USD/EACH

Qty:

EW-06344-20 15 mL conical bottom disposable plastic centrifuge tubes, screw cap/rim seal, PS, 500/bag

15 mL conical bottom disposable plastic centrifuge tubes, screw cap/rim seal, PS, 500/bag

Product Type Conical-bottom Tubes,Color clear,Dimensions OD (mm) 17
EW-06344-20
In stock.
$123.00 USD/CASE

Qty:

EW-17306-52 Fixed-angle Rotors; 6 place x 50mL

Fixed-angle Rotor; 6 place x 50mL

Product Type Centrifuge Rotor,Capacity six x 50-mL,Max rpm 6000
EW-17306-52
In stock.
$751.90 USD/EACH

Qty:
See all Rotors

EW-44175-02 Cole-Parmer<small><sup>&reg;</sup></small> Orbital Chilling/Heating Block, 100 to 230 VAC

Cole-Parmer Orbital Chilling/Heating Block, 100 to 230 VAC

Product Type Chilling/Heating Blocks,Description Orbital Mixing Chilling/Heating Block,Power VAC 100 to 230
EW-44175-02
Usually ships in 11 days.
$2,205.00 USD/EACH

Qty:

EW-17710-46 Save over $5200 with this centrifuge bundle!
EW-17710-46
Usually ships in 6 days.
$13,455.00 USD/EACH

Qty:

Representative photo only High Speed Programmable Refrigerated Universal Centrifuge 220V

High-Speed Programmable Refrigerated Universal Centrifuge; 220V

Product Type Refrigerated Centrifuge,Max rpm 17,000,Power VAC 220
EW-17303-15
Call 1-888-358-4705 for availability.
$10,155.00 USD/EACH

Qty:

Representative photo only Cole Parmer Microcentrifuge Tube 2 0mL natural non sterile

Cole-Parmer Microcentrifuge Tube, 2.0mL, natural, non sterile

Product Type Microcentrifuge Tubes,Color Natural,Capacity mL 2.0
EW-06333-70
In stock.
$29.50 USD/PACKAGE

Qty:

Representative photo only Thermo Scientific Hereaus X1 Centrifuge to 15 200 rpm 120VAC

Thermo Scientific Hereaus X1 Centrifuge, to 15,200 rpm, 120VAC

Product Type Centrifuge,Max rpm 15,200,Power VAC 120
EW-17709-02
Usually ships in 13 days.
$5,682.00 USD/EACH

Qty:

Representative photo only 50 mL self standing tube PPCO Non sterile 20 bags of 25

50 mL self-standing tube, PPCO, Non-sterile, 20 bags of 25

Product Type Conical-bottom Tubes,Sterile Yes,Color clear
EW-06344-25
In stock.
$201.00 USD/CASE

Qty:

Representative photo only Thermo Scientific Nunc Lab Tek Sterile Petri Dishes PS Standard 100mm x 15 mm 320 case

Thermo Scientific Nunc Lab-Tek Sterile Petri Dishes, PS, Standard, 100mm x 15 mm; 320/case

Product Type Petri Dishes,Material Disposable polystyrene,Description Standard
EW-14007-01
In stock.
$165.00 USD/CASE

Qty:

1290 results (showing 1 - 15 )
Per Page 

View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title