Subtotal = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
 

Quinaldic acid

Quinaldic acid
Choose from our comprehensive line of Quinaldic acid and other Qa to Qz products.
 Product Detail
Sort
 Availability
 Price
Representative photo only Quinaldic acid 99 10g

Quinaldic acid 99% (10g)

Product Type Chemical Reagents
EW-88182-80
In stock.
$161.00 USD/EACH

Qty:
See all Quinaldic acid

1 results (showing 1 - 1 )
Per Page 

View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title