Home  >  Safety Products > Radios
Radios
Radios
VHF two-way radios are excellent ways for lab professionals to maintain communications, especially indoors. VHF two-way radios operate on a low frequency and are less affected by weather, interference and other conditions than the high-frequency UHF two-way radios. There are VHF two-way radios that operate at different frequency ranges and power levels to match you application and budget. For small to mid-sized buildings, VHF radios with ranges up to five miles and coverage up to 150,000 square feet and 8 floors are ideal. For mid-sized to large buildings, the UHF radio may be more appropriate with ranges up to 6 miles and coverage up to 250,000 square feet and 20 floors.
Cole-Parmer’s standard radios operate with one watt of power to provide nominal coverage. If extended range is needed, then our radios with two watts of output power will be a better choice. Other features to consider when making your selection are channels provided and VOX capability. Radios with multiple channels allow different communicating groups to operate within the same area without cross-talk interference. A voice activated (VOX) radio provides a convenient means of communicating hands free when the need arises. If you need help selecting VHF two-way radios for your workplace contact a Cole-Parmer Technical Expert today.
 Product Detail
Sort
 Availability
 Price
Representative photo only Single Channel VHF radio
Single-Channel VHF radio
Description VHF 1-Watt,Channels 1,Model XV1100
EW-86399-00
Usually ships in 7 days.
$225.00 USD/EACH
Qty:
EW-86462-00 CLS Two-Way Radio, UHF, 1 watt, 1 channel
CLS Two-Way Radio, UHF, 1 watt, 1 channel
Description UHF 1-Watt,Channels 1,Model CLS1110
EW-86462-00
Usually ships in 6 days.
$189.00 USD/EACH
Qty:
Representative photo only Remote Speaker Microphone for Motorola Spirit radios
Remote Speaker Microphone for Motorola Spirit radios
Description Shoulder mount microphone,Brand Motorola,Model 53862
EW-08983-38
Usually ships in 10 days.
$110.00 USD/EACH
Qty:
EW-86399-04 Six-channel UHF radio
Six-channel UHF radio
Description UHF 2-Watt,Channels 6,Model XU2600
EW-86399-04
Usually ships in 6 days.
$349.00 USD/EACH
Qty:
EW-86462-10 CLS Two-Way Radio, UHF, 1 watt, 4 channel
CLS Two-Way Radio, UHF, 1 watt, 4 channel
Description UHF 1-Watt,Channels 4,Model CLS1410
EW-86462-10
Usually ships in 5 days.
$209.00 USD/EACH
Qty:
5 results (showing 1 - 5 )
Per Page 
Home  >  Safety Products > Radios
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title