United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Category:
ClearPAGE Electrophoresis Precast Gels
(15)
Amresco reagents
(2)
Brand:
FINGERTIGHT III
(2)
Product Type:
Electrophoresis Accessories
(15)
Price Range:
$0 to $100
(15)
$100 to $299
(1)
$300 to $499
(1)
Availability:
In Stock
(1)
Gel size (cm L):
8
(9)
Gel size (cm W):
8.5
(9)
No. of wells:
12
(7)
17
(2)
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Home
>
Life Science
>
Electrophoresis Products
> Reagents
Reagents
A cleanroom cleaner may seem like an oxymoron but such rooms have a unique sanitary standard that most other spaces couldn’t dream of living up to. For example, a single strand of human hair is about 200 times larger than the particle which caused the famous Hubble Telescope to malfunction. So a cleanroom cleaner is essential to keeping such controlled environments operating without costly and devastating delays. When you are selecting the perfect cleanroom cleaner you need to look for products that eliminate the main enemies of restricted environments: airborne particles, fiber, static and residue left by other cleaning products. The particles float from people, products, places and equipment. Since you can’t eliminate those items, look for products with nonvolatile residue (NVR) which are usually in common cleaning products. Make sure your cleanroom cleaner comes to you in sterile packaging. The last thing you want is to bring contamination into your controlled environment through your cleanroom cleaner.
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Compare
Product Detail
Sort
Most Popular
Name
Name Descending
Price (Lowest First)
Price (Highest First)
Availability
Price
Instant Blue Stain, 1 liter
Product Type Electrophoresis Accessories
EW-28570-59
In stock.
$209.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
ClearPAGE Electrophoresis Precast Gels
LDS Sample Buffer 10ml, 4x
Product Type Electrophoresis Accessories
EW-28570-49
Usually ships in 6 days.
$19.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
ClearPAGE Electrophoresis Precast Gels
ClearPAGE
SDS Gel, 4-20% Acrylamide, 12 well, 10 per Package
Product Type Electrophoresis Accessories,No. of wells 12,Gel size cm W 8.5
EW-28570-44
Usually ships in 6 days.
$96.00 USD/PACKAGE
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
ClearPAGE Electrophoresis Precast Gels
ClearPAGE
SDS Gel, 16% Acrylamide, 12 well, 10 per Package
Product Type Electrophoresis Accessories,No. of wells 12,Gel size cm W 8.5
EW-28570-38
Usually ships in 6 days.
$96.00 USD/PACKAGE
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
ClearPAGE Electrophoresis Precast Gels
Turbo Buffer 50mL
Product Type Electrophoresis Accessories
EW-28570-53
Usually ships in 6 days.
$11.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
ClearPAGE Electrophoresis Precast Gels
Agarose III
Electrophoresis Solution, 250g
Description Agarose III
,Brand FINGERTIGHT III
EW-21850-64
Usually ships in 7 days.
$375.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Amresco reagents
ClearPAGE
SDS Gel, 12% Acrylamide, 17 well, 10 per Package
Product Type Electrophoresis Accessories,No. of wells 17,Gel size cm W 8.5
EW-28570-36
Usually ships in 6 days.
$96.00 USD/PACKAGE
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
ClearPAGE Electrophoresis Precast Gels
Turbo Buffer 500 mL
Product Type Electrophoresis Accessories
EW-28570-55
Usually ships in 6 days.
$66.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
ClearPAGE Electrophoresis Precast Gels
ClearPAGE
SDS Gel, 10% Acrylamide, 12 well, 10 per Package
Product Type Electrophoresis Accessories,No. of wells 12,Gel size cm W 8.5
EW-28570-32
Usually ships in 6 days.
$96.00 USD/PACKAGE
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
ClearPAGE Electrophoresis Precast Gels
SDS-Non Reducing Run Buffer 50ml, 20x
Product Type Electrophoresis Accessories
EW-28570-51
Usually ships in 6 days.
$11.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
ClearPAGE Electrophoresis Precast Gels
ClearPAGE
SDS Gel, 4-20% Acrylamide, 17 well, 10 per Package
Product Type Electrophoresis Accessories,No. of wells 17,Gel size cm W 8.5
EW-28570-46
Usually ships in 6 days.
$96.00 USD/PACKAGE
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
ClearPAGE Electrophoresis Precast Gels
Agarose III
Electrophoresis Solution, 25g
Description Agarose III
,Brand FINGERTIGHT III
EW-21850-60
Usually ships in 7 days.
$49.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Amresco reagents
ClearPAGE
SDS Gel, 8% Acrylamide, 12 well, 10 per Package
Product Type Electrophoresis Accessories,No. of wells 12,Gel size cm W 8.5
EW-28570-30
Usually ships in 6 days.
$96.00 USD/PACKAGE
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
ClearPAGE
Electrophoresis Precast Gels
ClearPAGE
SDS Gel, 12% Acrylamide, 12 well, 10 per Package
Product Type Electrophoresis Accessories,No. of wells 12,Gel size cm W 8.5
EW-28570-34
Usually ships in 6 days.
$96.00 USD/PACKAGE
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
ClearPAGE Electrophoresis Precast Gels
ClearPAGE
SDS Gel, 4-12% Acrylamide, 12 well, 10 per Package
Product Type Electrophoresis Accessories,No. of wells 12,Gel size cm W 8.5
EW-28570-42
Usually ships in 6 days.
$96.00 USD/PACKAGE
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
ClearPAGE Electrophoresis Precast Gels
17
results (showing
1
-
15
)
1
2
Next >
>>
Per Page
5
10
15
25
50
Home
>
Life Science
>
Electrophoresis Products
> Reagents
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title