United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Leasing Available
Financing is avaliable for many of our products.
See Details
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Home
>
Lab Equipment
>
Mixers and Blenders
>
Mills
>
Retsch Mixer Mills
> Retsch Mixer Mill
Retsch Mixer Mill
Quick and effectivemost materials can be pulverized and mixed at ambient temperature without any cooling!
Suitable for dry, wet, and cryogenic grinding as well as cell disruption for DNA/RNA recovery
Two grinding stations and short processing times provide high sample throughput
Reproducible results by digital preselection of grinding time and vibrational frequency
This high-performance mixer mill can pulverize two samples from 0.2 to 20 mL at the same time. The compact benchtop model is suitable for size reduction, mixing and pulverization of hard, medium-hard, brittle, soft, elastic and fibrous samples. It is also useful for mixing and homogenization of powders.
The grinding jars perform oscillations in a horizontal position. The inertia of the grinding balls causes them to impact with high energy on the sample material at the rounded ends of the grinding jars and pulverize it. Also, the movement of the grinding jars combined with the movement of the balls result in the intensive mixing of the sample. As a function of frequency, up to 1800 impacts per minute are achieved resulting in a high degree of pulverization in very short times. The degree of mixing can be increased even further by using several smaller balls. If many very small balls are used (e.g. glass beads) then, for example, biological cells can be disrupted. The large frictional impact effects between the beads ensure effective cell disruption.
Order screw-top grinding jars and balls separately. Convenient self-locking clamp allows the use of large jars up to 50 mL.
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Compare
Product Detail
Sort
Most Popular
Name
Name Descending
Price (Lowest First)
Price (Highest First)
Availability
Price
Retsch Mixer Mill MM400; 100-240 VAC, 50/60 Hz
Product Type Mixer Mills,CE Compliance Yes,Brand Retsch
EW-04182-09
Usually ships in 7 days.
$8,260.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Retsch Mixer Mill
1
results (showing
1
-
1
)
Per Page
5
10
15
25
50
Home
>
Lab Equipment
>
Mixers and Blenders
>
Mills
>
Retsch Mixer Mills
> Retsch Mixer Mill
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title