log in or register
Price Range:
Availability:
Mass (g):
10 (1)
30 (1)
50 (1)
100 (1)
150 (1)
200 (1)
300 (1)
400 (1)
500 (1)
1000 (1)
1500 (1)
2000 (1)
3000 (1)
4000 (1)
5000 (1)
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
Home  >  Balances & Scales > Calibration Masses > Rice Lake Calibration Masses > Rice Lake Class 1 Individual Masses with Certificate of Calibration
Rice Lake Class 1 Individual Masses with Certificate of Calibration
Rice Lake Class 1 Individual Masses with Certificate of Calibration
  • Ensure balance accuracy with calibration masses from Rice Lake
  • Weights conform to NIST, ASTM or OIML (unlike internal calibration masses)
  • 4 day turnaround time on new masses and mass recalibrations
  • Individual masses and sets from Class 0 through Class 4
  • Electronic balance calibration masses
  • Specialty masses and custom designs
  • Succinct traceable calibration certificates
Ensure balance accuracy with calibration masses from Rice Lake. Dedicated to excellence and service, Rice Lake offers a 4-day turnaround on masses and calibrations.
Masses are made of polished stainless steel and include a glove, case and lifter unless otherwise indicated. As the leader in service, selection and speed – Rice Lake stands behind our offerings with a commitment beyond measurement.
 Product Detail
Sort
 Availability
 Price
Representative photo only Class 1 calibration mass with certificate of calibration 500 g
Class 1 calibration mass with certificate of calibration, 500 g
Product Type Calibration Mass,Class Class 1,Mass g 500
EW-10150-09
Usually ships in 8 days.
$250.00 USD/EACH
Qty:
Representative photo only Class 1 calibration mass with certificate of calibration 3000 g
Class 1 calibration mass with certificate of calibration, 3000 g
Product Type Calibration Mass,Class Class 1,Mass g 3000
EW-10150-13
Usually ships in 12 days.
$465.00 USD/EACH
Qty:
Representative photo only Class 1 calibration mass with certificate of calibration 1000 g
Class 1 calibration mass with certificate of calibration, 1000 g
Product Type Calibration Mass,Class Class 1,Mass g 1000
EW-10150-10
Usually ships in 15 days.
$275.00 USD/EACH
Qty:
Representative photo only Class 1 calibration mass with certificate of calibration 5000 g
Class 1 calibration mass with certificate of calibration, 5000 g
Product Type Calibration Mass,Class Class 1,Mass g 5000
EW-10150-15
Usually ships in 4 days.
$750.00 USD/EACH
Qty:
Representative photo only Class 1 calibration mass with certificate of calibration 200 g
Class 1 calibration mass with certificate of calibration, 200 g
Product Type Calibration Mass,Class Class 1,Mass g 200
EW-10150-06
Usually ships in 9 days.
$225.00 USD/EACH
Qty:
Representative photo only Class 1 calibration mass with certificate of calibration 4000 g
Class 1 calibration mass with certificate of calibration, 4000 g
Product Type Calibration Mass,Class Class 1,Mass g 4000
EW-10150-14
Usually ships in 7 days.
$680.00 USD/EACH
Qty:
Representative photo only Class 1 calibration mass with certificate of calibration 300 g
Class 1 calibration mass with certificate of calibration, 300 g
Product Type Calibration Mass,Class Class 1,Mass g 300
EW-10150-07
In stock.
$240.00 USD/EACH
Qty:
Representative photo only Class 1 calibration mass with certificate of calibration 2000 g
Class 1 calibration mass with certificate of calibration, 2000 g
Product Type Calibration Mass,Class Class 1,Mass g 2000
EW-10150-12
Usually ships in 8 days.
$330.00 USD/EACH
Qty:
Representative photo only Class 1 calibration mass with certificate of calibration 10 g
Class 1 calibration mass with certificate of calibration, 10 g
Product Type Calibration Mass,Class Class 1,Mass g 10
EW-10150-01
Usually ships in 12 days.
$130.00 USD/EACH
Qty:
EW-10150-04 Class 1 calibration mass with certificate of calibration, 100 g
Class 1 calibration mass with certificate of calibration, 100 g
Product Type Calibration Mass,Class Class 1,Mass g 100
EW-10150-04
Usually ships in 10 days.
$140.00 USD/EACH
Qty:
Representative photo only Class 1 calibration mass with certificate of calibration 400 g
Class 1 calibration mass with certificate of calibration, 400 g
Product Type Calibration Mass,Class Class 1,Mass g 400
EW-10150-08
Usually ships in 9 days.
$250.00 USD/EACH
Qty:
Representative photo only Class 1 calibration mass with certificate of calibration 50 g
Class 1 calibration mass with certificate of calibration , 50 g
Product Type Calibration Mass,Class Class 1,Mass g 50
EW-10150-03
Usually ships in 7 days.
$130.00 USD/EACH
Qty:
Representative photo only Class 1 calibration mass with certificate of calibration 1500 g
Class 1 calibration mass with certificate of calibration, 1500 g
Product Type Calibration Mass,Class Class 1,Mass g 1500
EW-10150-11
Usually ships in 4 days.
$310.00 USD/EACH
Qty:
Representative photo only Class 1 calibration mass with certificate of calibration 150 g
Class 1 calibration mass with certificate of calibration, 150 g
Product Type Calibration Mass,Class Class 1,Mass g 150
EW-10150-05
Usually ships in 4 days.
$220.00 USD/EACH
Qty:
Representative photo only Class 1 calibration mass with certificate of calibration 30 g
Class 1 calibration mass with certificate of calibration, 30 g
Product Type Calibration Mass,Class Class 1,Mass g 30
EW-10150-02
Usually ships in 11 days.
$130.00 USD/EACH
Qty:
15 results (showing 1 - 15 )
Per Page 
Home  >  Balances & Scales > Calibration Masses > Rice Lake Calibration Masses > Rice Lake Class 1 Individual Masses with Certificate of Calibration
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title