United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, Fittings, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Category:
Gloves
(601)
Coveralls
(115)
Lab Coats
(82)
META Unisex Scrubs
(40)
Aprons
(24)
Footwear
(20)
Safety Apparel Sleeves
(11)
Head & Face Covers
(10)
Brand:
Microflex
(109)
Best Glove
(56)
Ansell
(55)
[+]
Show more
(17)
Meta
(40)
META
(40)
High Five
(37)
Tyvek
(32)
Cryo-Glove
(23)
Safeskin
(20)
AURELIA
(19)
QRP Incorporated
(16)
Tychem
(12)
Sperian
(10)
Kleenguard
(9)
North
(9)
Pro/Clean
(8)
Touch-N-Tuff
(8)
Cole-Parmer
(7)
Sol-Vex
(7)
N-DEX
(7)
[-]
Show less
Product Type:
Gloves
(581)
Coveralls
(91)
Lab Coats
(80)
[+]
Show more
(17)
Scrubs
(40)
Aprons
(24)
Finger cots
(12)
Sleeves
(10)
Coveralls with elastic wrists and ankles
(10)
Head & Face Covers
(10)
Coveralls with elastic wrists, hood, and boots
(8)
Shoe & Boot Covers
(8)
Accessory
(4)
Suits
(3)
Hip Boots
(3)
Coveralls with elastic wrists
(2)
Polypropylene Shoe Covers
(2)
Polylatex Shoe Covers
(2)
Chest Waders
(2)
Boot Covers
(1)
Coveralls with elastic wrists and hood
(1)
[-]
Show less
Price Range:
$0 to $100
(725)
$100 to $299
(162)
$300 to $499
(14)
[+]
Show more
(1)
$500 to $799
(1)
[-]
Show less
Availability:
In Stock
(540)
Material:
Nitrile
(180)
Latex
(117)
Rubber
(89)
[+]
Show more
(17)
Polyethylene
(63)
Polyester/Cotton
(60)
65% polyester/35% cotton poplin
(40)
Nylon
(32)
Cotton
(25)
Neoprene
(23)
Nylon exterior, jersey liner
(23)
Leather
(22)
Polypropylene
(21)
Vinyl
(18)
Flame-retardant hydro-entangled wood pulp and polyester
(18)
Butyl
(12)
Non-latex
(10)
Nitrile, neoprene, rubber
(8)
Kevlar
(8)
Chloroprene
(8)
Dyneema
®
/Spectra
(8)
[-]
Show less
Color:
White
(108)
Blue
(28)
Black
(18)
[+]
Show more
(10)
Yellow
(17)
Gray
(13)
Hunter
(10)
Navy blue
(10)
Ceil
(10)
Navy
(9)
Silver
(4)
Royal blue
(4)
Green
(3)
Clear
(1)
[-]
Show less
Thickness (mil):
5
(58)
4
(31)
6
(28)
[+]
Show more
(17)
5.5
(17)
3.5
(16)
5.1
(13)
3
(12)
6.3
(12)
15
(11)
11
(10)
3.1
(9)
5.9
(8)
8
(8)
5.2
(8)
8.3
(7)
30
(6)
28
(6)
4.0
(6)
20
(5)
50
(5)
[-]
Show less
Men's size:
Extra large
(3)
Universal
(2)
11
(2)
[+]
Show more
(2)
12
(2)
10
(1)
[-]
Show less
Womens size:
Extra large
(3)
Universal
(2)
13
(2)
[+]
Show more
(2)
14
(2)
12
(1)
[-]
Show less
CE Compliance:
Yes
(1)
Learn more:
Isolating Genomic DNA from Whole Blood
ABCs of DMMs
[-]
Show Less
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Home
>
Safety Products
> Safety Apparel
Safety Apparel
Using safety apparel is an easy way to protect workers from a variety of hazards. In certain cases, OSHA requirements have to be met to ensure compliance and avoid penalties and fines. Cole-Parmer carries a wide selection of safety apparel to protect workers from spills, chemicals, and damage to personal clothing. Safety clothing that protects workers in laboratories and industrial environments is available in a variety of materials including Tyvek clothing. The type of safety apparel varies. Coveralls offer a physical barrier from spills and dirt while providing the greatest coverage to the worker. Some coveralls come with hoods and boots for head-to-toe protection. Lab coats provide protection to the laboratory worker from chemicals spills and other accidents; flame-resistant lab coats are ideal for use with flammable chemicals. Aprons provide some protection while allowing more freedom of movement and more ventilation. Head and face covers, such as bouffant, hair nets, and face masks protect the worker but also protects what you are working with from hair or germs. Sleeves provide arm protection usually in conjunction with gloves. Shoe covers protect the feet but also prevent contamination from dirt carried on shoes. The best safety apparel is clothing that is comfortable, easily worn and does not hamper a worker’s productivity.
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Compare
Product Detail
Sort
Most Popular
Name
Name Descending
Price (Lowest First)
Price (Highest First)
Availability
Price
Kimberly-Clark Nitrile Gloves, 9.5" length, Medium; 100/box
Product Type Gloves,Material Nitrile,Color Purple,Thickness mil 5
EW-86313-21
In stock.
$20.50 USD/BOX of 100
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Kimberly-Clark
Nitrile Exam Gloves
Kimberly-Clark Nitrile Gloves, 9.5" length, Large; 100/box
Product Type Gloves,Material Nitrile,Thickness mil 5,Color Purple
EW-86313-22
In stock.
$20.50 USD/BOX of 100
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Kimberly-Clark Nitrile Gloves
Microflex FreeForm
SE Nitrile Gloves, large, 100/Box
Product Type Gloves,Material Nitrile,Thickness mil 5.1,Color Blue
EW-86445-02
In stock.
$17.25 USD/BOX
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Microflex FreeForm SE Nitrile Gloves
Microflex Diamond Grip
Latex Gloves; size, large
Product Type Gloves,Material Latex,Color Clear,Thickness mil 7.1
EW-86231-32
In stock.
$18.75 USD/BOX of 100
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Microflex Diamond Grip Latex Gloves
Microflex Diamond Grip
Latex Gloves; size, extra-large
Product Type Gloves,Material Latex,Color Clear,Thickness mil 7.1
EW-86231-33
In stock.
$18.75 USD/BOX of 100
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Microflex Diamond Grip Latex Gloves
Ansell Touch-N-Tuff
Nitrile Gloves, Powder-free, Large
Product Type Gloves,Material Nitrile,Color Green,Thickness mil 4
EW-81602-65
In stock.
$23.00 USD/BOX of 100
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Ansell Touch-N-Tuff Nitrile Gloves
Face mask, ear loop, white, 50/pk
Product Type Head & Face Covers,Description Ear loop,Color White
EW-86495-20
In stock.
$12.50 USD/PACKAGE
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Face Masks
Kimberly-Clark Nitrile Gloves, 9.5" length, Small, 100/box
Product Type Gloves,Material Nitrile,Color Purple,Thickness mil 5
EW-86313-20
In stock.
$20.50 USD/BOX of 100
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Kimberly-Clark Nitrile Gloves
Microflex Diamond Grip
Latex Gloves; size, medium
Product Type Gloves,Material Latex,Color Clear,Thickness mil 7.1
EW-86231-31
In stock.
$18.75 USD/BOX of 100
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Microflex Diamond Grip Latex Gloves
ZetexPlus High-Temperature Gloves, up to 2000
F, one size fits all, 14"L, one pair
Product Type Gloves,Material Cotton,Color White
EW-86364-10
In stock.
$76.00 USD/PAIR
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
ZetexPlus
High-Temperature Gloves
KIMTECH Pure G3 Sterile Critical Nitrile Gloves, medium, bag of 20 pair
Product Type Gloves,Material Nitrile,Color White,Thickness mil 5
EW-86486-01
In stock.
$32.00 USD/BAG
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
KIMTECH Pure G3 Sterile Critical Nitrile Gloves
Microflex FreeForm
SE Nitrile Gloves, medium, 100/Box
Product Type Gloves,Material Nitrile,Thickness mil 5.1,Color Blue
EW-86445-01
In stock.
$17.25 USD/BOX
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Microflex FreeForm SE Nitrile Gloves
High Five A+ Aloe
Nitrile Gloves, Medium, 100/bx
Product Type Gloves,Material Nitrile,Color Green,Thickness mil 5
EW-86480-12
In stock.
$18.00 USD/BOX
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
High Five A+ Aloe Nitrile Gloves
Microflex FreeForm
SE Nitrile Gloves, X-large, 100/Box
Product Type Gloves,Material Nitrile,Color Blue,Thickness mil 5.1
EW-86445-03
In stock.
$17.25 USD/BOX
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Microflex FreeForm SE Nitrile Gloves
Microflex NeoPro
Chloroprene gloves, medium, box of 100
Product Type Gloves,Material Chloroprene,Thickness mil 6,Color Green
EW-86232-51
In stock.
$19.00 USD/BOX
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Microflex
NeoPro
Chloroprene gloves
902
results (showing
1
-
15
)
1
2
3
4
5
6
Next >
>>
Per Page
5
10
15
25
50
Home
>
Safety Products
> Safety Apparel
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title