log in or register
2,600 pages
of answers
FREE
2013/14 Cole-Parmer
Sourcebook
7 pages of safety glasses, plus even more eye protection products are contained in our 2009/10 general catalog. Cole-Parmer carries the coolest styles and latest trends in safety glasses from industry leaders such as Uvex, Wilson, Smith & Wesson, DeWalt, and more.
Request your copy today.
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
Safety Glasses
Safety Glasses
When it comes to safety glasses, Cole-Parmer offers a large selection of types and styles from leading brands such as Uvex, Wilson, Smith & Wesson, DeWalt, AOSafety, and more. Whether your need for safety glasses is in the lab, factory, industrial setting, or to keep your visitors safe, Cole-Parmer has what you need.
OSHA requirements state that protective eye and face devices must comply with ANSI Z87.1 (American National Standard Practice for Occupational and Educational Eye and Face Protection). ANSI Z87.1 requires that all lenses be permanently imprinted with the manufacturer's monogram and all frames marked "Z87". Cole-Parmer's entire selection of safety glasses is ANSI compliant.
Types of Eyewear
  • Safety glasses have impact-resistant lenses and strong frames to protect you from flying particles.
  • Safety goggles are designed to provide a shield around the entire eye area to protect against hazards from many directions.
  • Face shields give you full face protection against sparks, splashes, and splatter. They provide secondary protection and must be worn with protective glasses or goggles.
Lens Tints
  • ClearNormal lighting conditions
  • GrayFor use in traffic safety and general protection against sunlight
  • Amber/yellowLow light and enhanced contrast
  • MirroredOutdoors, anti-glare, and UV protection
  • Indoor/OutdoorEqual visibility protection in natural or fluorescent lighting
  • AntifogProtection from fogging, scratching, and static
  • Welding shade 3.0 and 5.0Welding sites or brazing and cutting
In addition to safety glasses, our eye protection selection also includes Safety Goggles and Face Shields. Need protection from lasers? Cole-Parmer can help there too. We also have options in Laser Eyewear.
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
 Product Detail
Sort
 Availability
 Price
Representative photo only Smith Wesson Magnum Glasses Fog free Clear Lens
Smith & Wesson Magnum Glasses, Fog-free Clear Lens
Product Type Glasses,Color Black,Lens Clear/Anti-fog
EW-86285-01
In stock.
$10.00 USD/EACH
Qty:
Representative photo only 3M Virtua Safety Eyewear Clear frame Clear lens
3M Virtua Safety Eyewear, Clear frame, Clear lens
Product Type Glasses,Color Clear,Lens Clear
EW-86472-70
In stock.
$2.25 USD/EACH
Qty:
EW-86325-70 Black frame glasses with clear lenses
Uvex Genesis Black frame glasses with clear lenses
Product Type Glasses,Color Black,Lens Clear
EW-86325-70
In stock.
$8.75 USD/EACH
Qty:
EW-86472-60 3M<small><sup>&reg;</sup></small> BX<small><sup>&#153;</sup></small> Eyewear, Silver/Black frame, Clear lens
3M BX Eyewear, Silver/Black frame, Clear lens
Product Type Glasses,Color Black and Silver,Lens Clear/Anti-fog
EW-86472-60
In stock.
$5.75 USD/EACH
Qty:
EW-86285-00 Smith & Wesson<small><sup>&reg;</sup></small> Magnum<small><sup>&#153;</sup></small> Glasses, Clear Lens
Smith & Wesson Magnum Glasses, Clear Lens
Product Type Glasses,Color Black,Lens Clear
EW-86285-00
In stock.
$8.25 USD/EACH
Qty:
EW-86321-71 Uvex Ignite Safety Glasses, Black/Silver Frame, Hardcoat Lens Coating, Clear Lens
Uvex Ignite Safety Glasses, Black/Silver Frame, Hardcoat Lens Coating, Clear Lens
Product Type Glasses,Color Black and Silver,Lens Clear
EW-86321-71
In stock.
$7.00 USD/EACH
Qty:
Representative photo only Uvex Seismic Safety Eyewear Clear lens Dual antifog antiscratch coating
EW-86497-62
Usually ships in 10 days or sooner.
$20.00 USD/EACH
Qty:
Representative photo only 3M Tour Guard Visitors Glasses Regular 20 box
3M Tour-Guard Visitors' Glasses, Regular, 20/box
Product Type Glasses,Color Clear,Lens Clear
EW-48496-00
In stock.
$42.00 USD/PACKAGE of 20
Qty:
Representative photo only Encon Veratti 1000 Protective Eyewear Clear Lens Silver Frame
Encon Veratti 1000 Protective Eyewear, Clear Lens, Silver Frame
Product Type Glasses,Color Silver,Lens Clear
EW-86052-82
Usually ships in 8 days or sooner.
$5.00 USD/EACH
Qty:
Representative photo only Smith Wesson Magnum Elite Glasses Clear Lens
Smith & Wesson Magnum Elite Glasses, Clear Lens
Product Type Glasses,Color Platinum,Lens Clear
EW-86285-10
Usually ships in 9 days or sooner.
$8.75 USD/EACH
Qty:
EW-81659-00 Sperian<small><sup>&reg;</sup></small> Polysafe<small><sup>&reg;</sup></small> Visitors' Glasses, Clear Temples
Sperian Polysafe Visitors' Glasses, Clear Temples
Product Type Glasses,Color Clear,Lens Clear
EW-81659-00
In stock.
$2.75 USD/EACH
Qty:
EW-86474-22 Olympic Optical magnifying safety glasses, black frame w/clear lens, +2.0 diopter
EW-86474-22
In stock.
$14.75 USD/EACH
Qty:
EW-86497-01 DeWalt<small><sup>&reg;</sup></small> Protector<small><sup>&#153;</sup></small> Safety Glasses, Clear lens
DeWalt Protector Safety Glasses, Clear lens
Product Type Glasses,Color Clear and Yellow,Lens Clear
EW-86497-01
In stock.
$2.75 USD/EACH
Qty:
Representative photo only Encon Veratti 1000 Protective Eyewear Clear Lens Black Frame
Encon Veratti 1000 Protective Eyewear, Clear Lens, Black Frame
Product Type Glasses,Color Black,Lens Clear
EW-86052-80
In stock.
$5.50 USD/EACH
Qty:
Representative photo only Allsafe SMC Nemesis Safety Glasses Black frame I O lens
Allsafe SMC Nemesis Safety Glasses, Black frame, I/O lens
Product Type Glasses,Color Black,Lens Smoke
EW-86475-12
In stock.
$5.25 USD/EACH
Qty:
118 results (showing 1 - 15 )
Per Page 
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title