log in or register
2,600 pages
of answers
FREE
2013/14 Cole-Parmer
Sourcebook
Browse our complete selection of safety goggles and other eye protection products in our 2009/10 general catalog. We have what you need to keep you safe.
Request your copy today.
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
Safety Goggles
Safety Goggles
Safety goggles are designed to provide a shield around the entire eye area to protect against hazards from many directions. They offer higher impact protection, greater dust, chemical splash, and welding light protection.
Safety goggles for splash or fine dust protection should have indirect venting. Use direct vented goggles for less fogging when working with large particles. Those designed after ski type goggles with high air flow minimize fogging while providing better particle and splash protection. No matter what you need to protect your eyes from, Cole-Parmer has the safety goggles to help.
Use safety goggles, or other eye protection, to protect your eyes from common hazards and injuries:
Common Hazards
  • Dust, concrete, and metal particles
  • Falling or shifting debris, building materials, glass
  • Smoke, noxious/poisonous gases
  • Chemicals (acids, bases, fuels, solvents, lime, wet or dry cement powder)
  • Welding light and electrical arc
  • Thermal hazards and fires
  • Bloodborne pathogens
Common Injuries
  • Corneal abrasions and conjunctivitis (red eyes)
  • Concrete or metal particles or slivers embedded in the eye
  • Chemical splash or burn
  • Eyeball laceration
  • Facial contusion and black eye
  • Bloodborne pathogen exposure
In addition to safety goggles, our eye protection selection also includes safety glasses and face shields. Let Cole-Parmer help protect your eyes with our complete line of safety eyewear.
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
 Product Detail
Sort
 Availability
 Price
EW-86281-01 Sellstrom Odyssey<small><sup>&reg;</sup></small> II Autoclavable Safety Goggles, Nonvented
Sellstrom Odyssey II Autoclavable Safety Goggles, Nonvented
Product Type Safety Goggles,Color Gray,Lens Clear/Anti-fog
EW-86281-01
In stock.
$10.00 USD/EACH
Qty:
EW-86326-00 Uvex<small><sup>&reg;</sup></small> STEALTH Safety Goggles, Clear Lens
Uvex STEALTH Safety Goggles, Clear Lens
Product Type Safety Goggles,Color Gray,Lens Clear
EW-86326-00
In stock.
$15.00 USD/EACH
Qty:
Representative photo only Allsafe MonoGoggle Large Clear Lens
Allsafe MonoGoggle, Large, Clear Lens
Product Type Safety Goggles,Color Smoke,Lens Clear
EW-81657-11
In stock.
$12.75 USD/EACH
Qty:
EW-86326-10 Uvex<small><sup>&reg;</sup></small> FUTURA Safety Goggles
Uvex FUTURA Safety Goggles
Product Type Safety Goggles,Color White,Lens Clear
EW-86326-10
In stock.
$10.25 USD/EACH
Qty:
Representative photo only Encon 500 Series Spherical Goggles Clear Antifogging Lens
Encon 500-Series Spherical Goggles, Clear Antifogging Lens
Product Type Safety Goggles,Color Clear,Lens Clear
EW-01662-14
In stock.
$8.55 USD/EACH
Qty:
Representative photo only Sellstrom Odyssey II Autoclavable Safety Goggles Top vented
Sellstrom Odyssey II Autoclavable Safety Goggles, Top vented
Product Type Safety Goggles,Color White,Lens Clear
EW-86281-02
In stock.
$10.00 USD/EACH
Qty:
Representative photo only 3M Maxim Safety Goggles Over the GlassClear Lens
3M Maxim Safety Goggles Over the GlassClear Lens
Product Type Safety Goggles,Color Black,Lens Clear
EW-86478-09
In stock.
$17.75 USD/EACH
Qty:
EW-86281-52 Odyssey<small><sup>&#153;</sup></small> Goggles, Clear/Fog Free Lens
Odyssey Goggles, Clear/Fog Free Lens
Product Type Safety Goggles,Color Clear,Lens Clear/Anti-fog
EW-86281-52
In stock.
$11.00 USD/EACH
Qty:
EW-86326-22 Uvex Strategy Safety Goggles with Neoprene Band, Gray Body with Indirect Vent, Clear Lens
EW-86326-22
In stock.
$8.00 USD/EACH
Qty:
EW-86527-00 Flex Seal Safety Goggle, clear lens
Flex Seal Safety Goggle, clear lens
Product Type Safety Goggles,Color Gray,Lens Clear
EW-86527-00
In stock.
$19.00 USD/EACH
Qty:
EW-81657-18 Allsafe<small><sup>&reg;</sup></small> Shield/MonoGoggle XTR<small><sup>&#153;</sup></small>, Blue Frame, Clear Lens
Allsafe Shield/MonoGoggle XTR, Blue Frame, Clear Lens
Product Type Safety Goggles,Color Blue,Lens Clear/Anti-fog
EW-81657-18
In stock.
$22.00 USD/EACH
Qty:
Representative photo only U S SAFETY Safety Flex Goggles Indirect vent
U.S.SAFETY Safety-Flex Goggles, Indirect-vent
Product Type Safety Goggles,Color Green,Lens Clear
EW-81652-22
In stock.
$3.75 USD/EACH
Qty:
EW-86326-20 Uvex Strategy Safety Goggles with Fabric Band, Gray Body with Direct Vents, Clear Lens
EW-86326-20
In stock.
$7.25 USD/EACH
Qty:
EW-81652-20 U.S.SAFETY<small><sup>&reg;</sup></small> Safety-Flex Goggles, Unvented
U.S.SAFETY Safety-Flex Goggles, Unvented
Product Type Safety Goggles,Color Green,Lens Clear
EW-81652-20
In stock.
$3.50 USD/EACH
Qty:
Representative photo only Flex Seal Safety Goggle gray lens
Flex Seal Safety Goggle, gray lens
Product Type Safety Goggles,Color Gray,Lens Clear
EW-86527-02
In stock.
$20.00 USD/EACH
Qty:
17 results (showing 1 - 15 )
Per Page 
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title