Item Total = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
Category:
Brand:
Accuform (675)
Salisbury (128)
Product Type:
Gloves (487)
Signs (317)
Availability:
In Stock (705)
Shop Items By:
CE Compliance:
Yes (144)
Capacity (Liters):
1 to 5 (5)
10 to 50 (14)
Capacity (gal.):
1-5 (29)
10-20 (19)
Color:
White (94)
Blue (31)
Black (24)
Dewpoint:
Yes (1)
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 8 ext. 138-8456
 
Home  >  Safety Products

Safety Products

Choose from our comprehensive line of Safety Products products.
Air/Gas Analysis for Environmental Health and Safety
Alarms for Environmental Health and Safety
Apparel for Environmental Health and Safety
Electrical Safety for Environmental Health and Safety
Eyewash for Environmental Health and Safety
Eye protection and accessories for Environmental Health and Safety
Floor Mats for Environmental Health and Safety
Gloves for Environmental Health and Safety
Hearing Protection for Environmental Health and Safety
Heat Stress Monitors for Environmental Health and Safety
Light Meter for Environmental Health and Safety
Lockout/Tagout for Environmental Health and Safety
Personal Protection Equipment (PPE) for Environmental Health and Safety
Radiation Meters for Environmental Health and Safety
Respirators for Environmental Health and Safety
Signs and Labels for Environmental Health and Safety
Sound Meters for Environmental Health and Safety
Spill Control & Waste Disposal for Environmental Health and Safety
Storage Cabinets & Safety Cans for Environmental Health and Safety
 

Shop for occupational safety supplies and safety equipment, including personal protective equipment (PPE), equipment for hazardous materials and protection from electrical and fall hazards.

Buy More Save More on many Safety products

Choose the right glove for the chemicals you handle

Enhance workplace safety

 Product Detail
Sort
 Availability
 Price

Ansell TNT Blue Nitrile Gloves, Powder Free, Large

  • Product Type: Gloves
  • Material: Nitrile
  • Size: Large
EW-81601-06
In stock.
$15.00 USD/BOX

Qty:

Ansell 565717 TNT Blue Nitrile Gloves, Powder Free, Medium

  • Product Type: Gloves
  • Material: Nitrile
  • Size: Medium
EW-81601-05
In stock.
$15.00 USD/BOX
$12.00 USD/BOX

Qty:

Ansell TNT Blue Nitrile Gloves, Powder Free, Small

  • Product Type: Gloves
  • Material: Nitrile
  • Size: Small
EW-81601-04
In stock.
$15.00 USD/BOX
$12.00 USD/BOX

Qty:

MAPA Temp-Tec Heat-Insulated Neoprene Gloves, Large

  • Product Type: Gloves
  • Material: Neoprene
  • Size: Large
EW-40211-31
In stock.
$31.00 USD/PAIR

Qty:

25 Pound Solid PP Autoclavable Cable/Zip Ties, 8" L, Black; 1000/Pk

  • Product Type: Cable Ties
  • Bundle dia: 1.75 in
  • Min loop strength: 25 lb
EW-06830-95
In stock.
$181.00 USD/PACKAGE

Qty:

40 Pound Solid Nylon Cable/Zip Ties, 5.5" L, White; 1000/Pk

  • Product Type: Cable Ties
  • Bundle dia: 1.25 in (3.2 cm)
  • Min loop strength: 40 lb
EW-06830-54
In stock.
$37.50 USD/BAG of 1000

Qty:

Ansell TNT Blue Nitrile Gloves, Powder Free, X-Large

  • Product Type: Gloves
  • Material: Nitrile
  • Size: X-Large
EW-81601-07
In stock.
$15.00 USD/BOX
$12.00 USD/BOX

Qty:

Solid-design 4 in cable tie, white 1000/bag

  • Product Type: Cable Ties
  • Bundle dia: 0.9375 in
  • Min loop strength: 18 lb
EW-06830-52
In stock.
$25.75 USD/BAG of 1000

Qty:

50 Pound Solid Nylon Cable/Zip Ties, 7.5" L, White; 1000/Pk

  • Product Type: Cable Ties
  • Bundle dia: 1.75 in
  • Min loop strength: 50 lb
EW-06830-56
In stock.
$57.00 USD/BAG of 1000

Qty:

Digi-Sense Data Logging Light Meter with NIST Traceable Calibration

  • Product Type: Light Meters
  • Foot Candle Range: 0 to 40,000
  • LUX Range: 400/4000/40,000/400,000
EW-20250-00
In stock.
$192.00 USD/EACH

Qty:

Tedlar Sampling Bags With Combination Valve, 1L

  • Product Type: Sampling Bag
  • Material: PVF
  • Qty/cs: 10
EW-86561-00
In stock.
$117.00 USD/CASE

Qty:

11 Pound Solid PP Autoclavable Cable/Zip Ties, 4" L, White; 1000/Pk

  • Product Type: Cable Ties
  • Bundle dia: 0.75 in
  • Min loop strength: 11 lb
EW-06830-85
Usually ships in 5 days or sooner.
$102.00 USD/PACKAGE
$96.90 USD/PACKAGE

Qty:

Over-the-Ear Safety Face Mask, White Woven Polypropylene; 50/Pk

  • Product Type: Head & Face Covers
  • Description: Ear loop
  • Color: White
EW-86495-20
In stock.
$14.00 USD/PACKAGE

Qty:

Safety Eyewear Organizer, Acrylic, 12 Slots, Wall-Mount/Benchtop

  • Product Type: Glass Holder
  • Capacity: 12 pair
  • Dimensions: 12-1/4 " W
EW-86307-00
In stock.
$108.00 USD/EACH

Qty:

Ansell Thermaprene Heat-Resistant, Lined Neoprene Gloves, size 10, 26"L

  • Product Type: Gloves
  • Material: Neoprene
  • Length: 26 in
EW-81609-06
In stock.
$44.50 USD/PAIR

Qty:

3059 results (showing 1 - 15 )
Per Page 
#PRODUCTTITLE#

#PRODUCTHEADER#

#IAMGESTICKER#
#CROSSOUTPRICE#
#PRICE#
#PRICEWITHVAT#
#AVAILABILITY#
Qty
Quantity Discounts
Qty: #QTY10COUNT# #QTY15COUNT# #QTY20COUNT#
Price: #QTY10PRICE# #QTY15PRICE# #QTY20PRICE#
#PRODUCTIMAGE#
#POWERREVIEW#
#MARKETINGTEXT#
#ICONS#
#SPECS#
Home  >  Safety Products