log in or register
Max system pressure (bar):
8.6 (6)
Max flow rate (GPM):
50 (2)
60 (2)
32 (2)
115 (2)
55 (2)
110 (2)
80 (1)
125 (1)
145 (1)
Max head (ft):
29 (2)
46 (2)
15 (2)
65 (2)
73 (2)
92 (1)
38 (1)
115 (1)
138 (1)
105 (1)
Max flow rate (LPM):
227.1 (1)
435.3 (1)
208.2 (1)
Max head (m):
8.8 (1)
4.6 (1)
14.0 (1)
Leasing Available
Financing is avaliable for many of our products.
See Details
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
Home  >  Pumps > Centrifugal Pumps > Specialty Pumps - Sanitary / Petroleum / Utility > Sanitary Centrifugal Pumps, Heavy-Duty
Sanitary Centrifugal Pumps, Heavy-Duty
Sanitary Centrifugal Pumps, Heavy-Duty
  • Clean-in-place (CIP) 3A sanitary transfer pump
  • 316L stainless steel liquid end construction
  • Ideal for RO water and pharmaceutical manufacturing
  • Casing is twice as thick as the 76710 series of pumps
  • Designed for steam-in-place (SIP)
Piping changes are eliminated due to the interchangeable design that these economical pumps offer when compared to other pumps. Models are available with inlets up to 6" contact our Application Specialists for assistance in sizing the proper pump for your system.
 Product Detail
Sort
 Availability
 Price
Representative photo only Sanitary Centrifugal Pump Heavy Duty 32 GPM 1 HP Stainless Steel Motor 1800 RPM
Sanitary Centrifugal Pump, Heavy-Duty, 32 GPM, 1 HP, Stainless Steel Motor, 1800 RPM
Product Type Sanitary Centrifugal Pumps,Max flow rate GPM 32,Max flow rate LPM 121.1
EW-76713-05
Usually ships in 14 days.
$3,212.00 USD/EACH
Qty:
Representative photo only Sanitary Centrifugal Pump Heavy Duty 32 GPM 1 HP White Epoxy Motor 1800 RPM
Sanitary Centrifugal Pump, Heavy-Duty, 32 GPM, 1 HP, White Epoxy Motor, 1800 RPM
Product Type Sanitary Centrifugal Pumps,Max flow rate GPM 32,Max head ft 15
EW-76712-05
Usually ships in 14 days.
$2,859.00 USD/EACH
Qty:
Representative photo only Sanitary Centrifugal Pump Heavy Duty 60 GPM 1 HP White Epoxy Motor 1800 RPM
Sanitary Centrifugal Pump, Heavy-Duty, 60 GPM, 1 HP, White Epoxy Motor, 1800 RPM
Product Type Sanitary Centrifugal Pumps,Max flow rate GPM 60,Max head ft 29
EW-76712-25
Usually ships in 14 days.
$3,029.00 USD/EACH
Qty:
Representative photo only Sanitary Centrifugal Pump Heavy Duty 50 GPM 1 HP Stainless Steel Motor 3600 RPM
Sanitary Centrifugal Pump, Heavy-Duty, 50 GPM, 1 HP, Stainless Steel Motor, 3600 RPM
Product Type Sanitary Centrifugal Pumps,Max flow rate GPM 50,Max flow rate LPM 189.3
EW-76713-00
Usually ships in 14 days.
$3,187.00 USD/EACH
Qty:
Representative photo only Sanitary Centrifugal Pump Heavy Duty 60 GPM 1 HP Stainless Steel Motor 1800 RPM
Sanitary Centrifugal Pump, Heavy-Duty, 60 GPM, 1 HP, Stainless Steel Motor, 1800 RPM
Product Type Sanitary Centrifugal Pumps,Max flow rate GPM 60,Max flow rate LPM 227.1
EW-76713-25
Usually ships in 14 days.
$3,387.00 USD/EACH
Qty:
Representative photo only Sanitary Centrifugal Pump Heavy Duty 115 GPM 5 HP White Epoxy Motor 1800 RPM
Sanitary Centrifugal Pump, Heavy-Duty, 115 GPM, 5 HP, White Epoxy Motor, 1800 RPM
Product Type Sanitary Centrifugal Pumps,Max flow rate GPM 115,Max head ft 115
EW-76712-40
Usually ships in 14 days.
$3,680.00 USD/EACH
Qty:
Representative photo only Sanitary Centrifugal Pump Heavy Duty 55 GPM 1 5 HP White Epoxy Motor 3600 RPM
Sanitary Centrifugal Pump, Heavy-Duty, 55 GPM, 1.5 HP, White Epoxy Motor, 3600 RPM
Product Type Sanitary Centrifugal Pumps,Max flow rate GPM 55,Max head ft 65
EW-76712-10
Usually ships in 14 days.
$2,934.00 USD/EACH
Qty:
Representative photo only Sanitary Centrifugal Pump Heavy Duty 115 GPM 5 HP Stainless Steel Motor 1800 RPM
Sanitary Centrifugal Pump, Heavy-Duty, 115 GPM, 5 HP, Stainless Steel Motor, 1800 RPM
Product Type Sanitary Centrifugal Pumps,Max flow rate GPM 115,Max flow rate LPM 435.3
EW-76713-40
Usually ships in 14 days.
$3,960.00 USD/EACH
Qty:
Representative photo only Sanitary Centrifugal Pump Heavy Duty 110 GPM 3 HP White Epoxy Motor 3600 RPM
Sanitary Centrifugal Pump, Heavy-Duty, 110 GPM, 3 HP, White Epoxy Motor, 3600 RPM
Product Type Sanitary Centrifugal Pumps,Max flow rate GPM 110,Max head ft 73
EW-76712-30
Usually ships in 14 days.
$3,326.00 USD/EACH
Qty:
Representative photo only Sanitary Centrifugal Pump Heavy Duty 110 GPM 3 HP Stainless Steel Motor 3600 RPM
Sanitary Centrifugal Pump, Heavy-Duty, 110 GPM, 3 HP, Stainless Steel Motor, 3600 RPM
Product Type Sanitary Centrifugal Pumps,Max flow rate GPM 110,Max flow rate LPM 416.4
EW-76713-30
Usually ships in 14 days.
$3,691.00 USD/EACH
Qty:
Representative photo only Sanitary Centrifugal Pump Heavy Duty 145 GPM 10 HP White Epoxy Motor 3600 RPM
Sanitary Centrifugal Pump, Heavy-Duty 145 GPM, 10 HP, White Epoxy Motor, 3600 RPM
Product Type Sanitary Centrifugal Pumps,Max flow rate GPM 145,Max head ft 138
EW-76712-70
Usually ships in 14 days.
$4,489.00 USD/EACH
Qty:
EW-76712-00 Sanitary Centrifugal Pump, Heavy-Duty, 50 GPM, 1 HP, White Epoxy Motor, 3600 RPM
Sanitary Centrifugal Pump, Heavy-Duty, 50 GPM, 1 HP, White Epoxy Motor, 3600 RPM
Product Type Sanitary Centrifugal Pumps,Max flow rate GPM 50,Max head ft 46
EW-76712-00
Usually ships in 14 days.
$2,834.00 USD/EACH
Qty:
Representative photo only Sanitary Centrifugal Pump Heavy Duty 55 GPM 1 5 HP Stainless Steel Motor 3600 RPM
Sanitary Centrifugal Pump, Heavy-Duty, 55 GPM, 1.5 HP, Stainless Steel Motor, 3600 RPM
Product Type Sanitary Centrifugal Pumps,Max flow rate GPM 55,Max flow rate LPM 208.2
EW-76713-10
Usually ships in 14 days.
$3,341.00 USD/EACH
Qty:
Representative photo only Sanitary Centrifugal Pump Heavy Duty 80 GPM 1 5 HP White Epoxy Motor 1800 RPM
Sanitary Centrifugal Pump, Heavy-Duty, 80 GPM, 1.5 HP, White Epoxy Motor, 1800 RPM
Product Type Sanitary Centrifugal Pumps,Max flow rate GPM 80,Max head ft 38
EW-76712-55
Usually ships in 14 days.
$3,249.00 USD/EACH
Qty:
Representative photo only Sanitary Centrifugal Pump Heavy Duty 125 GPM 5 HP White Epoxy Motor 3600 RPM
Sanitary Centrifugal Pump, Heavy-Duty, 125 GPM, 5 HP, White Epoxy Motor, 3600 RPM
Product Type Sanitary Centrifugal Pumps,Max flow rate GPM 125,Max head ft 92
EW-76712-60
Usually ships in 14 days.
$3,890.00 USD/EACH
Qty:
15 results (showing 1 - 15 )
Per Page 
Home  >  Pumps > Centrifugal Pumps > Specialty Pumps - Sanitary / Petroleum / Utility > Sanitary Centrifugal Pumps, Heavy-Duty
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title