log in or register
Max pressure:
Max flow rate (LPM):
219.6 (4)
98.4 (4)
136.3 (4)
Leasing Available
Financing is avaliable for many of our products.
See Details
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
Home  >  Pumps > Positive Displacement Pumps > Rotary Pumps > Rotary Lobe Pumps > Sanitary Wash-Down Rotary Lobe Pumps
Sanitary Wash-Down Rotary Lobe Pumps
Sanitary Wash-Down Rotary Lobe Pumps
  • Low maintenance rotary lobe pumps conform to 3A sanitary standards
  • Front mounted shaft seals are fully accessible from the front of the pump for easy replacement without disturbing piping in the process line.
  • Bi-wing rotors require no timing adjustments and provide a high volumetric efficiency.
  • Rugged design incorporates heavy-duty shafts for high-pressure capability.
Pumps are approved to EHEDG (European Hygienic Equipment Design Group) and CIP/SIP standards. Remove pump casing for quick maintenance and cleaning. Reduced noise and cavitation provide for a safe and comfortable working environment. Risk of particles being shed into fluid stream is reduced with noncontacting rotors.
Mount pump heads either vertically or horizontally (must be mentioned at time of order which orientation is needed).
Pumps include NEMA 4 wash-down motors and NEMA 4 wash-down variable-speed controllers. Variable-speed operation is digital and via a keypad mounted to the pump.
 Product Detail
Sort
 Availability
 Price
Representative photo only Sanitary Wash Down Rotary Lobe Pump 13 GPM 230 VAC Single phase
Sanitary Wash-Down Rotary Lobe Pump, 13 GPM, 230 VAC, Single-phase
Product Type Rotary Lobe Pumps,Max flow rate LPM 49.2,Max pressure 220 psi
EW-75201-02
Call 1-888-358-4705 for availability.
$8,030.00 USD/EACH
Qty:
Representative photo only Sanitary Wash down Rotary Lobe Pump 26 GPM 460 VAC Three phase
Sanitary Wash-down Rotary Lobe Pump, 26 GPM, 460 VAC, Three-phase
Product Type Rotary Lobe Pumps,Max flow rate LPM 98.4,Max pressure 115 psi
EW-75201-17
Usually ships in 14 days.
$9,174.00 USD/EACH
Qty:
Representative photo only Sanitary Wash down Rotary Lobe Pump 26 GPM 230 VAC Three phase
Sanitary Wash-down Rotary Lobe Pump, 26 GPM, 230 VAC, Three-phase
Product Type Rotary Lobe Pumps,Max flow rate LPM 98.4,Max pressure 115 psi
EW-75201-15
Usually ships in 14 days.
$9,174.00 USD/EACH
Qty:
Representative photo only Sanitary Wash down Rotary Lobe Pump 58 GPM 115 VAC Single phase
Sanitary Wash-down Rotary Lobe Pump, 58 GPM, 115 VAC, Single-phase
Product Type Rotary Lobe Pumps,Max flow rate LPM 219.6,Max pressure 115 psi
EW-75201-30
Usually ships in 14 days.
$12,398.00 USD/EACH
Qty:
EW-75201-00 Sanitary Wash-Down Rotary Lobe Pump, 13 GPM, 115VAC, Single-phase
Sanitary Wash-Down Rotary Lobe Pump, 13 GPM, 115VAC, Single-phase
Product Type Rotary Lobe Pumps,Max flow rate LPM 49.2,Max pressure 220 psi
EW-75201-00
Usually ships in 11 days.
$8,072.00 USD/EACH
Qty:
EW-75201-20 Analog controller-no digital due to motor
Sanitary Wash-down Rotary Lobe Pump, 36 Gpm, 115 Vac, Single-phase (no Digital Controller Due To 2 Hp 1 pHase Motor)
Product Type Rotary Lobe Pumps,Max flow rate LPM 136.3,Max pressure 225 psi
EW-75201-20
Usually ships in 14 days.
$10,648.00 USD/EACH
Qty:
Representative photo only Sanitary Wash down Rotary Lobe Pump 58 GPM 460 VAC Three phase
Sanitary Wash-down Rotary Lobe Pump, 58 GPM, 460 VAC, Three-phase
Product Type Rotary Lobe Pumps,Max flow rate LPM 219.6,Max pressure 115 psi
EW-75201-37
Usually ships in 14 days.
$11,726.00 USD/EACH
Qty:
Representative photo only Sanitary Wash down Rotary Lobe Pump 58 GPM 230 VAC Single phase
Sanitary Wash-down Rotary Lobe Pump, 58 GPM, 230 VAC, Single-phase
Product Type Rotary Lobe Pumps,Max flow rate LPM 219.6,Max pressure 115 psi
EW-75201-32
Usually ships in 14 days.
$12,398.00 USD/EACH
Qty:
Representative photo only Sanitary Wash down Rotary Lobe Pump 58 GPM 230 VAC Three phase
Sanitary Wash-down Rotary Lobe Pump, 58 GPM, 230 VAC, Three-phase
Product Type Rotary Lobe Pumps,Max flow rate LPM 219.6,Max pressure 115 psi
EW-75201-35
Usually ships in 14 days.
$11,726.00 USD/EACH
Qty:
Representative photo only Sanitary Wash down Rotary Lobe Pump 36 GPM 230 VAC Three phase
Sanitary Wash-down Rotary Lobe Pump, 36 GPM, 230 VAC, Three-phase
Product Type Rotary Lobe Pumps,Max flow rate LPM 136.3,Max pressure 225 psi
EW-75201-25
Usually ships in 14 days.
$10,424.00 USD/EACH
Qty:
Representative photo only Sanitary Wash down Rotary Lobe Pump 36 GPM 460 VAC Three phase
Sanitary Wash-down Rotary Lobe Pump, 36 GPM, 460 VAC, Three-phase
Product Type Rotary Lobe Pumps,Max flow rate LPM 136.3,Max pressure 225 psi
EW-75201-27
Usually ships in 14 days.
$10,424.00 USD/EACH
Qty:
Representative photo only Sanitary Wash down Rotary Lobe Pump 13 GPM 460 VAC Three phase
Sanitary Wash-down Rotary Lobe Pump, 13 GPM, 460 VAC, Three-phase
Product Type Rotary Lobe Pumps,Max flow rate LPM 49.2,Max pressure 220 psi
EW-75201-07
Usually ships in 14 days.
$7,903.00 USD/EACH
Qty:
Representative photo only Sanitary Wash down Rotary Lobe Pump 26 GPM 115 VAC Single phase
Sanitary Wash-down Rotary Lobe Pump, 26 GPM, 115 VAC, Single-phase
Product Type Rotary Lobe Pumps,Max flow rate LPM 98.4,Max pressure 115 psi
EW-75201-10
Usually ships in 14 days.
$9,401.00 USD/EACH
Qty:
Representative photo only Sanitary Wash down Rotary Lobe Pump 26 GPM 230 VAC Single phase
Sanitary Wash-down Rotary Lobe Pump, 26 GPM, 230 VAC, Single-phase
Product Type Rotary Lobe Pumps,Max flow rate LPM 98.4,Max pressure 115 psi
EW-75201-12
Usually ships in 14 days.
$9,401.00 USD/EACH
Qty:
Representative photo only Sanitary Wash down Rotary Lobe Pump 36 GPM 230 VAC Single phase
Sanitary Wash-down Rotary Lobe Pump, 36 GPM, 230 VAC, Single-phase
Product Type Rotary Lobe Pumps,Max flow rate LPM 136.3,Max pressure 225 psi
EW-75201-22
Usually ships in 14 days.
$10,648.00 USD/EACH
Qty:
16 results (showing 1 - 15 )
Per Page 
Home  >  Pumps > Positive Displacement Pumps > Rotary Pumps > Rotary Lobe Pumps > Sanitary Wash-Down Rotary Lobe Pumps
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title