Home  >  Fluid & Liquid Handling > Piping > Schedule 40 PVC Piping
Schedule 40 PVC Piping
Schedule 40 PVC Piping
  • PVC and CPVC piping will not rust or corrode
  • Easy to install
  • Offers significantly lower installation costs compared to metal piping systems
PVC is generally inert to most mineral acids, bases, salts and paraffinic hydrocarbon solutions, and can handle fluids at service temperatures up to 140° F (60°C), allowing a wide range of process applications, including corrosive fluids.
 Product Detail
Sort
 Availability
 Price
Representative photo only Schedule 40 PVC piping 5 5
Schedule 40 PVC piping, 5", 5'
Product Type Piping,Wall thickness in. 0.258,ID in. 5.017,Material PVC
EW-33312-44
Usually ships in 5 days.
$43.50 USD/EACH
Qty:
Representative photo only Schedule 40 PVC piping 1 4 5
Schedule 40 PVC piping, 1/4", 5'
Product Type Piping,Wall thickness in. 0.088,ID in. 0.354,Material PVC
EW-33312-00
Usually ships in 14 days.
$6.35 USD/EACH
Qty:
Representative photo only Schedule 40 PVC piping 2 5 5
Schedule 40 PVC piping, 2.5", 5'
Product Type Piping,Wall thickness in. 0.203,ID in. 2.445,Material PVC
EW-33312-32
Usually ships in 13 days.
$17.50 USD/EACH
Qty:
Representative photo only Schedule 40 PVC piping 1 25 5
Schedule 40 PVC piping, 1.25", 5'
Product Type Piping,Wall thickness in. 0.14,ID in. 1.364,Material PVC
EW-33312-20
Usually ships in 5 days.
$5.55 USD/EACH
Qty:
Representative photo only Schedule 40 PVC piping 3 4 5
Schedule 40 PVC piping, 3/4", 5'
Product Type Piping,Wall thickness in. 0.113,ID in. 0.81,Material PVC
EW-33312-12
Usually ships in 4 days.
$4.40 USD/EACH
Qty:
Representative photo only Schedule 40 PVC piping 4 5
Schedule 40 PVC piping, 4", 5'
Product Type Piping,Wall thickness in. 0.237,ID in. 3.998,Material PVC
EW-33312-40
Usually ships in 6 days.
$25.00 USD/EACH
Qty:
Representative photo only Schedule 40 PVC piping 1 5 5
Schedule 40 PVC piping, 1.5", 5'
Product Type Piping,Wall thickness in. 0.145,ID in. 1.592,Material PVC
EW-33312-24
Usually ships in 3 days.
$6.35 USD/EACH
Qty:
Representative photo only Schedule 40 PVC piping 6 5
Schedule 40 PVC piping, 6", 5'
Product Type Piping,Wall thickness in. 0.28,ID in. 6.031,Material PVC
EW-33312-48
Usually ships in 6 days.
$45.50 USD/EACH
Qty:
Representative photo only Schedule 40 PVC piping 6 9
Schedule 40 PVC piping, 6", 9'
Product Type Piping,Wall thickness in. 0.28,ID in. 6.031,Material PVC
EW-33312-50
Call 1-888-358-4705 for availability.
$89.50 USD/EACH
Qty:
Representative photo only Schedule 40 PVC piping 2 5 9
Schedule 40 PVC piping, 2.5", 9'
Product Type Piping,Wall thickness in. 0.203,ID in. 2.445,Material PVC
EW-33312-34
Usually ships in 3 days.
$31.50 USD/EACH
Qty:
Representative photo only Schedule 40 PVC piping 3 9
Schedule 40 PVC piping, 3", 9'
Product Type Piping,Wall thickness in. 0.216,ID in. 3.042,Material PVC
EW-33312-38
Usually ships in 3 days.
$33.50 USD/EACH
Qty:
Representative photo only Schedule 40 PVC piping 1 2 5
Schedule 40 PVC piping, 1/2", 5'
Product Type Piping,Wall thickness in. 0.109,ID in. 0.608,Material PVC
EW-33312-08
Call 1-888-358-4705 for availability.
$19.00 USD/EACH
Qty:
Representative photo only Schedule 40 PVC piping 5 9
Schedule 40 PVC piping, 5", 9'
Product Type Piping,Wall thickness in. 0.258,ID in. 5.017,Material PVC
EW-33312-46
Call 1-888-358-4705 for availability.
$86.50 USD/EACH
Qty:
Representative photo only Schedule 40 PVC piping 2 5
Schedule 40 PVC piping, 2", 5'
Product Type Piping,Wall thickness in. 0.154,ID in. 2.049,Material PVC
EW-33312-28
Usually ships in 2 days.
$8.55 USD/EACH
Qty:
Representative photo only Schedule 40 PVC piping 4 9
Schedule 40 PVC piping, 4", 9'
Product Type Piping,Wall thickness in. 0.237,ID in. 3.998,Material PVC
EW-33312-42
Usually ships in 3 days.
$49.00 USD/EACH
Qty:
17 results (showing 1 - 15 )
Per Page 
Home  >  Fluid & Liquid Handling > Piping > Schedule 40 PVC Piping
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title