Subtotal = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
Leasing Available
Financing is avaliable for many of our products.

See Details
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
 
Home  >  Lab Equipment > Titrators > Volumetric and Potentiometric > Schott Automatic Volumetric Titrator

Schott Automatic Volumetric Titrator

Schott Automatic Volumetric Titrator

  • Automatically titrate to your endpoints using a keypad for digital control
  • Eliminates the difficult task of reading a burette and judging color changes at endpoints
  • Automatic titrators provide added convenience over manual methods
Autotitrator stops at your preselected pH endpoint or equivalence point every time. There’s no need to worry about overshooting your endpoint. Use the keypad to program pre-titration volume and speed.

Use the autotitrator with any combination pH electrode. The pH buffer solutions are stored in memory to make calibration easy. Access the eight pH buffers—2.00, 4.00, 4.01, 6.87, 7.00, 9.18, 10.00, and 12.45—through the keypad.

For high-accuracy dosing, the glass cylinder features an automatic valve system. The integrated system fills without prompting for easy dosing. For incremental dosing operations, adjust the volume
and waiting time between volume increments.

Connect FEP tubing (24906-60) and bottletop adapter (24906-55) to 1-L reagent reservoir (24906-52) and titrator to add reagent. Order reservoir, adapter, and tubing separately.
 Product Detail
Sort
 Availability
 Price
EW-24906-25 Schott<small><sup>&reg;</sup></small> automatic volumetric titrator, 20-mL capacity, 230 VAC

Schott automatic volumetric titrator, 20-mL capacity, 230 VAC

Product Type Volumetric Titrators,Power VAC 230,Output RS-232
EW-24906-25
Call 1-888-358-4705 for availability.
$5,500.00 USD/EACH

Qty:

Representative photo only Schott automatic volumetric titrator 20 mL capacity 115 VAC

Schott automatic volumetric titrator, 20-mL capacity, 115 VAC

Product Type Volumetric Titrators,Power VAC 115,Output RS-232
EW-24906-20
Call 1-888-358-4705 for availability.
$3,950.00 USD/EACH

Qty:

2 results (showing 1 - 2 )
Per Page 
Home  >  Lab Equipment > Titrators > Volumetric and Potentiometric > Schott Automatic Volumetric Titrator

View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title