United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Leasing Available
Financing is avaliable for many of our products.
See Details
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Home
>
Cooling Equipment
>
Ice Machines
>
Scotsman Cubed-and Flaked-Ice Machines
> Scotsman Flaked Ice Makers
Scotsman Flaked Ice Makers
Self-contained units eliminate the need to order a separate ice storage bin
Compact size allows convenient placement under standard 40"H laboratory benches
Slide-away door provides convenient and easy access to ice
These self-contained units provide maximum efficiency and performance under continuous operating conditions. Choose either air- or water-cooled condenser units depending on your laboratory room conditions. Removable front cabinetry facilitates easy access for maintenance and replacement of air filters, and water pump. Included 6" legs raise the ice maker off the floor for easy cleaning or leveling on uneven floors.
Flaked-Ice Makers produce up to 395 lbs of flaked ice per day and store 140 lbs in the self-contained bin. Flaked ice quickly conforms to your container or sample tubes for greater cooling capabilities than other ice forms and is perfect for short-term packing and shipping of laboratory samples.
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Compare
Product Detail
Sort
Most Popular
Name
Name Descending
Price (Lowest First)
Price (Highest First)
Availability
Price
Flaked-ice maker with air-cooled condenser, 395 Lbs/day production, 115 VAC
Product Type Flaked-Ice Makers,Description Self-Contained Flaker,Capacity lbs 80
EW-55100-03
Usually ships in 5 days.
$5,460.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Scotsman Flaked Ice Makers
Flaked-ice maker with water-cooled condenser, 395 Lbs/day production, 115 VAC
Product Type Flaked-Ice Makers,Description Self-Contained Flaker,Capacity lbs 80
EW-55100-04
Usually ships in 13 days.
$5,460.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Scotsman Flaked Ice Makers
2
results (showing
1
-
2
)
Per Page
5
10
15
25
50
Home
>
Cooling Equipment
>
Ice Machines
>
Scotsman Cubed-and Flaked-Ice Machines
> Scotsman Flaked Ice Makers
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title