Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Home
>
Laboratory Cleaning Supplies
>
Biosafety Disposal
> Sharps Containers
Sharps Containers
Proper disposal of sharps containers is important
Containers for contaminated sharps must be puncture-resistant. The sides and the bottom must be leakproof. They must be appropriately labeled or color-coded red to warn everyone that the contents are hazardous. Containers for disposable sharps must be closable (that is, have a lid, flap, door, or other means of closing the container), and they must be kept upright to keep the sharps and any liquids from spilling out of the container. The containers must be replaced routinely and not be overfilled, which can increase the risk of needle sticks or cuts. Sharps disposal containers that are reusable must not be opened, emptied, or cleaned manually or in any other manner that would expose workers to the risk of sharps injury. Employers also must ensure that reusable sharps that are contaminated are not stored or processed in a manner that requires workers to reach by hand into the containers where these sharps have been placed.
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Compare
Product Detail
Sort
Most Popular
Name
Name Descending
Price (Lowest First)
Price (Highest First)
Availability
Price
B-D Polypropylene Sharps Collectors, 5.4 qt, 12/case
Product Type Sharps Collector,Material Polypropylene,Capacity Liters 5.1
EW-07940-75
Usually ships in 13 days.
$90.00 USD/CASE
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
BD Sharps Collectors, Polypropylene
B-D Polypropylene Sharps Collectors, 2 gal., 10/case
Product Type Sharps Collector,Material Polypropylene,Capacity Liters 7.6
EW-07940-76
In stock.
$94.00 USD/CASE of 10
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
BD Sharps Collectors, Polypropylene
2
results (showing
1
-
2
)
Per Page
5
10
15
25
50
Home
>
Laboratory Cleaning Supplies
>
Biosafety Disposal
> Sharps Containers
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity