Item Total = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
Maximum magnification:
20x (1)
40x (1)
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 8 ext. 138-8456
Shop Microscopes
Shop Microscopes
Once the only microscope, monocular microscopes are looked upon as quaint now. A monocular microscope is made for just one eye. While larger more sophisticated microscopes have moved into the laboratory experience the monocular microscope still has its uses. Portable, easy-to-use and durable a monocular microscope, such as the Cole-Parmer Shop Microscopes, can be excellent for field work, inspections and routine or unexpected applications. With these instruments you just need batteries. Just turn them on, no calibration is necessary, and begin working. Most models are cast in an aluminum frame, a small size, and rugged carrying case. Select from four models, choosing the magnification that meets your needs. Swivel-mount penlight illuminator ensures proper lighting when needed. Take measurements with the built-in reticle, showing inch and millimeter scales. If you need help selecting the right monocular microscope speak with a Cole-Parmer Technical Expert today.
 Product Detail
Sort
 Availability
 Price
Cole-Parmer Shop Microscope; 40x magnification
  • Product Type: Specialty Microscopes
  • Magnification: 40x
  • Maximum magnification: 40x
EW-03890-20
Usually ships in 12 days.
$196.00 USD/EACH
Qty:
Cole-Parmer Shop Microscope; 20x magnification
  • Product Type: Specialty Microscopes
  • Magnification: 20x
  • Maximum magnification: 20x
EW-03890-10
Usually ships in 8 days.
$196.00 USD/EACH
Qty:
2 results (showing 1 - 2 )
Per Page 
#PRODUCTTITLE#
#PRODUCTHEADER#
#IAMGESTICKER#
#CROSSOUTPRICE#
#PRICE#
#PRICEWITHVAT#
#AVAILABILITY#
Qty
Quantity Discounts
Qty: #QTY10COUNT# #QTY15COUNT# #QTY20COUNT#
Price: #QTY10PRICE# #QTY15PRICE# #QTY20PRICE#
#PRODUCTIMAGE#
#POWERREVIEW#
#MARKETINGTEXT#
#ICONS#
#SPECS#