Home  >  Material Testing & Physical Testing > Surface Testing > Hardness Testers > Shore D Model of Digital Hardness Tester
Shore D Model of Digital Hardness Tester
Shore D Model of Digital Hardness Tester
  • Conform to ASTM D2240, DIN 53505, ISO R/868, and JIS K7215 Shore scale hardness testing standards
These hardness testers are available in low-cost analog or digital versions. Simply position tester on sample and apply constant pressure. Choose an economical analog model or a digital model with 3 1/2-digit LCD.
Shore D models 59790-02 and -06 have a 30° sharp-point penetrator for testing hard materials such as hard rubber, acrylic, glass, and dense wood.
Each hardness tester includes reference test block, instructions, and hard carrying case.
Order bench stand 59790-50 separately to increase the accuracy and repeatability of your tests.
 Product Detail
Sort
 Availability
 Price
Representative photo only Shore D digital hardness tester
Shore D digital hardness tester
Product Type Durometer,Dimensions " W 3.625,Dimensions " H 7.125
EW-59790-06
In stock.
$1,933.00 USD/EACH
Qty:
1 results (showing 1 - 1 )
Per Page 
Home  >  Material Testing & Physical Testing > Surface Testing > Hardness Testers > Shore D Model of Digital Hardness Tester
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title