United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Home
>
Pumps
>
Metering Pumps
>
Piston Pumps
>
Series Q Valveless Piston Pump Systems
> Single-Head, Variable-Speed AC Drives with Separate Speed Controller
Single-Head, Variable-Speed AC Drives with Separate Speed Controller
Interchangeable with all pump heads on facing page
Set flow anywhere from 0 to 100% of maximum flow rate—drives offer better than ±1% accuracy
Rated for continuous duty
Drives with explosion proof motors are designed for applications in Class I, Group D, Division 1 hazardous areas
For additional speed control, choose drives that have a separate speed controller for precise and repeatable setting of stroke rates from 0.1 to 99.9%. Controllers accept 4 to 20 mA input for remote control or a digital potentiometer for manual control. Pedestal drive (model 07104-40) is for chain-, belt-, or shaft-coupling to rotating machine elements or to your motor.
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Compare
Product Detail
Sort
Most Popular
Name
Name Descending
Price (Lowest First)
Price (Highest First)
Availability
Price
Series Q single head, variable-speed AC drive with controller, 1800 spm maximum, 110 VAC
Product Type Piston Metering Pumps,Max pressure 100 psi,Motor hp 1/27
EW-07104-32
Usually ships in 7 days.
$2,185.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Single-Head, Variable-Speed AC Drives with Separate Speed Controller
1
results (showing
1
-
1
)
Per Page
5
10
15
25
50
Home
>
Pumps
>
Metering Pumps
>
Piston Pumps
>
Series Q Valveless Piston Pump Systems
> Single-Head, Variable-Speed AC Drives with Separate Speed Controller
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title