United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Price Range:
$100 to $150
(1)
$150 to $200
(1)
$200 to $250
(1)
Availability:
In Stock
(2)
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Home
>
Lab Equipment
>
Extraction Apparatus
>
Soxhlet Extraction Systems
> Soxhlet Extractor Body
Soxhlet Extractor Body
Improved design prevents the trapping of liquid
Use these Soxhlet extraction components to extract soluble components from a solid sample into an organic solvent. Place a sample in the thimble and heat the solvent contained in the flask. As the solvent vapors rise (passing through the extraction thimble), they enter the water-cooled condensor, and liquify. When the liquid level in the extractor reaches the top of the siphon tube, siphoning action returns the extract-enriched solvent to the flask.
Soxhlet-type extraction is used for continuous extraction of solids with a suitable solvent. The improved design includes a protected siphon tube, an enlarged vapor tube, and sloping seals to prevent the trapping of liquid. TS joints connect the condenser to the extractor and the extractor to the flask. Uses paper, alundum, or extraction thimbles (order separately). Purchase Soxhlet glassware as a complete assembly or as individual components.<br>
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Compare
Product Detail
Sort
Most Popular
Name
Name Descending
Price (Lowest First)
Price (Highest First)
Availability
Price
250 ml Soxhlet extractor body
EW-99037-40
In stock.
$185.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Soxhlet Extractor Body
300 ml Soxhlet extractor body
EW-99037-42
Call 1-888-358-4705 for availability.
$203.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Soxhlet Extractor Body
125 ml Soxhlet extractor body
EW-99037-38
Usually ships in 14 days.
$148.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Soxhlet Extractor Body
3
results (showing
1
-
3
)
Per Page
5
10
15
25
50
Home
>
Lab Equipment
>
Extraction Apparatus
>
Soxhlet Extraction Systems
> Soxhlet Extractor Body
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title