Subtotal = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
Leasing Available
Financing is avaliable for many of our products.

See Details
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
 

Specialty spectrophotometers

Specialty spectrophotometers

Specialty spectrometers allow you to deliver quick and accurate measurements in the field. Some of these specialty products include an oil content analyzer and cold-vapor mercury analyzer. These specialty spectrometers are geared towards environmental testing- particularly in the water industry for drinking, surface, saline and waste waters. Choose these products when surveying water quality and hazardous wastewater monitoring is of concern. Cole-Parmer offers you a wide selection of spectroscopy equipment and would like to be your solution for offering unique and hard-to-find products. It is our specialty! If you cannot find the product you need or are looking for assistance, please contact us. Selecting the right equipment for your needs can be difficult. If you need help choosing the specialty spectrometers contact a Cole-Parmer Technical Expert today.
 Product Detail
Sort
 Availability
 Price
Representative photo only Solvent extractant S 316 1 5 kg for Horiba Oil Content Analyzer

Solvent extractant S-316, 1.5 kg, for Horiba Oil Content Analyzer

Product Type Petroleum Testing Spectroscopes,Model 100690,Brand Horiba
EW-82900-10
In stock.
$420.00 USD/EACH

Qty:

EW-82900-00 Horiba Oil Content Analyzer, 115 V

Horiba Oil Content Analyzer, 115 V

Product Type Petroleum Testing Spectroscopes,Output RS-232C and centronics printer port,Display LCD
EW-82900-00
Usually ships in 7 days.
$7,000.00 USD/EACH

Qty:

Representative photo only Horiba Oil Content Analyzer 230 V

Horiba Oil Content Analyzer, 230 V

Product Type Petroleum Testing Spectroscopes,Output RS-232C and centronics printer port,Display LCD
EW-82900-05
Usually ships in 12 days.
$7,000.00 USD/EACH

Qty:

Representative photo only Solvent reclaimer SR 300 for recycling Horiba Oil Content Analyzer extractant

Solvent reclaimer SR-300, for recycling Horiba Oil Content Analyzer extractant

Product Type Petroleum Testing Spectroscopes,Model SR-305,Brand Horiba
EW-82900-20
Usually ships in 12 days.
$1,855.00 USD/EACH

Qty:

EW-28750-25 Cold Vapor Mercury Analyzer, 115 V

Cold Vapor Mercury Analyzer, 115 V

Product Type Specialty Spectroscopes,Output RS-232, analog, parallel port,Model 410
EW-28750-25
Call 1-888-358-4705 for availability.
$10,350.00 USD/EACH

Qty:

Representative photo only Cold Vapor Mercury Analyzer 230 V

Cold Vapor Mercury Analyzer, 230 V

Product Type Specialty Spectroscopes,Output RS-232, analog, parallel port,Model 410-220V
EW-28750-28
Call 1-888-358-4705 for availability.
$10,350.00 USD/EACH

Qty:

6 results (showing 1 - 6 )
Per Page 

View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title