log in or register
Price Range:
Availability:
Flask size (mL):
250 (2)
500 (2)
1000 (2)
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
Spherical Heating Mantles
Spherical Heating Mantles
New Spherical heating mantles cover both top and bottom of one-, two-,or three-neck spherical flasks. Mantles are made from flexible fabrics for effective heating and protection of glass vessels. Wrap around zipper holds the mantle halves snugly together to prevent heat loss. Top split allow mantle removal without disturbing the glass vessels. Maximum heating temperature is 842°F (450°C). Mantles include detachable 4-ft, two wire-cord with locking connector. Order a power or temperature controller from the accessories.
 Product Detail
Sort
 Availability
 Price
Representative photo only Spherical heating mantle for 1000 ml flask 380 watts lower half 140 watts upper half 230 VAC
Spherical heating mantle for 1000-ml flask, 380 watts lower half, 140 watts upper half, 230 VAC
Product Type Heating Mantle,Flask size mL 1000,Max temperature º F 842
EW-36226-25
In stock.
$597.00 USD/EACH
Qty:
Representative photo only Spherical heating mantle for 5000 ml flask 600 watts lower half 300 watts upper half 115 VAC
Spherical heating mantle for 5000-ml flask, 600 watts lower half, 300 watts upper half, 115 VAC
Product Type Heating Mantle,Flask size mL 5000,Max temperature º F 842
EW-36226-50
Usually ships in 12 days.
$758.00 USD/EACH
Qty:
Representative photo only Spherical heating mantle for 3000 ml flask 500 watts lower half 200 watts upper half 115 VAC
Spherical heating mantle for 3000-ml flask, 500 watts lower half, 200 watts upper half, 115 VAC
Product Type Heating Mantle,Flask size mL 3000,Max temperature º F 842
EW-36226-40
Usually ships in 14 days.
$638.00 USD/EACH
Qty:
Representative photo only Spherical heating mantle for 2000 ml flask 500 watts lower half 200 watts upper half 230 VAC
Spherical heating mantle for 2000-ml flask, 500 watts lower half, 200 watts upper half, 230 VAC
Product Type Heating Mantle,Flask size mL 2000,Max temperature º F 842
EW-36226-35
Usually ships in 14 days.
$622.00 USD/EACH
Qty:
Representative photo only Spherical heating mantle for 1000 ml flask 380 watts lower half 140 watts upper half 115 VAC
Spherical heating mantle for 1000-ml flask, 380 watts lower half, 140 watts upper half, 115 VAC
Product Type Heating Mantle,Flask size mL 1000,Max temperature º F 842
EW-36226-20
Usually ships in 11 days.
$580.00 USD/EACH
Qty:
Representative photo only Spherical heating mantle for 2000 ml flask 500 watts lower half 200 watts upper half 115 VAC
Spherical heating mantle for 2000-ml flask, 500 watts lower half, 200 watts upper half, 115 VAC
Product Type Heating Mantle,Flask size mL 2000,Max temperature º F 842
EW-36226-30
Usually ships in 8 days.
$612.00 USD/EACH
Qty:
EW-36226-00 Spherical heating mantle for 250-ml flask, 180 watts lower half, 0 watts upper half, 115 VAC
Spherical heating mantle for 250-ml flask, 180 watts lower half, 0 watts upper half, 115 VAC
Product Type Heating Mantle,Flask size mL 250,Max temperature º F 842
EW-36226-00
Call 1-888-358-4705 for availability.
$304.00 USD/EACH
Qty:
Representative photo only Spherical heating mantle for 500 ml flask 270 watts lower half 0 watts upper half 230 VAC
Spherical heating mantle for 500-ml flask, 270 watts lower half, 0 watts upper half, 230 VAC
Product Type Heating Mantle,Flask size mL 500,Max temperature º F 842
EW-36226-15
Call 1-888-358-4705 for availability.
$360.00 USD/EACH
Qty:
Representative photo only Spherical heating mantle for 250 ml flask 180 watts lower half 0 watts upper half 230 VAC
Spherical heating mantle for 250-ml flask, 180 watts lower half, 0 watts upper half, 230 VAC
Product Type Heating Mantle,Flask size mL 250,Max temperature º F 842
EW-36226-05
Usually ships in 12 days.
$319.00 USD/EACH
Qty:
Representative photo only Spherical heating mantle for 3000 ml flask 500 watts lower half 200 watts upper half 230 VAC
Spherical heating mantle for 3000-ml flask, 500 watts lower half, 200 watts upper half, 230 VAC
Product Type Heating Mantle,Flask size mL 3000,Max temperature º F 842
EW-36226-45
Usually ships in 13 days.
$747.00 USD/EACH
Qty:
Representative photo only Spherical heating mantle for 5000 ml flask 600 watts lower half 300 watts upper half 230 VAC
Spherical heating mantle for 5000-ml flask, 600 watts lower half, 300 watts upper half, 230 VAC
Product Type Heating Mantle,Flask size mL 5000,Max temperature º F 842
EW-36226-55
Usually ships in 14 days.
$758.00 USD/EACH
Qty:
Representative photo only Spherical heating mantle for 500 ml flask 270 watts lower half 0 watts upper half 115 VAC
Spherical heating mantle for 500-ml flask, 270 watts lower half, 0 watts upper half, 115 VAC
Product Type Heating Mantle,Flask size mL 500,Max temperature º F 842
EW-36226-10
Usually ships in 13 days.
$350.00 USD/EACH
Qty:
12 results (showing 1 - 12 )
Per Page 
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title