log in or register
Capacity (cu ft):
Leasing Available
Financing is avaliable for many of our products.
See Details
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
Home  >  Cooling Equipment > Freezers > StableTemp Lab Freezers
StableTemp Lab Freezers
StableTemp Lab Freezers
  • Providing reliability and security essential for the protection of sensitive lab samples
  • Quiet refrigeration system keeps the noise level down
  • Range of sizes to suit your lab needs
  • Available with or without chart recorders
Our line of general-purpose freezers is ideal for the demanding requirements of daily lab applications from -12 to -30°C (10.4 to -22°F). These freezers deliver the quality, security, and performance that you need with basic features such as a digital temperature controller with intuitive interface, fluorine-free high-density insulation, and automatic defrost. Standard security features include keyed power switch, high/low temperature alarm, easily accessible contacts for backup monitoring, door ajar alarm, and a control panel with key lock.
Quality painted steel exterior, anodized aluminum interior and solid stainless steel door make these freezers durable and easy to clean. Easily move, level, and stabilize with the included locking casters. The 1" rear access point makes it simple to monitor a sample with an external probe.
 Product Detail
Sort
 Availability
 Price
EW-44261-50 StableTemp freezer, single solid door model
StableTemp 24 cu ft freezer stainless steel exterior w/alarms, access port, solid door and casters; 115V
Product Type General Purpose Freezer,Capacity cu ft 24,Capacity Liters 679
EW-44261-50
Usually ships in 13 days.
$7,680.00 USD/EACH
Qty:
EW-44261-53 StableTemp freezer, single solid door model
EW-44261-53
Usually ships in 13 days.
$10,240.00 USD/EACH
Qty:
EW-44261-54 StableTemp freezer, single solid door model
StableTemp 48 cu ft freezer stainless steel exterior w/alarms, access port, dual solid doors and casters; 208-230V
Product Type General Purpose Freezer,Capacity cu ft 48,Capacity Liters 1359
EW-44261-54
Usually ships in 13 days.
$9,090.00 USD/EACH
Qty:
EW-44261-55 StableTemp freezer, single solid door model
EW-44261-55
Usually ships in 13 days.
$10,125.00 USD/EACH
Qty:
EW-44261-52 StableTemp freezer, single solid door model
StableTemp 34 cu ft freezer stainless steel exterior w/alarms, access port, solid door and casters; 115V
Product Type General Purpose Freezer,Capacity cu ft 34,Capacity Liters 962
EW-44261-52
Usually ships in 13 days.
$9,310.00 USD/EACH
Qty:
EW-44261-51 StableTemp freezer, single solid door model
EW-44261-51
Usually ships in 13 days.
$8,610.00 USD/EACH
Qty:
6 results (showing 1 - 6 )
Per Page 
Home  >  Cooling Equipment > Freezers > StableTemp Lab Freezers
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title