Home  >  Process Equipment & Instrumentation > Steam Handling
Steam Handling
Steam Handling
Stainless steel steam traps are an important accessory to any lab that wants to prevent condensation. Steam traps keep steam space free of condensate to provide maximum system efficiency and are available in thermodynamic, thermostatic, and inverted bucket configurations. Steam trap materials of construction include stainless steel or cast iron with 1/2" to 1" NPT connections and 1/2" and 3/4" Tri-clamp connections. Y-strainers are installed in-line in vertical or horizontal pipelines for added protection against dirt and pipe scale. Included 20 mesh screens remove pipeline debris and protect pipeline components such as valves, pumps, and heat transfer equipment in steam systems. Y-strainers are available in bronze or cast iron with 1/4" to 2" NPT connections. Air eliminators improve the circulation of pressurized liquids by eliminating air and other non-condensable gases; mount at high points to provide immediate removal of air and gases. Cast-iron air eliminators feature stainless steel internal components and are available with 3/4" and 1" NPT connections. If you have a question about stainless steel stream traps or any other equipment contact a Technical Expert.
 Product Detail
Sort
 Availability
 Price
Representative photo only Thermo Dynamic steam trap 3 4 NPT F
Thermo-Dynamic steam trap, 3/4"NPT(F)
Product Type Steam handling fittings,Material Stainless Steel,Max temperature 800F (427C)
EW-07600-06
Usually ships in 7 days.
$444.00 USD/EACH
Qty:
EW-07600-92 Air eliminator; 3/4" NPT(F) x 3/8" NPT(F)
Air eliminator; 3/4" NPT(F) x 3/8" NPT(F)
Product Type Steam Handling fittings,Material Cast iron,Max temperature º F 450
EW-07600-92
Usually ships in 6 days.
$208.00 USD/EACH
Qty:
EW-07600-30 Cast iron float and thermostatic steam trap (parallel configuration); 3/4" NPT(F), 15 psig
Cast iron float and thermostatic steam trap (parallel configuration); 3/4" NPT(F), 15 psig
Product Type Steam handling fittings,Material Cast iron,Max temperature 45F (25C) of superheat
EW-07600-30
Usually ships in 7 days.
$252.00 USD/EACH
Qty:
Representative photo only Air eliminator 1 NPT F x 3 8 NPT F
Air eliminator; 1" NPT(F) x 3/8" NPT(F)
Product Type Steam Handling fittings,Material Cast iron,Max temperature º F 450
EW-07600-94
Usually ships in 7 days.
$176.00 USD/EACH
Qty:
Representative photo only Cast iron Y strainer 3 4 NPT F
Cast iron "Y" strainer, 3/4" NPT(F)
Product Type Steam handling fittings,Material Cast iron,Max temperature 406F (208C)
EW-07600-72
Usually ships in 10 days.
$38.00 USD/EACH
Qty:
Representative photo only Cast iron inverted bucket steam trap 3 4 NPT F 30 psig
Cast iron inverted bucket steam trap; 3/4" NPT(F), 30 psig
Product Type Steam Handling fittings,Material Cast iron,Max temperature º F 450
EW-07600-64
Usually ships in 7 days.
$134.50 USD/EACH
Qty:
Representative photo only Bronze Y strainer 3 4 NPT F
Bronze "Y" strainer, 3/4" NPT(F)
Product Type Steam handling fittings,Material Bronze,Max temperature 406F (208C)
EW-07600-88
Usually ships in 7 days.
$115.00 USD/EACH
Qty:
Representative photo only Cast iron float and thermostatic steam trap parallel configuration 3 4 NPT F 125 psig
Cast iron float and thermostatic steam trap (parallel configuration); 3/4" NPT(F), 125 psig
Product Type Steam handling fittings,Material Cast iron,Max temperature 45F (25C) of superheat
EW-07600-36
Usually ships in 7 days.
$418.00 USD/EACH
Qty:
EW-07600-38 Cast iron float and thermostatic steam trap (in-line horizontal configuration); 1/2" NPT(F), 15 psig
Cast iron float and thermostatic steam trap (in-line horizontal configuration); 1/2" NPT(F), 15 psig
Product Type Steam handling fittings,Material Cast iron,Max temperature 45F (25C) of superheat
EW-07600-38
Usually ships in 11 days.
$224.00 USD/EACH
Qty:
Representative photo only Cast iron float and thermostatic steam trap in line horizontal configuration 3 4 NPT F 15 psig
Cast iron float and thermostatic steam trap (in-line horizontal configuration); 3/4" NPT(F), 15 psig
Product Type Steam handling fittings,Material Cast iron,Max temperature 45F (25C) of superheat
EW-07600-46
Usually ships in 7 days.
$261.00 USD/EACH
Qty:
Representative photo only ASCO Red Hat steam and hot water valve 1 2 300F
ASCO Red-Hat steam and hot water valve, 1/2", 300°F
Product Type Solenoid Electric Valves (actuated),Cv 4,Max temperature º F 210
EW-01340-80
Usually ships in 8 days.
$151.00 USD/EACH
Qty:
Representative photo only Thermo Dynamic steam trap 1 NPT F
Thermo-Dynamic steam trap, 1"NPT(F)
Product Type Steam handling fittings,Material Stainless Steel,Max temperature 800F (427C)
EW-07600-08
Usually ships in 8 days.
$611.00 USD/EACH
Qty:
EW-07600-02 Thermo-Dynamic steam trap, 3/8"NPT(F)
Thermo-Dynamic steam trap, 3/8"NPT(F)
Product Type Steam handling fittings,Material Stainless Steel,Max temperature 800F (427C)
EW-07600-02
Usually ships in 10 days.
$249.00 USD/EACH
Qty:
Representative photo only Cast iron inverted bucket steam trap 3 4 NPT F 75 psig
Cast iron inverted bucket steam trap; 3/4" NPT(F), 75 psig
Product Type Steam Handling fittings,Material Cast iron,Max temperature º F 450
EW-07600-66
Usually ships in 7 days.
$126.00 USD/EACH
Qty:
Representative photo only Bronze Y strainer 3 8 NPT F
Bronze "Y" strainer, 3/8" NPT(F)
Product Type Steam handling fittings,Material Bronze,Max temperature 406F (208C)
EW-07600-84
Usually ships in 7 days.
$61.00 USD/EACH
Qty:
36 results (showing 1 - 15 )
Per Page 
Home  >  Process Equipment & Instrumentation > Steam Handling
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title