Subtotal = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
Price Range:
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
 
Home  >  Laboratory Supplies & Consumables > Bottles > Sample Bottles > Sterilized Sampling Bottle Assemblies

Sterilized Sampling Bottle Assemblies

Sterilized Sampling Bottle Assemblies

  • Safely transfer, sample, and store biopharmaceutical products and reagents in applications from research to process
  • Secure, single-piece cap/tubing assembly eliminates contamination and product loss due to loose connections
  • Assemblies are pre-sterilized by gamma irradiation—eliminates labor and validation costs associated with assembly and cleaning
Cap assembly includes a seamless liner with premolded C-FLEX® fill and vent tubes to create an unobstructed fluid path. The 18" fill tube is plugged at the terminal end to maintain assembly sterility until use. Vent tube includes a 0.2-micron Pall® Acrodisc PTFE vent filter to prevent infiltration by contaminants. Vent filter is hydrophobic to guard against loss of costly product or media during filling or emptying. The one-piece liner eliminates the risk of bacterial entrapment and contamination and forms an elastomeric seal between cap and bottle rim.

C-FLEX Tubing is manufactured according to cGMP and meets USP Class VI and USDA requirements. Tubing can withstand temperatures from –100 to 275°F (–73 to 135°C). C-Flex tubing has exceptional tensile and tear strength, extremely low extractables, and excellent biocompatibility. The smooth inner surface reduces absorption and protein binding.
 Product Detail
Sort
 Availability
 Price
Representative photo only Sterilized Sampling Bottle Assembly 2 L

Sterilized Sampling Bottle Assembly, 2 L

Product Type Sterilized Sampling Bottle Assemblies
EW-95644-08
In stock.
$145.00 USD/EACH

Qty:

Representative photo only Sterilized Sampling Bottle Assembly 500 mL

Sterilized Sampling Bottle Assembly, 500 mL

Product Type Sterilized Sampling Bottle Assemblies
EW-95644-04
In stock.
$61.00 USD/EACH

Qty:

Representative photo only Sterilized Sampling Bottle Assembly 1 L

Sterilized Sampling Bottle Assembly, 1 L

Product Type Sterilized Sampling Bottle Assemblies
EW-95644-06
In stock.
$68.00 USD/EACH

Qty:

Representative photo only Sterilized Sampling Bottle Assembly 250 mL

Sterilized Sampling Bottle Assembly, 250 mL

Product Type Sterilized Sampling Bottle Assemblies
EW-95644-02
In stock.
$61.00 USD/EACH

Qty:

EW-95644-00 Sterilized Sampling Bottle Assembly, 125 mL

Sterilized Sampling Bottle Assembly, 125 mL

Product Type Sterilized Sampling Bottle Assemblies
EW-95644-00
In stock.
$58.00 USD/EACH

Qty:

5 results (showing 1 - 5 )
Per Page 
Home  >  Laboratory Supplies & Consumables > Bottles > Sample Bottles > Sterilized Sampling Bottle Assemblies

View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title