United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Category:
Scienceware Stirbars
(42)
Mono-Mold Standard Stir Bars
(14)
Egg-Shaped Stir Bars
(8)
Thermo Scientific Stirring Bars
(7)
Slim-Line Stir Bars
(6)
Micro Stir Bars
(5)
Starburst Stirring Heads
(5)
PTFE-Coated Disposable Stir Bars
(4)
Glass-Encased Stir Bars
(4)
Double Stir Bars
(3)
Color-Coded Stir Bars, PTFE
(2)
Stir Bar Retrievers
(2)
Large-Volume Stir Bars
(2)
Cole-Parmer Mono-Mold Stir Bar Kits
(2)
Semi-Circular Stir Bar
(1)
Brand:
Mono-Mold
(20)
Scienceware
(17)
Cole-Parmer
(7)
[+]
Show more
(1)
Thermo Scientific
(7)
[-]
Show less
Product Type:
Stir bars
(65)
Price Range:
$0 to $100
(98)
$100 to $299
(9)
Availability:
In Stock
(54)
Learn more:
Equipping a Laboratory: A Study in Customer Service Excellence
[-]
Show Less
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Home
>
Laboratory Supplies & Consumables
> Stir Bars
Stir Bars
Choose from our comprehensive line of Magnetic Stir Bars and other Stirrers products.
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Compare
Product Detail
Sort
Most Popular
Name
Name Descending
Price (Lowest First)
Price (Highest First)
Availability
Price
Mono-Mold Standard Stir Bar with Spinning Ring, 3"L x 1/2" dia
Product Type Stir bars,Brand Mono-Mold
EW-04612-30
In stock.
$21.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Mono-Mold
Standard Stir Bars
MONO-MOLD Standard Stir Bar with Spinning Ring, 1"L x 3/8" dia
Product Type Stir bars,Brand Mono-Mold
EW-08550-00
In stock.
$6.25 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Mono-Mold
Standard Stir Bars
MONO-MOLD Standard Stir Bar with Spinning Ring, 2"L x 3/8" dia
Product Type Stir bars,Brand Mono-Mold
EW-08552-00
In stock.
$10.25 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Mono-Mold
Standard Stir Bars
Large-Volume Stir Bar, 6-1/4"L x 1-1/16"dia
Product Type Stir bars
EW-04627-10
In stock.
$142.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Large-Volume Stir Bars
Mono-Mold Slim-Line Stir Bar with Spinning Ring, 2"L x 5/16" dia
Product Type Stir bars,Brand Mono-Mold
EW-04775-65
In stock.
$7.75 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Mono-Mold
Slim-Line
Stir Bars
Cole-Parmer 3" Egg-Shaped Stir Bar
Product Type Stir bars,Diameter 3/4",Brand Cole-Parmer
EW-04768-85
In stock.
$29.50 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
EGG-SHAPED STIR BARS
Cole-Parmer Mono-Mold With Spinning Ring Stir Bar Kit, 12 bars
Product Type Stir bars,Brand Cole-Parmer,Model F37182-5001
EW-04775-99
In stock.
$86.00 USD/BOX of 12
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Cole-Parmer
Mono-Mold Stir Bar Kits
MONO-MOLD Standard Stir Bar, 2"L x 3/8" dia
Product Type Stir bars,Brand Mono-Mold
EW-04770-60
In stock.
$9.75 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Mono-Mold
Standard Stir Bars
MONO-MOLD Standard Stir Bar with Spinning Ring, 1-1/2"L x 3/8" dia
Product Type Stir bars,Brand Mono-Mold
EW-08551-00
In stock.
$7.60 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Mono-Mold
Standard Stir Bars
Cole-Parmer Mono-Mold Without Spinning Ring Stir Bar Kit, 18 bars
Product Type Stir bars,Brand Cole-Parmer,Model F 37184-5000
EW-04775-98
In stock.
$87.00 USD/BOX of 18
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Cole-Parmer
Mono-Mold Stir Bar Kits
Micro Stir Bar, 10mm L x 3mm dia
Product Type Stir bars
EW-08546-00
In stock.
$7.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Micro Stir Bars
Mono-Mold Slim-Line Stir Bar with Spinning Ring, 1"L x 5/16" dia
Product Type Stir bars,Brand Mono-Mold
EW-04775-45
In stock.
$5.25 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Mono-Mold
Slim-Line
Stir Bars
Large-Volume Stir Bar, 4-1/2"L x 1-1/16"dia
Product Type Stir bars
EW-04627-00
In stock.
$114.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Large-Volume Stir Bars
1/2" X 1/8" Disposable Stir Bars
Product Type Stir bars,Model 80037-1065
EW-04769-02
In stock.
$147.00 USD/PACKAGE
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
PTFE-Coated Disposable Stir Bars
Cole-Parmer 2 1/2" Egg-Shaped Stir Bar
Product Type Stir bars,Diameter 3/4",Brand Cole-Parmer
EW-04768-75
In stock.
$25.75 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
EGG-SHAPED STIR BARS
107
results (showing
1
-
15
)
1
2
3
4
5
6
Next >
>>
Per Page
5
10
15
25
50
Home
>
Laboratory Supplies & Consumables
> Stir Bars
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title