Subtotal = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register

Learn more about
Safety Can and Safety Storage Cabinet

2,600 pages
of answers

FREE
2013/14 Cole-Parmer
Sourcebook
Our safety can, safety cabinet and safety storage cabinet selection covers over 8 pages and contains options that are OSHA compliant and FM approved from leading brands such as Justrite and Eagle.
Request your copy today.

Leasing Available
Financing is avaliable for many of our products.

See Details
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
 

Storage and Handling

Storage and Handling

Cole-Parmer carries a wide selection of safety storage and handling options, including safety cans, safety cabinets, and safety storage cabinets. Many of the products we offer will help you comply with OSHA requirements and are FM approved. Look for these designations on the individual products.

Safety Cabinets

safety storage cabinet

Safety cabinets provide a convenient and secure means of storage for flammables, corrosives, acids, paints, and ink. To properly assess your storage needs, you must determine the quantity and class of the chemicals you need to house, and ascertain whether they have any special storage requirements. Refer to the Material Safety Data Sheet or container label for the specific requirements of each chemical. Additional guidelines on storing hazardous materials are available from your local fire marshall or OSHA office. These regulations will help determine the size and type of storage cabinet you require.

Proper segregation of stored chemicals is necessary to prevent incompatible materials from inadvertently coming into contact, in which case fire, explosion, violent reactions, or toxic gases could result. A physical barrier and/or distance, as provided by safety cabinets, is effective for proper segregation. You must also consider the compatibility of the chemical with the cabinet. For example, strong acids and caustics will corrode untreated, metal cabinets. It is best to store these chemicals in non-metallic wood or HDPE cabinets, or in epoxy-coated cabinets designed to prevent rapid deterioration caused by corrosive fluids or vapors.

Safety cabinets serve several critical functions, including:

  • Reducing the risk of fire posed by hazardous chemicals, protecting both personnel and facilities
  • Helping you identify, organize, and segregate dangerous liquids
  • Providing compliance with federal OSHA regulations and state and local fire codes
  • Improving efficiency by allowing solvents to be stored near points of use
  • Improving security with keyed locking mechanisms

Safety Cans

safety can

Safety cans are designed to permit safe handling, storage, transport, or disposal of flammable liquids or waste by controlling any flammable vapors that may be present.

Safety Requirements

To meet OSHA requirements, a safety storage can must:

  • Have a flame arrester
  • Be leakproof
  • Automatically vent vapor
  • Self-close after filling or pouring
  • Have a maximum containment volume of 5 gal (18.9L)

In addition, any portable safety can containing flammable liquids with a flash point at or below 26.7C (80F) must also be emblazoned with a yellow band or stenciled identification marker.

Need help in selecting the best safety can or safety cabinet for your requirements? Let our highly trained Application Specialists provide you with additional product specifications, guide you through safety cabinet selection and maintenance, and offer support throughout the life of the safety cabinet you purchase. They are thoroughly schooled on the products we sell and can answer the most challenging questions.

Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
 Product Detail
Sort
 Availability
 Price
EW-81781-10 SciMatCo<small><sup>&reg;</sup></small> Flammable Storage Cabinet, 4 gallon capacity, manual-latching door

SciMatCo Flammable Storage Cabinet, 4 gallon capacity, manual-latching door

Product Type Flammable Storage Cabinets,Description Benchtop,Capacity gal. 4 x 1,Color Yellow
EW-81781-10
Usually ships in 7 days.
$469.00 USD/EACH

Qty:

EW-81780-15 SciMatCo<small><sup>&reg;</sup></small> Polypropylene Acid Storage Cabinet, 6 X 2.5 L bottle capacity

SciMatCo Polypropylene Acid Storage Cabinet, 6 X 2.5 L bottle capacity

Product Type Acid Storage Cabinets,Capacity gal. 4,Color White
EW-81780-15
Usually ships in 4 days.
$505.00 USD/EACH

Qty:

EW-81791-44 81791-45 shown

Justrite Safety Can for Flammables, Type II, 1 gallon

Product Type Safety Cans,Capacity Liters 4,Capacity gal. 1
EW-81791-44
Call 1-888-358-4705 for availability.
$90.00 USD/EACH

Qty:

EW-81791-43 81791-42 shown with optional funnel 81791-50

Justrite Safety Can for Flammables, Type I, 5 gallon

Product Type Safety Cans,Capacity Liters 18.9,Capacity gal. 5
EW-81791-43
Call 1-888-358-4705 for availability.
$48.00 USD/EACH

Qty:

Representative photo only SciMatCo Full Size Acid Storage Cabinet twenty four 2 5 L bottle capacity

SciMatCo Full-Size Acid Storage Cabinet, twenty four 2.5 L bottle capacity

Product Type Acid Storage Cabinets,Description Full-size,Capacity gal. 24 x 2.5 L,Color Blue
EW-81780-00
Usually ships in 5 days.
$871.00 USD/EACH

Qty:

EW-09419-05 Space Saver Acid Storage Cabinet, Manual-Latching Door, 12 Gallon

Eagle Space Saver Acid Storage Cabinet, Manual-Latching Door, 12 Gallon

Product Type Acid Storage Cabinets,Description Space saver,Capacity gal. 12,Color Blue
EW-09419-05
Usually ships in 7 days.
$667.00 USD/EACH

Qty:

EW-86446-20 Vented Storage Cabinet, 200L Double, Organic Filter

Vented Storage Cabinet, 200L Double, Organic Filter

Product Type Chemical Storage Cabinets,Description Double w/ organic vapor filter,Capacity gal. 53,Color White
EW-86446-20
Call 1-888-358-4705 for availability.
$5,619.00 USD/EACH

Qty:

Representative photo only Eagle Acid Storage Cabinet Self Closing Door 60 Gallon

Eagle Acid Storage Cabinet, Self-Closing Door, 60 Gallon

Product Type Acid Storage Cabinets,Capacity gal. 60,Color Blue
EW-09419-01
Usually ships in 6 days.
$1,581.00 USD/EACH

Qty:

Representative photo only Justrite HDPE Benchtop Acid Storage Cabinet 8 L

Justrite HDPE Benchtop Acid Storage Cabinet, 8 L

Product Type Acid Storage Cabinets,Description Benchtop,Capacity gal. 2,Color Blue
EW-81790-00
Usually ships in 4 days.
$444.00 USD/EACH

Qty:

Representative photo only SciMatCo Stackable Cabinet Acid

SciMatCo Stackable Cabinet, Acid

Product Type Acid Storage Cabinets,Description Acid,Capacity gal. 22 x 2.5 L,Color Blue
EW-81781-00
Usually ships in 4 days.
$936.00 USD/EACH

Qty:

EW-81773-00 Acid And Corrosive Cabinets, 4 Gal, Blue

Eagle PE Acid And Corrosive Cabinets, 4 Gal, Blue

Product Type Acid Storage Cabinets,Description Benchtop,Capacity gal. 4,Color Blue
EW-81773-00
In stock.
$515.00 USD/EACH

Qty:

Representative photo only Eagle Flammable Safety Cabinet Manual latching 4 gallon

Eagle Flammable Safety Cabinet, Manual-latching, 4 gallon

Product Type Flammable Storage Cabinets,Capacity gal. 4,Color Yellow
EW-09418-21
Usually ships in 7 days.
$502.00 USD/EACH

Qty:

EW-81400-14 Thermo Scientific Nalgene Fluorinated HDPE Safety Waste System

Thermo Scientific Nalgene Fluorinated HDPE Safety Waste System

Product Type Safety Cans,Capacity Liters 10,Capacity gal. 2.6
EW-81400-14
In stock.
$149.00 USD/EACH

Qty:

EW-81790-42 Model 81790-41 shown

Justrite Flammable Storage Safety Cabinet, 60 gallons, sure-grip handle

Product Type Flammable Storage Cabinets,Capacity gal. 60,Color Yellow
EW-81790-42
Usually ships in 4 days.
$1,190.00 USD/EACH

Qty:

Representative photo only SciMatCo Benchtop Acid Storage Cabinet five 2 5 L bottle capacity

SciMatCo Benchtop Acid Storage Cabinet, five 2.5 L bottle capacity

Product Type Acid Storage Cabinets,Description Benchtop,Capacity gal. 5 x 2.5 L,Color Blue
EW-81780-05
Usually ships in 4 days.
$580.00 USD/EACH

Qty:

160 results (showing 1 - 15 )
Per Page 

View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title