Item Total = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
Category:
Product Type:
Price Range:
Capacity (kg):
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 8 ext. 138-8456
 
Home  >  Storage and Utility Cabinets

Storage and Utility Cabinets

Storage and Utility Cabinets
Choose from our comprehensive line of Storage and Utility Cabinets and other Material Handling products.
 Product Detail
Sort
 Availability
 Price

Storage Cabinet, 2 Door, Plexiglass Windows; 36"W x 78" H x 24"D

  • Product Type: Storage Cabinets
  • Dimensions: 36 " W
  • Dimensions: 78 " H
EW-47546-62
Available for order
or call 1-888-358-4705 to order
$961.00 USD/EACH

Qty:

Wall-Mount Cabinet, 2 Door, Plexiglass Windows; 30"W x 26" H x 12"D

  • Product Type: Storage Cabinets
  • Dimensions: 30 " W
  • Dimensions: 26 " H
EW-47546-64
Usually ships in 15 days.
$499.00 USD/EACH

Qty:

Mobile Laboratory Bench, 3-Drawer/1-Door, 45" x 22", 230 lbs Cap

  • Product Type: Utility Cart
  • Dimensions: 45 in W x 22 in D x 34 in H
  • Dimensions: 45 " W
EW-47546-66
Usually ships in 11 days.
$1,057.00 USD/EACH

Qty:

3 results (showing 1 - 3 )
Per Page 
#PRODUCTTITLE#

#PRODUCTHEADER#

#IAMGESTICKER#
#CROSSOUTPRICE#
#PRICE#
#PRICEWITHVAT#
#AVAILABILITY#
Qty
Quantity Discounts
Qty: #QTY10COUNT# #QTY15COUNT# #QTY20COUNT#
Price: #QTY10PRICE# #QTY15PRICE# #QTY20PRICE#
#PRODUCTIMAGE#
#POWERREVIEW#
#MARKETINGTEXT#
#ICONS#
#SPECS#
Home  >  Storage and Utility Cabinets