Home  >  Fluid & Liquid Handling > Syringes
Syringes
Syringes
Syringes have been around for centuries. From ancient India to modern Rome, from the hospitals of ninth century Egypt to the late nineteenth century laboratories of Ireland,syringes have advanced along with man. Today, the array of syringes available is dizzying. In most cases the syringe and needle are sold separately, allowing for customization of the complete set. There are disposable, single-use products available in sterilized and nonsterilized versions. The Hamilton Gastight High-Performance Micro and Macro syringes are ideal for chromatography applications, as they are corrosive resistant and are gas- and liquid-tight, up to 200 psi. The Hamilton Microliter syringe is low-volume with needle is ideal for accurate dispensing of fluids at minute volumes. The Hamilton standard syringes are appropriate for a large range of applications. Cadence, formerly Popper, has a syringe that is available in large volumes, with capacities up to 250 mL. Cadence standard ones are also available in a variety of tip types to meet your connection needs. Needle types also can be chosen to meet specific requirements. The Micro-Emulsifying needles are ideal for mixing two liquids with different viscosity. If you have questions about syringes or needles, why not speak with one of our Technical Experts.
 Product Detail
Sort
 Availability
 Price
Representative photo only 60 mL B D plastic disposable syringe luer lock tip 40 box
60 mL B-D plastic disposable syringe, luer-lock tip, 40 / box
Material Polycarbonate,Product Type Chromatography syringe,Capacity mL 60
EW-07940-30
Usually ships in 15 days.
$54.00 USD/BOX
Qty:
EW-07940-12 B-D<small><sup>&#153;</sup></small> Disposable Syringes, Luer-Lock Tips, 10 mL
B-D Disposable Syringes, Luer-Lock Tips, 10 mL
Material Polycarbonate,Product Type Chromatography syringe,Capacity mL 10
EW-07940-12
Usually ships in 15 days.
$26.00 USD/BOX of 100
Qty:
EW-07940-00 B-D<small><sup>&#153;</sup></small> Disposable Syringes, Luer-Lock Tips, 1 mL
B-D Disposable Syringes, Luer-Lock Tips, 1 mL
Material Polycarbonate,Product Type Chromatography syringe,Capacity mL 1
EW-07940-00
Call 1-888-358-4705 for availability.
$66.00 USD/BOX of 100
Qty:
Representative photo only B D Disposable Syringes Luer Lock Tips 3 mL
B-D Disposable Syringes, Luer-Lock Tips, 3 mL
Material Polycarbonate,Product Type Chromatography syringe,Capacity mL 3
EW-07940-02
Call 1-888-358-4705 for availability.
$31.00 USD/BOX of 100
Qty:
Representative photo only BD bulk syringes non sterile clean 60mL Luer Lok tip 125 case
BD bulk syringes, non-sterile clean, 60mL Luer-Lok tip, 125/case
Material Polycarbonate,Product Type Syringe,Capacity mL 60
EW-07945-28
In stock.
$140.00 USD/CASE
Qty:
Representative photo only 60 mL B D plastic disposable syringe slip tip 40 box
60 mL B-D plastic disposable syringe, slip tip, 40 / box
Material Polycarbonate,Product Type Chromatography syringe,Capacity mL 60
EW-07940-28
Call 1-888-358-4705 for availability.
$51.00 USD/BOX
Qty:
Representative photo only BD bulk syringes non sterile clean 20mL Luer Lok tip 325 case
BD bulk syringes, non-sterile clean, 20mL Luer-Lok tip, 325/case
Material Polycarbonate,Product Type Syringe,Capacity mL 20
EW-07945-16
Usually ships in 14 days or sooner.
$190.00 USD/CASE
Qty:
Representative photo only BD bulk syringes non sterile clean 5mL Luer Lok tip 1400 case
BD bulk syringes, non-sterile clean, 5mL Luer-Lok tip, 1400/case
Material Polycarbonate,Product Type Syringe,Capacity mL 5
EW-07945-06
In stock.
$225.00 USD/CASE
Qty:
Representative photo only BD bulk syringes non sterile clean 3mL Luer Lok tip 1600 case
BD bulk syringes, non-sterile clean, 3mL Luer-Lok tip, 1600/case
Material Polycarbonate,Product Type Syringe,Capacity mL 3
EW-07945-02
In stock.
$150.00 USD/CASE
Qty:
Representative photo only B D Disposable Syringes Slip Tips 10 mL
B-D Disposable Syringes, Slip Tips, 10 mL
Material Polycarbonate,Product Type Chromatography syringe,Capacity mL 10
EW-07940-14
Call 1-888-358-4705 for availability.
$26.00 USD/BOX of 100
Qty:
Representative photo only B D Disposable Syringes Luer Lock Tips 5 mL
B-D Disposable Syringes, Luer-Lock Tips, 5 mL
Material Polycarbonate,Product Type Chromatography syringe,Capacity mL 5
EW-07940-06
Usually ships in 15 days.
$29.00 USD/BOX of 100
Qty:
Representative photo only BD bulk syringes non sterile clean 10mL Luer Lok tip 850 case
BD bulk syringes, non-sterile clean, 10mL Luer-Lok tip, 850/case
Material Polycarbonate,Product Type Syringe,Capacity mL 10
EW-07945-12
In stock.
$170.00 USD/CASE
Qty:
Representative photo only Cadence Science Jumbo Syringe syringe capacity 50mL
Cadence Science® Jumbo Syringe; syringe capacity; 50mL
Material Glass,Product Type Jumbo Syringes,Capacity mL 50
EW-25701-00
Usually ships in 6 days.
$105.00 USD/EACH
Qty:
Representative photo only BD bulk syringes non sterile clean 60mL Slip Tip 125 case
BD bulk syringes, non-sterile clean, 60mL Slip-Tip, 125/case
Material Polycarbonate,Product Type Syringe,Capacity mL 60
EW-07945-30
In stock.
$140.00 USD/CASE
Qty:
Representative photo only Syringes with PTFE luer lock syringe capacity 50 mL needle gauge 22
Syringes with PTFE luer lock; syringe capacity; 50 mL; needle gauge; 22
Material Glass,Product Type Chromatography syringe,Capacity mL 50
EW-07939-86
In stock.
$205.00 USD/EACH
Qty:
140 results (showing 1 - 15 )
Per Page 
Home  >  Fluid & Liquid Handling > Syringes
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title