log in or register
Availability:
Max sample size (mL):
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
Tapered microtips
Tapered microtips
Microtips can process small samples at very high intensity. The tapered microtip screws in the 1/2" (13 mm) threaded end probe in place of the replaceable tip. The stepped microtip assembly consists of two parts—a microtip and a coupler. The stepped microtip assembly can process volumes as small as 250 µL.
 Product Detail
Sort
 Availability
 Price
Representative photo only Tapered microtips 1 4
Tapered microtips; 1/4"
Product Type Ultrasonic Processor Probes,Description Tapered microtips; 1/4",Max sample size mL 25
EW-04710-46
In stock.
$272.50 USD/EACH
Qty:
EW-04710-42 Tapered microtips; 1/8"
Tapered microtips; 1/8"
Product Type Ultrasonic Processor Probes,Description Tapered microtips; 1/8",Max sample size mL 10
EW-04710-42
In stock.
$272.50 USD/EACH
Qty:
Representative photo only Tapered microtips 3 16
Tapered microtips; 3/16"
Product Type Ultrasonic Processor Probes,Description Tapered microtips; 3/16",Max sample size mL 15
EW-04710-44
Call 1-888-358-4705 for availability.
$272.50 USD/EACH
Qty:
3 results (showing 1 - 3 )
Per Page 
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title