United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Home
>
Calibration
>
Calibration Equipment
>
Pressure Calibrators
> Taylor Low Pressure Calibrators
Taylor Low Pressure Calibrators
Small size allows you to test the output pressure of a pneumatic instrument and make calibration adjustments at the same time
This calibrator allows a technician to test the output or differential pressure (flow) readings of a pneumatic instrument with one hand allowing the other hand to make calibration adjustments on the instrument being tested. The calibrator weighs only 8 oz. as compared to 3 or more lbs. for equipment marketed in kit form or one made up by the technician from parts.
The calibrator consists of sphygmomanometer-type (blood pressure) gauge accurate to 1% with a squeeze bulb for applying pressure manually and a precision-machined hand-adjusted valve to hold and relieve the calibrating pressure. A flexible hose and a 3/16" tube-1/4" NPT compression fitting is supplied for quick and easy connection to unit being tested.
CALIBRATES
Pneumatic controllers
Low pressure gauges
Pneumatic transmitters
Flow transmitters
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Compare
Product Detail
Sort
Most Popular
Name
Name Descending
Price (Lowest First)
Price (Highest First)
Availability
Price
44S390 : Pressure Calibrator, 0 to 18 psig with Case
Product Type Pressure/Vacuum Calibrators,Brand Taylor,Model 44S390
EW-68454-75
In stock.
$309.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Taylor Low Pressure Calibrators
44S369 : Pressure Calibrator, 0 to 160" H2O with Case
Product Type Pressure/Vacuum Calibrators,Brand Taylor,Model 44S369
EW-68454-74
In stock.
$309.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Taylor Low Pressure Calibrators
2
results (showing
1
-
2
)
Per Page
5
10
15
25
50
Home
>
Calibration
>
Calibration Equipment
>
Pressure Calibrators
> Taylor Low Pressure Calibrators
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title