United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Price Range:
$100 to $299
(2)
$800 to $1,099
(1)
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Home
>
Cooling Equipment
>
Cryogenics
>
Cryogenic Storage Dewars
>
Taylor Wharton Cryogenic Storage Dewars
> Taylor-Wharton Roller Base for Cryogenic Storage Dewars
Taylor-Wharton Roller Base for Cryogenic Storage Dewars
Easily move storage Dewars and Dewars with canister storage systems. Sturdy cast aluminum frame has five wheels with ball-bearing swivel casters. Use model 03773-65 for Dewars with 15.6" diameter base; use 03773-71 for Dewars with 18.8" diameter base. Shpg wt 8.0 lb (3.7 kg).
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Compare
Product Detail
Sort
Most Popular
Name
Name Descending
Price (Lowest First)
Price (Highest First)
Availability
Price
Roller base for Dewars with 15-5/8 in. dia base
Product Type Dewars,Brand Taylor-Wharton,Model R018-8C00
EW-03773-65
In stock.
$223.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Taylor-Wharton Roller Base for Cryogenic Storage Dewars
Taylor Wharton Storage Dewars Tipping Stand
Product Type Dewars,Brand Taylor-Wharton,Model D025-8C00
EW-03773-70
In stock.
$910.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Taylor-Wharton Roller Base for Cryogenic Storage Dewars
Roller base for Dewars with 18.8
Product Type Dewars,Brand Taylor-Wharton,Model R033-8C00
EW-03773-71
In stock.
$226.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Taylor-Wharton Roller Base for Cryogenic Storage Dewars
3
results (showing
1
-
3
)
Per Page
5
10
15
25
50
Home
>
Cooling Equipment
>
Cryogenics
>
Cryogenic Storage Dewars
>
Taylor Wharton Cryogenic Storage Dewars
> Taylor-Wharton Roller Base for Cryogenic Storage Dewars
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title