Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Price Range:
$1,100 to $1,999
(1)
$2,000 and Over
(3)
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Home
>
Life Science
>
PCR (Polymerase Chain Reaction)
>
Thermal Cyclers
>
Techne Prime Large-Format Thermal Cyclers
> Techne Prime Large-Format Thermal Cyclers Accessories
Techne Prime Large-Format Thermal Cyclers Accessories
Choose from a reliable and robust selection of thermal cycling options based on your needs
Colorful touch screen allows for quick set up and easy programming with intuitive software
Uses the Peltier effect—a chosen method for the most trusted thermal cyclers on the market
Heated lid prevents condensation in your samples
Easily upgrade your unit to gradient cycling, now or later
These high-performance thermal cyclers provide robust and reliable operation that is user friendly and easy on the pocketbook. Intuitive programming feature allows you to easily set up to 1000 programs (unlimited with USB) with password protection to minimize inadvertent changes in settings. Use the color touch screen to create even the most complex programs.
Heated lid has an adjustable temperature range of 100 to 115°C to prevent condensation in your samples. All units have the ability to transfer programs via the USB port between instruments or to your PC. An industry-leading 4-year warranty ensures years of reliable results.
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Compare
Product Detail
Sort
Most Popular
Name
Name Descending
Price (Lowest First)
Price (Highest First)
Availability
Price
PRIME Base block, 96x0.2ML
EW-93945-51
Usually ships in 4 days.
$2,200.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Techne Prime Large-Format Thermal Cyclers Accessories
Prime USB Gradient Upgrade
EW-93945-57
Usually ships in 4 days.
$1,560.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Techne Prime Large-Format Thermal Cyclers Accessories
PRIME Base block, 60x0.5ML
EW-93945-53
Usually ships in 4 days.
$2,200.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Techne Prime Large-Format Thermal Cyclers Accessories
Prime Base Block, 384 Well Plates
EW-93945-55
Usually ships in 4 days.
$2,200.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Techne Prime Large-Format Thermal Cyclers Accessories
4
results (showing
1
-
4
)
Per Page
5
10
15
25
50
Home
>
Life Science
>
PCR (Polymerase Chain Reaction)
>
Thermal Cyclers
>
Techne Prime Large-Format Thermal Cyclers
> Techne Prime Large-Format Thermal Cyclers Accessories
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity