Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Home
>
Laboratory Cleaning Supplies
>
Mops
> Texwipe Alphamop
Texwipe Alphamop
Ideal for use in ultraclean production environments
Alphamop system features easily, changeable, laundered polyester heads that fit over a flat-padded head assembly. Users can rapidly change wiping surfaces as they mop to adhere to strict wiping protocols. Polyester head provides a clean substrate ideal for use on walls, floors, and ceilings. Foam pad conforms head to textured surfaces. Lightweight, fiberglass handle enables superior control and prevents arm fatigue during cleaning. Order mop components separately below.
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Compare
Product Detail
Sort
Most Popular
Name
Name Descending
Price (Lowest First)
Price (Highest First)
Availability
Price
Alphamop, pads (6) and covers (150)
Product Type Mops,Description Alphamop pads (6) and head covers (150),Brand TEXWIPE
EW-86518-02
Usually ships in 15 days.
$561.00 USD/CASE
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Texwipe Alphamop
1
results (showing
1
-
1
)
Per Page
5
10
15
25
50
Home
>
Laboratory Cleaning Supplies
>
Mops
> Texwipe Alphamop
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity