log in or register
Leasing Available
Financing is avaliable for many of our products.
See Details
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
Thermal Cyclers
Thermal Cyclers
PCR is a method that is used for various biological applications. The technique typically relies on thermal cyclers to amplify strands of DNA in order to replicate up to millions of copies of a DNA sequence.
Thermal cyclers, also known as PCR machine or DNA amplifiers, still represent a significant purchase in the laboratory. Please use this guide to assist you in selecting the most appropriate unit for your specific requirements.
One of the most important features in thermal cyclers is its ability to run numerous PCR samples simultaneously. Low-throughput thermocyclers can run typically between 20 and 48 wells. Typically, they are less costly and require less bench space. Medium throughput thermal cyclers are the most common, allowing between 48 and 96 wells. Typically this is adequate for most laboratories. High-throughput (384) + wells are required in some circumstances.
Another important feature when choosing a thermal cycler is thermal gradient. Some thermal cyclers incorporate a gradient function, allowing different temperatures to be applied in various parts of the block. A thermal gradient function is particularly useful when testing suitable annealing temperatures for primers. If the goal is uniform temperature across the entire thermal cycler block, a standard thermal cycler with no thermal gradient is usually adequate.
Heated lids are another potentially important critical feature when selecting a thermal cycler. Heated lids allow a sample to be heated more efficiently-minimizing formation of non-specific annealing and primer-dimers. This function also stymies water condensation on the lid and evaporation of your samples.
Other features to consider:
Flexibility- if your lab is expected to grow, it is comforting to know your cyclers have the ability to link easily together and be controlled with one device.
Intuitive programming- the ability to quickly edit your protocols and programs can quicken your testing.
Ramp rates- Heating and cooling rates of cyclers influience the speed of a reaction. The quicker the ramp rate, the faster the protocol is finished.
Quantitive PCR or qPCR is being used for such measurements such as genotyping, drug target validation, and other applications require quantification of the amont of DNA in a starting sample.
Real time PCR- requires a thermal cycler that is able to raise and lower the temperature in an appropriate way for the analysis.
Other PCR products: It is critical that consumables used to with PCR experimentation do not contain any impurities which could adversely affect your samples. In particular, amplification of minute quantities of genetic substances should be protected from degradation. Look for DNA/RNAse free and non-pyrogenic products.
 Product Detail
Sort
 Availability
 Price
Representative photo only Techne Prime Base Thermal Cycler 96 x 0 2ml
Techne­ Prime Base Thermal Cycler, 96 x 0.2ml
Product Type Large-Format Thermal Cycler, Prime Base,Display 5.7" color touch screen with graphical display,CE Compliance Yes
EW-93945-13
Usually ships in 4 days.
$5,750.00 USD/EACH
Qty:
Representative photo only Techne 3 Prime X Thermal Personal Cyclers with Extended Capacity 48 x 0 2ml
Techne 3 Prime X Thermal Personal Cyclers with Extended Capacity, 48 x 0.2ml
Product Type Personal Thermal Cycler with extended capacity,Display 3.2" color touch screen with graphical display,CE Compliance Yes
EW-93945-05
Usually ships in 4 days.
$4,100.00 USD/EACH
Qty:
Representative photo only Techne 3 Prime Base Personal Thermal Cycler 24 x 0 2ml
Techne 3 Prime Base Personal Thermal Cycler, 24 x 0.2ml
Product Type Personal Thermal Cycler,Display 3.2" color touch screen with graphical display,CE Compliance Yes
EW-93945-01
Usually ships in 4 days.
$3,400.00 USD/EACH
Qty:
Representative photo only PRIME Base block 96x0 2ML
EW-93945-51
Usually ships in 4 days.
$2,200.00 USD/EACH
Qty:
Representative photo only Eppendorf Mastercycler Nexus Thermal Cycler
Eppendorf Mastercycler Nexus Thermal Cycler
Capacity 96 x 0.2 mL PCR tubes, 1 PCR plate (8 x 12 array), or up to 77 x 0.5 mL PCR tubes,Dimensions " W 9-3/4,Dimensions " H 12-1/2
EW-93944-32
Usually ships in 10 days.
$7,200.00 USD/EACH
Qty:
Representative photo only Techne 3 Prime G Thermal Personal Cyclers with Thermal Gradient 48 x 0 2ml
Techne 3 Prime G Thermal Personal Cyclers with Thermal Gradient, 48 x 0.2ml
Product Type Personal Thermal Cycler with thermal gradient,Display 3.2" color touch screen with graphical display,CE Compliance Yes
EW-93945-09
Usually ships in 4 days.
$4,750.00 USD/EACH
Qty:
Representative photo only Eppendorf Mastercycler Nexus Thermal Cycler Flat
Eppendorf Mastercycler Nexus Thermal Cycler Flat
Capacity 96 x 0.2 mL PCR tubes, 1 PCR plate (8 x 12 array), or up to 77 x 0.5 mL PCR tubes,Dimensions " W 9-3/4,Dimensions " H 12-1/2
EW-93944-30
Usually ships in 10 days.
$7,400.00 USD/EACH
Qty:
Representative photo only Techne PrimeQ Real time PCR System
Techne PrimeQ Real-time PCR System
Product Type PCR System,CE Compliance Yes,Description 8 x Peltier block employing quad-circuit technology to enhance performance
EW-93948-00
Usually ships in 5 days.
$23,000.00 USD/EACH
Qty:
EW-93945-11 Image is representative of item.
Techne 3 Prime G Thermal Personal Cyclers with Thermal Gradient, 30 X 0.5ML
Product Type Personal Thermal Cycler with thermal gradient,Display 3.2" color touch screen with graphical display,CE Compliance Yes
EW-93945-11
Usually ships in 4 days.
$4,750.00 USD/EACH
Qty:
See all
Representative photo only 3 PRIME Base block 18 x0 5ML
3 PRIME Base block, 18 x0.5ML
Product Type Interchangeable Block,Brand Techne,Model 3PRIMEBASE/05/B
EW-93945-39
Usually ships in 4 days.
$1,170.00 USD/EACH
Qty:
Representative photo only Techne Prime G Gradient Thermal Cycler 384 Wells
Techne­ Prime G Gradient Thermal Cycler, 384 Wells
Product Type Large-Format Thermal Cycler, Prime G,Display 5.7" color touch screen with graphical display,CE Compliance Yes
EW-93945-23
Usually ships in 4 days.
$7,300.00 USD/EACH
Qty:
Representative photo only Prime Base Block 384 Well Plates
EW-93945-55
Usually ships in 4 days.
$2,200.00 USD/EACH
Qty:
Representative photo only FAM multiplex 460nm excitation wavelength 500nm emission wavelength
EW-93948-05
Call 1-888-358-4705 for availability.
$1,805.00 USD/EACH
Qty:
Representative photo only Yellow HEX 530nm excitation wavelength 560nm emission wavelength
EW-93948-07
Call 1-888-358-4705 for availability.
$975.00 USD/EACH
Qty:
Representative photo only Techne Prime Base Thermal Cycler 384 Well Plates
Techne­ Prime Base Thermal Cycler, 384 Well Plates
Product Type Large-Format Thermal Cycler, Prime Base,Display 5.7" color touch screen with graphical display,CE Compliance Yes
EW-93945-17
Usually ships in 4 days.
$5,750.00 USD/EACH
Qty:
40 results (showing 1 - 15 )
Per Page 
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title