log in or register
Availability:
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
Home  >  Material Testing & Physical Testing > Physical Property Testing > Viscometers > Thermo Scientific HAAKE Portable Viscometers
Thermo Scientific HAAKE Portable Viscometers
Thermo Scientific HAAKE Portable Viscometers
  • Small, battery-operated design for quick field tests and comparative measurements
  • Rotational viscometers provide quick, exact, and reliable measurements for a variety of fluid types ranging from paints and shampoos to sauces and oils
  • Simple, one button operation eliminates set-up time and allows hassle-free testing
These lightweight digital viscometers can be operated handheld or on a stand (order separately) for your convenience. Converting the handheld to use on a stand is only a matter of seconds - simply place in the special instrument holder. Use for quick viscosity tests for process optimization, machine adjustment, or to ensure your product is within specification.
By displaying viscosity on a digital display, human error associated with manual timing and misreading analog dials are eliminated. Possible handling errors as well as service information are also shown on the display.
Pushing a button tp turn the unit on; push the button again to select the rotor type, then begin measuring. A rotor rotating at a constant speed is immersed in the fluid to be tested; the fluid"s resistance to the rotation is correlated to measuring the fluid viscosity. Three different-sized rotors are included with the meter for multiple application capabilities.
 Product Detail
Sort
 Availability
 Price
Representative photo only Thermo Scientific HAAKE High range portable viscometer range 30 to 400 000 cP
Thermo Scientific HAAKE High-range portable viscometer, range 30 to 400,000 cP
Product Type Rotational Viscometer,rpm 65.5,Max temperature 302F (150C)
EW-08705-02
Call 1-888-358-4705 for availability.
$2,764.00 USD/EACH
Qty:
Representative photo only Thermo Scientific HAAKE Low range portable viscometer range 1 5 to 330 cP
Thermo Scientific HAAKE Low-range portable viscometer, range 1.5 to 330 cP
Product Type Rotational Viscometer,rpm 65.5,Max temperature 302F (150C)
EW-08705-01
Call 1-888-358-4705 for availability.
$2,764.00 USD/EACH
Qty:
2 results (showing 1 - 2 )
Per Page 
Home  >  Material Testing & Physical Testing > Physical Property Testing > Viscometers > Thermo Scientific HAAKE Portable Viscometers
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title