log in or register
Price Range:
Availability:
Number of Blocks:
1 (2)
2 (2)
3 (2)
4 (2)
6 (2)
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
Home  >  Heating Equipment > Block Heaters > Thermo Scientific Multi-Blok Dry Block Heaters and Blocks > Thermo Scientific Multi-Blok Dry Block Heaters (Analog)
Thermo Scientific Multi-Blok Dry Block Heaters (Analog)
Thermo Scientific Multi-Blok Dry Block Heaters (Analog)
  • Temperature uniformity within ±0.5degC (±0.9degF)
  • Three-position switch—high, low, or off
  • Low-range between ambient and 60degC (140degF) and
  • high-range to 130degC (266degF)
These versatile heating systems uniformly heat a wide range of test tubes simultaneously. Systems offer different combination of blocks for your specific needs. A complete heater system consists of a standard heater base and heating blocks (sold separately). Each block heater base features a stepless thermostat dial marked from 0 to 10 for repeatable settings. A separate power switch with power-on light operates independently of the thermostat dial.
Metal-sheathed safety heaters ensure temperature uniformity and rapid heating. Heater bases have steel housing with baked enamel finish, and include 6-ft, three-wire power cord; 115 VAC models include a plug.
 Product Detail
Sort
 Availability
 Price
Representative photo only Thermo Scientific Multi Blok Analog Dry Block Heater Base 1 block 120VAC
Thermo Scientific Multi-Blok Analog Dry Block Heater Base, 1-block, 120VAC
Product Type Dry Block Heaters-Analog,Max temperature º F 266,Power VAC 120
EW-36402-08
In stock.
$525.00 USD/EACH
Qty:
Representative photo only Thermo Scientific Multi Blok Analog Dry Block Heater Base 4 block 120 VAC
Thermo Scientific Multi-Blok Analog Dry Block Heater Base, 4-block, 120 VAC
Product Type Dry Block Heaters-Analog,Max temperature º F 266,Power VAC 120
EW-36402-14
Call 1-888-358-4705 for availability.
$1,110.00 USD/EACH
Qty:
Representative photo only Thermo Scientific Multi Blok Analog Dry Block Heater Base 3 block 120VAC
Thermo Scientific Multi-Blok Analog Dry Block Heater Base, 3-block, 120VAC
Product Type Dry Block Heaters-Analog,Max temperature º F 266,Power VAC 120
EW-36402-12
Usually ships in 8 days.
$905.00 USD/EACH
Qty:
Representative photo only Thermo Scientific Multi Blok Analog Dry Block Heater Base 2 block 120VAC
Thermo Scientific Multi-Blok Analog Dry Block Heater Base, 2-block, 120VAC
Product Type Dry Block Heaters-Analog,Max temperature º F 266,Power VAC 120
EW-36402-10
Call 1-888-358-4705 for availability.
$675.00 USD/EACH
Qty:
Representative photo only Thermo Scientific Multi Blok Analog Dry Block Heater Base 2 block 230 VAC
Thermo Scientific Multi-Blok Analog Dry Block Heater Base, 2-block, 230 VAC
Product Type Dry Block Heaters-Analog,Max temperature º F 266,Power VAC 230
EW-36402-11
Call 1-888-358-4705 for availability.
$675.00 USD/EACH
Qty:
Representative photo only Thermo Scientific Multi Blok Analog Dry Block Heater Base 1 block 230 VAC
Thermo Scientific Multi-Blok Analog Dry Block Heater Base, 1-block, 230 VAC
Product Type Dry Block Heaters-Analog,Max temperature º F 266,Power VAC 230
EW-36402-09
Call 1-888-358-4705 for availability.
$525.00 USD/EACH
Qty:
Representative photo only Thermo Scientific Multi Blok Analog Dry Block Heater Base 3 block 230 VAC
Thermo Scientific Multi-Blok Analog Dry Block Heater Base, 3-block, 230 VAC
Product Type Dry Block Heaters-Analog,Max temperature º F 266,Power VAC 230
EW-36402-13
Call 1-888-358-4705 for availability.
$905.00 USD/EACH
Qty:
Representative photo only Thermo Scientific Multi Blok Analog Dry Block Heater Base 6 block 120 VAC
Thermo Scientific Multi-Blok Analog Dry Block Heater Base, 6-block, 120 VAC
Product Type Dry Block Heaters-Analog,Max temperature º F 266,Power VAC 120
EW-36402-16
Usually ships in 11 days.
$1,375.00 USD/EACH
Qty:
Representative photo only Thermo Scientific Multi Blok Analog Dry Block Heater Base 4 block 230 VAC
Thermo Scientific Multi-Blok Analog Dry Block Heater Base, 4-block, 230 VAC
Product Type Dry Block Heaters-Analog,Max temperature º F 266,Power VAC 230
EW-36402-15
Call 1-888-358-4705 for availability.
$1,110.00 USD/EACH
Qty:
Representative photo only Thermo Scientific Multi Blok Analog Dry Block Heater Base 6 block 230 VAC
Thermo Scientific Multi-Blok Analog Dry Block Heater Base, 6-block, 230 VAC
Product Type Dry Block Heaters-Analog,Max temperature º F 266,Power VAC 230
EW-36402-17
Call 1-888-358-4705 for availability.
$1,375.00 USD/EACH
Qty:
10 results (showing 1 - 10 )
Per Page 
Home  >  Heating Equipment > Block Heaters > Thermo Scientific Multi-Blok Dry Block Heaters and Blocks > Thermo Scientific Multi-Blok Dry Block Heaters (Analog)
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title