log in or register
Price Range:
Number of Blocks:
1 (2)
2 (2)
3 (2)
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
Home  >  Heating Equipment > Block Heaters > Thermo Scientific Multi-Blok Dry Block Heaters and Blocks > Thermo Scientific Multi-Blok Dry Block Heaters (Digital)
Thermo Scientific Multi-Blok Dry Block Heaters (Digital)
Thermo Scientific Multi-Blok Dry Block Heaters (Digital)
  • Temperature uniformity within ±0.3degC (±0.5degF)
  • Temperature control accuracy of ±0.2degC (±0.4degF)
  • Temperature range above ambient to 100degC (212degF)
These versatile heating systems uniformly heat a wide range of test tubes simultaneously. Systems offer different combination of blocks for your specific needs. A complete heater system consists of a LED heater base and heating blocks (sold separately). LED bases are microprocessor controlled. A separate power switch with power-on light operates independently of the temperature control.
Block heater bases feature metal-sheathed safety heaters that ensure temperature uniformity and rapid heating. Heater bases have steel housing with baked enamel finish, and include 6-ft, three-wire power cord; 115 VAC models include a plug.
 Product Detail
Sort
 Availability
 Price
Representative photo only Thermo Scientific Multi Blok Dry Block LED Display Heater Base one block 115 VAC 60 Hz
Thermo Scientific Multi-Blok Dry Block LED Display Heater Base; one-block, 115 VAC/60 Hz
Product Type Dry Block Heaters-Digital,Max temperature º F 266,Power VAC 115
EW-36290-00
Usually ships in 8 days.
$925.00 USD/EACH
Qty:
Representative photo only Thermo Scientific Multi Blok Dry Block LED Display Heater Base 2 block 120 VAC
Thermo Scientific Multi-Blok Dry Block LED Display Heater Base;, 2-block, 120 VAC
Product Type Dry Block Heaters-Digital,Max temperature º F 266,Power VAC 120
EW-36402-00
Call 1-888-358-4705 for availability.
$1,175.00 USD/EACH
Qty:
Representative photo only Thermo Scientific Multi Blok Dry Block LED Display Heater Base 4 block 230 VAC
Thermo Scientific Multi-Blok Dry Block LED Display Heater Base; 4-block, 230 VAC
Product Type Dry Block Heaters-Digital,Max temperature º F 266,Power VAC 240
EW-36402-06
Usually ships in 9 days.
$1,475.00 USD/EACH
Qty:
Representative photo only Thermo Scientific Multi Blok Dry Block LED Display Heater Base 4 block 120 VAC
Thermo Scientific Multi-Blok Dry Block LED Display Heater Base; 4-block, 120 VAC
Product Type Dry Block Heaters-Digital,Max temperature º F 266,Power VAC 120
EW-36402-04
Call 1-888-358-4705 for availability.
$1,475.00 USD/EACH
Qty:
Representative photo only Thermo Scientific Multi Blok Dry Block LED Display Heater Base one block 220 VAC 60 Hz
Thermo Scientific Multi-Blok Dry Block LED Display Heater Base; one-block, 220 VAC/60 Hz
Product Type Dry Block Heaters-Digital,Max temperature º F 212,Power VAC 240
EW-36290-02
Usually ships in 12 days.
$925.00 USD/EACH
Qty:
Representative photo only Thermo Scientific Multi Blok Dry Block LED Display Heater Base 2 block 230 VAC
Thermo Scientific Multi-Blok Dry Block LED Display Heater Base;, 2-block, 230 VAC
Product Type Dry Block Heaters-Digital,Max temperature º F 266,Power VAC 240
EW-36402-02
Call 1-888-358-4705 for availability.
$1,175.00 USD/EACH
Qty:
Representative photo only Thermo Scientific Multi Blok Dry Block LED Display Heater Base three block 220 VAC 60 Hz
Thermo Scientific Multi-Blok Dry Block LED Display Heater Base; three-block, 220 VAC/60 Hz
Product Type Dry Block Heaters-Digital,Max temperature º F 212,Power VAC 240
EW-36290-22
Call 1-888-358-4705 for availability.
$1,250.00 USD/EACH
Qty:
Representative photo only Thermo Scientific Multi Blok Dry Block LED Display Heater Base three block 115 VAC 600 Hz
Thermo Scientific Multi-Blok Dry Block LED Display Heater Base; three-block, 115 VAC/600 Hz
Product Type Dry Block Heaters-Digital,Max temperature º F 266,Power VAC 200
EW-36290-20
Call 1-888-358-4705 for availability.
$1,250.00 USD/EACH
Qty:
8 results (showing 1 - 8 )
Per Page 
Home  >  Heating Equipment > Block Heaters > Thermo Scientific Multi-Blok Dry Block Heaters and Blocks > Thermo Scientific Multi-Blok Dry Block Heaters (Digital)
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title