Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Leasing Available
Financing is avaliable for many of our products.
See Details
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Home
>
Material Testing & Physical Testing
>
Spectroscopy and Spectrophotometers
>
Visible/UV-Visible Spectroscopy
>
Thermo Scientific NanoDrop Spectrophotometers
> Thermo Scientific NanoDrop 2000 & 2000c Spectrophotometers
Thermo Scientific NanoDrop 2000 & 2000c Spectrophotometers
The NanoDrop 2000 is the perfect instrument for almost every lab dealing with sample analysis or preparation.
- Easy to use, simply pipette sample directly onto the pedestal
- Designed for micro-volume sample size (0.5 µL – 2.0 µL)
- Requires no dilutions, even for highly concentrated samples (up to 15,000 ng/ µL)
- Fast, less than five second measurement time
- Complete with user friendly software packed with features
The NanoDrop 2000c does everything the NanoDrop 2000 does, plus more. The NanoDrop 2000c combines micro-volume pedestal technology and cuvette capability in a single instrument.
- Dual modes – choose the measuring option right for your sample – pedestal or cuvette
- Very broad concentration range for measuring both very low concentrations or very high concentrations
- cuvette capability for kinetics studies and cell culture (OD 600) measurements
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Compare
Product Detail
Sort
Most Popular
Name
Name Descending
Price (Lowest First)
Price (Highest First)
Availability
Price
Thermo Scientific NanoDrop
ND-2000 1-position Spectrophotometer
Product Type UV/Visible Spectroscopes,Model ND-2000,Brand Thermo Scientific NanoDrop
EW-83061-00
Usually ships in 4 days.
$9,570.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Thermo Scientific NanoDrop
2000 & 2000c Spectrophotometers
Thermo Scientific NanoDrop
ND-2000c Spectrophotometer with Cuvette
Product Type UV/Visible Spectroscopes,Model ND-2000c,Brand Thermo Scientific NanoDrop
EW-83061-12
Usually ships in 3 days.
$11,300.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Thermo Scientific NanoDrop 2000 & 2000c Spectrophotometers
Thermo Scientific NanoDrop
ND-2000 Dual-mode UV-Vis Spectrophotometer with Laptop
Product Type UV/Visible Spectroscopes,Model ND-2000,Brand Thermo Scientific NanoDrop
EW-83061-52
Usually ships in 8 days.
$12,300.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Thermo Scientific NanoDrop 2000 & 2000c Spectrophotometers
Thermo Scientific NanoDrop
ND-2000 1-position Spectrophotometer with Laptop
Product Type UV/Visible Spectroscopes,Model ND-2000,Brand Thermo Scientific NanoDrop
EW-83061-50
Usually ships in 8 days.
$10,570.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Thermo Scientific NanoDrop 2000 & 2000c Spectrophotometers
4
results (showing
1
-
4
)
Per Page
5
10
15
25
50
Home
>
Material Testing & Physical Testing
>
Spectroscopy and Spectrophotometers
>
Visible/UV-Visible Spectroscopy
>
Thermo Scientific NanoDrop Spectrophotometers
> Thermo Scientific NanoDrop 2000 & 2000c Spectrophotometers
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity