Subtotal = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
Price Range:
Capacity (Liters):
11.3 (2)
26.5 (2)
CE Compliance:
Yes (2)
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
 
Home  >  Lab Equipment > Baths and Circulators > Specialty Baths > Steam Baths > Thermo Scientific Precision Concentric Ring Electrical Steaming Baths

Thermo Scientific Precision Concentric Ring Electrical Steaming Baths

Thermo Scientific Precision Concentric Ring Electrical Steaming Baths

  • Concentric rings can be removed from inner cover outward to accommodate various vessel shapes and sizes
  • Copper clad immersion heaters provide bath temperatures up to 100°C
  • Easy to operate with a 4-position switch and 3 heat settings
Thermo Scientific PRECISION Concentric Ring Steaming Baths are designed to gently warm samples, evaporate liquids, and melt solids such as agar. Concentric rings can be removed from inner cover outward to accommodate various vessel shapes and sizes. Four- and eight-hole models enlarge in 1” increments from 7/8” at the center, to a maximum opening diameter of 5 and 6 inches, respectively. Copper clad immersion heaters provide bath temperatures up to 100°C, with three variable heat settings.

Maintain bath depth at desired level with adjustable water level regulator. Epoxy polyester-coated steel support frame is resistant to corrosion. Stainless steel flanged cabinet top can be installed in the benchtop providing a permanent station. Baths must be hard- wired to the electrical system by a qualified electrician.
 Product Detail
Sort
 Availability
 Price
Representative photo only Thermo Scientific PRECISION Four hole steaming bath 240V

Thermo Scientific PRECISION Four-hole steaming bath, 240V

Product Type Water Bath,Bath type Special Purpose Water Baths,Capacity Liters 11.3
EW-24100-02
Call 1-888-358-4705 for availability.
$1,461.00 USD/EACH

Qty:

Representative photo only Thermo Scientific PRECISION Eight hole steaming bath 240V

Thermo Scientific PRECISION Eight-hole steaming bath, 240V

Product Type Water Bath,Bath type Special Purpose Water Baths,Capacity Liters 26.5
EW-24100-06
Call 1-888-358-4705 for availability.
$2,002.00 USD/EACH

Qty:

EW-24100-00 Thermo Scientific PRECISION Four-hole steaming bath, 120V

Thermo Scientific PRECISION Four-hole steaming bath, 120V

Product Type Water Bath,Bath type Special Purpose Water Baths,Capacity Liters 11.3
EW-24100-00
Call 1-888-358-4705 for availability.
$1,461.00 USD/EACH

Qty:

Representative photo only Thermo Scientific PRECISION Eight hole steaming bath 120V

Thermo Scientific PRECISION Eight-hole steaming bath, 120V

Product Type Water Bath,Bath type Special Purpose Water Baths,Capacity Liters 26.5
EW-24100-04
Call 1-888-358-4705 for availability.
$1,455.00 USD/EACH

Qty:

4 results (showing 1 - 4 )
Per Page 
Home  >  Lab Equipment > Baths and Circulators > Specialty Baths > Steam Baths > Thermo Scientific Precision Concentric Ring Electrical Steaming Baths

View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title