log in or register
Capacity (Liters):
1 to 5 (4)
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
Home  >  Lab Equipment > Baths and Circulators > Laboratory, Utility Water Baths > Thermo Scientific Precision General-Purpose Heated Baths > Thermo Scientific Precision General-Purpose Heated Digital Baths
Thermo Scientific Precision General-Purpose Heated Digital Baths
Thermo Scientific Precision General-Purpose Heated Digital Baths
  • Seamless 304 stainless steel interior and epoxy powder-coated exterior—corrosion resistant and easy to maintain
  • Fully insulated chamber with stainless steel diffuser shelf achieves desired temperature quickly.
  • Adjust temperature setpoint via front-mounted control panel
  • Over-temperature safety circuitry prevents thermal runaway.
Choose between analog and digital control, single-chamber and dual-chamber baths. All baths include 304 stainless steel gable covers except for models 12418-10, -15 and models 12428-10, -15; these baths include polypropylene gable covers. Covers allow for rapid heat up to the maximum temperature. All models include an 8-ft power cord with grounded plug (the 230 VAC models have a European plug).
Digital Baths offer high performance and reliability. Microprocessor-based controller and fiberglass insulation provide excellent heat control and uniformity. Program temperature setpoint with the push of a button. Each bath features LED readout, automatic over-temperature protection, and built-in calibration capabilities.
 Product Detail
Sort
 Availability
 Price
Representative photo only Thermo Scientific Precision General Purpose Heated Digital Bath 19 5 Liters 230V
Thermo Scientific Precision General-Purpose Heated Digital Bath; 19.5 Liters; 230V
Product Type Water Bath,Bath type Digital Water Bath,Capacity Liters 19.5
EW-12428-45
Usually ships in 13 days.
$1,423.00 USD/EACH
Qty:
Representative photo only Thermo Scientific Precision General Purpose Heated Digital Bath Dual Chamber 115V
Thermo Scientific Precision General-Purpose Heated Digital Bath; Dual-Chamber; 115V
Product Type Water Bath,Bath type Digital Water Bath,Capacity Liters 12
EW-12428-80
Usually ships in 5 days.
$2,092.00 USD/EACH
Qty:
Representative photo only Thermo Scientific Precision General Purpose Heated Digital Bath 19 5 Liters 115V
Thermo Scientific Precision General-Purpose Heated Digital Bath; 19.5 Liters; 115V
Product Type Water Bath,Bath type Digital Water Bath,Capacity Liters 19.5
EW-12428-40
Usually ships in 4 days.
$1,423.00 USD/EACH
Qty:
Representative photo only Thermo Scientific Precision General Purpose Heated Digital Bath 1 5 Liters 230V
Thermo Scientific Precision General-Purpose Heated Digital Bath; 1.5 Liters, 230V
Product Type Water Bath,Bath type Digital Water Bath,Capacity Liters 1.5
EW-12428-05
Call 1-888-358-4705 for availability.
$832.00 USD/EACH
Qty:
Representative photo only Thermo Scientific Precision General Purpose Heated Digital Bath 12 Liters 115V
Thermo Scientific Precision General-Purpose Heated Digital Bath; 12 Liters; 115V
Product Type Water Bath,Bath type Digital Water Bath,Capacity Liters 12
EW-12428-30
Usually ships in 8 days.
$1,226.00 USD/EACH
Qty:
Representative photo only Thermo Scientific Precision General Purpose Heated Digital Bath 43 Liters 230V
Thermo Scientific Precision General-Purpose Heated Digital Bath; 43 Liters; 230V
Product Type Water Bath,Bath type Digital Water Bath,Capacity Liters 43
EW-12428-65
Call 1-888-358-4705 for availability.
$2,032.00 USD/EACH
Qty:
Representative photo only Thermo Scientific Precision General Purpose Heated Digital Bath 5 5 Liters 230V
Thermo Scientific Precision General-Purpose Heated Digital Bath; 5.5 Liters; 230V
Product Type Water Bath,Bath type Digital Water Bath,Capacity Liters 5.5
EW-12428-25
Usually ships in 14 days.
$1,220.00 USD/EACH
Qty:
Representative photo only Thermo Scientific Precision General Purpose Heated Digital Bath 43 Liters 115V
Thermo Scientific Precision General-Purpose Heated Digital Bath; 43 Liters; 115V
Product Type Water Bath,Bath type Digital Water Bath,Capacity Liters 43
EW-12428-60
Usually ships in 7 days.
$2,032.00 USD/EACH
Qty:
Representative photo only Thermo Scientific Precision General Purpose Heated Digital Bath 12 Liters 230V
Thermo Scientific Precision General-Purpose Heated Digital Bath; 12 Liters; 230V
Product Type Water Bath,Bath type Digital Water Bath,Capacity Liters 12
EW-12428-35
Usually ships in 10 days.
$1,226.00 USD/EACH
Qty:
Representative photo only Thermo Scientific Precision General Purpose Heated Digital Bath 1 5 Liters 115V
Thermo Scientific Precision General-Purpose Heated Digital Bath; 1.5 Liters, 115V
Product Type Water Bath,Bath type Digital Water Bath,Capacity Liters 1.5
EW-12428-00
Usually ships in 9 days.
$832.00 USD/EACH
Qty:
Representative photo only Thermo Scientific Precision General Purpose Heated Digital Bath 2 5 Liters 115V
Thermo Scientific Precision General-Purpose Heated Digital Bath; 2.5 Liters, 115V
Product Type Water Bath,Bath type Digital Water Bath,Capacity Liters 2.5
EW-12428-10
Usually ships in 6 days.
$881.00 USD/EACH
Qty:
Representative photo only Thermo Scientific Precision General Purpose Heated Digital Bath 2 5 Liters 230V
Thermo Scientific Precision General-Purpose Heated Digital Bath; 2.5 Liters; 230V
Product Type Water Bath,Bath type Digital Water Bath,Capacity Liters 2.5
EW-12428-15
Call 1-888-358-4705 for availability.
$861.00 USD/EACH
Qty:
EW-12428-50 Thermo Scientific Precision General-Purpose Heated Digital Bath; 18 Liters; 115V
Thermo Scientific Precision General-Purpose Heated Digital Bath; 18 Liters; 115V
Product Type Water Bath,Bath type Digital Water Bath,Capacity Liters 18
EW-12428-50
Usually ships in 4 days.
$1,610.00 USD/EACH
Qty:
Representative photo only Thermo Scientific Precision General Purpose Heated Digital Bath 5 5 Liters 115V
Thermo Scientific Precision General-Purpose Heated Digital Bath; 5.5 Liters; 115V
Product Type Water Bath,Bath type Digital Water Bath,Capacity Liters 5.5
EW-12428-20
Usually ships in 5 days.
$1,189.00 USD/EACH
Qty:
Representative photo only Thermo Scientific Precision General Purpose Heated Digital Bath Dual Chamber 230V
Thermo Scientific Precision General-Purpose Heated Digital Bath; Dual-Chamber; 230V
Product Type Water Bath,Bath type Digital Water Bath,Capacity Liters 12
EW-12428-85
Call 1-888-358-4705 for availability.
$2,092.00 USD/EACH
Qty:
16 results (showing 1 - 15 )
Per Page 
Home  >  Lab Equipment > Baths and Circulators > Laboratory, Utility Water Baths > Thermo Scientific Precision General-Purpose Heated Baths > Thermo Scientific Precision General-Purpose Heated Digital Baths
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title