Product Type Prepared Media Plates,Description Irradiated. For cultivation of bacteria, yeast, and fungi.,Media Contact plate sterile tryptic soy agar with lecithin, polysorbate 80,Qty/pk 10
Usually ships in 4 days.
Product Type Prepared Media Plates,Description General-purpose medium for cultivation and isolation of microorganisms. Industrial reference:
AOAC, BAM, COMPF, IDF, USDA, USP, SMWW,Media Tryptic soy agar (TSA), soybean casein digest,Qty/pk 10
Usually ships in 8 days.
Product Type Prepared Media Plates,Description General-purpose medium for cultivation and isolation of microorganisms. Industrial reference: BAM,Media Blood agar (TSA with 5% sheep blood),Qty/pk 10
Usually ships in 6 days.
Product Type Prepared Media Plates,Description For cultivation, identification, and enumeration of yeast and fungi. Industrial reference: BAM, COMPF, USP,Media Potato dextrose agar, pH 5.6,Qty/pk 10
Usually ships in 11 days.
Product Type Prepared Media Plates,Description For selective and differential isolation of sorbitol-negative Escherichia coli associatedwith verotoxin production, especially O157:H7. Industrial reference: BAM, COMPF,Media MacConkey agar with sorbitol (SMAC),Qty/pk 10
Usually ships in 4 days.
Product Type Prepared Media Plates,Description For cultivation and isolation of yeast and fungi. Lower pH enhances growth of fungi, especially dermatophytes, and is slightly inhibitory to bacteria. Industrial reference: BAM, COMPF, USP,Media Sabouraud dextrose agar, pH 5.6,Qty/pk 10
Usually ships in 5 days.
Product Type Prepared Media Plates,Description For isolation and differentiation of gram-negative enteric microorganisms. Industrial reference: BAM, COMPF,Media Hektoen enteric (HE) agar,Qty/pk 10
Usually ships in 9 days.
Product Type Prepared Media Plates,Description For selective isolation and differentiation of gram-negative enteric bacilli from samples. Industrial reference: BAM, COMPF, IDF, USP,Media EMB agar, Levine,Qty/pk 10
Usually ships in 4 days.
Product Type Prepared Media Plates,Description For selective and differential isolation of gram-negative enteric pathogens. Industrial reference:
AOAC, BAM, COMPF, USP,Media XLD (xylose, lysine, desoxycholate) agar,Qty/pk 10
Usually ships in 5 days.
Product Type Prepared Media Plates,Description For cultivation of bacteria, yeast, and fungi. Manufactured in a cleanroom environment to provide the highest level of sterility assurance.,Media Contact I tryptic soy agar with lecithin, polysorbate 80,Qty/pk 20
Usually ships in 14 days.
Product Type Prepared Media Plates,Description For selective isolation and presumptive identification of Pseudomonas aeruginosa and othergram-negative, nonfermentative bacteria. Industrial reference: BAM, USP,Media Cetrimide selective agar,Qty/pk 10
Usually ships in 5 days.
Product Type Prepared Media Plates,Description For selective isolation, differentiation, and enumeration of staphylococci. Industrial reference: BAM, USP,Media Mannitol salt agar (MSA),Qty/pk 10
Usually ships in 5 days.
Product Type Prepared Media Plates,Description For microbial plate counts from milk, dairy products, food, water, and other specimens of sanitary importance.
Industrial reference: AOAC, BAM, COMPF, EPA, IDF, SMD, USDA,Media Plate count agar (Standard methods agar or Tryptone glucose yeast extract),Qty/pk 10
Call 1-888-358-4705 for availability.
Product Type Prepared Media Plates,Description General-purpose medium for cultivation of microorganisms,Media Contact I blood agar (tryptic soy agar with 5% sheep blood),Qty/pk 20
Usually ships in 5 days.
Product Type Prepared Media Plates,Description For isolation and presumptive identification of coagulase positive staphylococci. Uses tellurite reduction toproduce black colonies of Staphylococcus aureus. Industrial reference: AOAC, BAM, IDF, SMD, SMWW, USDA,Media Baird-Parker agar,Qty/pk 10
Usually ships in 5 days.