log in or register
Learn more:
[-] Show Less
Leasing Available
Financing is avaliable for many of our products.
See Details
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
Thermo Scientific Quantech Fluorometer
Thermo Scientific Quantech Fluorometer
  • Photomultiplier tube (PMT) and microprocessor control provide excellent detection sensitivity
  • Automatically sets gain (sensitivity) and range—eliminates time-consuming manual dial adjustments
  • Store up to nine multi-point standard curves in memory
  • Linear regression analysis permits direct concentration readings
  • Operates in English, Spanish, French, and German
  • RS-232 provides computer and printer connections
  • Retainer clip made of "memory metal" lets you use a variety of tube and cuvette sizes without special adapters
Compact and efficient, these fluorometers provide excellent analytical performance. Quantitate DNA and PCR products. Detect DNA contamination in drugs. Perform vitamin assays for pharmaceutical procedures. Environmental applications include the quantitative analysis of herbicides and pesticides in water and monitoring chlorophyll in lakes and water treatment plants.
Reduced source lamp replacement alone will show you the absolute economy of these fluorometers. For example, you can purchase a wide-band fluorometer (83075-20 or -25) with minimal operating costs for less than the cost of xenon or deuterium source lamp replacement for a single year.
 Product Detail
Sort
 Availability
 Price
Representative photo only Thermo Scientific Quantech Base Fluorometer 230 VAC
Thermo Scientific Quantech Base Fluorometer, 230 VAC
Product Type Fluorometers,Output RS-232,Applications Standard biotechnology; life sciences
EW-83075-05
Usually ships in 14 days.
$6,800.00 USD/EACH
Qty:
Representative photo only NB490 Excitation Filter for Turner Quantech Fluorometer narrow band
NB490 Excitation Filter for Turner Quantech Fluorometer; narrow band
Product Type Fluorometers,Applications RNA,Model LE1095X13
EW-83076-15
Usually ships in 14 days.
$275.00 USD/EACH
Qty:
Representative photo only Borosilicate Cuvettes for Turner Quantech Fluorometer Pack of Twelve
Borosilicate Cuvettes for Turner Quantech Fluorometer, Pack of Twelve
Product Type Fluorometers,Capacity 5 mL,Model 331775
EW-83075-46
Usually ships in 14 days.
$85.50 USD/PACKAGE
Qty:
Representative photo only SC515 Emission Filter for Turner Quantech Fluorometer sharp cut
SC515 Emission Filter for Turner Quantech Fluorometer; sharp cut
Product Type Fluorometers,Applications RNA,Model LE1095X23
EW-83076-65
Usually ships in 14 days.
$216.00 USD/EACH
Qty:
Representative photo only SC585 Emission Filter for Turner Quantech Fluorometer sharp cut
SC585 Emission Filter for Turner Quantech Fluorometer; sharp cut
Product Type Fluorometers,Applications Protein,Model LE1095X26
EW-83076-80
Usually ships in 14 days.
$216.00 USD/EACH
Qty:
Representative photo only SC450 Emission Filter for Turner Quantech Fluorometer sharp cut
SC450 Emission Filter for Turner Quantech Fluorometer; sharp cut
Product Type Fluorometers,Applications Histamine,Model LE1095X20
EW-83076-50
Usually ships in 14 days.
$216.00 USD/EACH
Qty:
Representative photo only SC430 Emission Filter for Turner Quantech Fluorometer sharp cut
SC430 Emission Filter for Turner Quantech Fluorometer; sharp cut
Product Type Fluorometers,Applications DNA,Model LE1095X19
EW-83076-45
Usually ships in 14 days.
$237.00 USD/EACH
Qty:
Representative photo only SC665 Emission Filter for Turner Quantech Fluorometer sharp cut
SC665 Emission Filter for Turner Quantech Fluorometer; sharp cut
Product Type Fluorometers,Applications Chlorophyll,Model LE1095X29
EW-83076-95
Usually ships in 14 days.
$237.00 USD/EACH
Qty:
Representative photo only Thermo Scientific Quantech Wide Band Fluorometer 120 VAC
Thermo Scientific Quantech Wide Band Fluorometer, 120 VAC
Product Type Fluorometers,Output RS-232,Applications Various
EW-83075-20
Call 1-888-358-4705 for availability.
$9,100.00 USD/EACH
Qty:
Representative photo only NB450 Excitation Filter for Turner Quantech Fluorometer narrow band
NB450 Excitation Filter for Turner Quantech Fluorometer; narrow band
Product Type Fluorometers,Applications Histamine,Model LE1095X11
EW-83076-05
Usually ships in 14 days.
$407.00 USD/EACH
Qty:
Representative photo only NB440 Excitation Filter for Turner Quantech Fluorometer narrow band
NB440 Excitation Filter for Turner Quantech Fluorometer; narrow band
Product Type Fluorometers,Applications Chlorophyll,Model LE1095X10
EW-83076-00
Usually ships in 14 days.
$278.00 USD/EACH
Qty:
Representative photo only Thermo Scientific Quantech Base Fluorometer 120 VAC
Thermo Scientific Quantech Base Fluorometer, 120 VAC
Product Type Fluorometers,Output RS-232,Applications Standard biotechnology; life sciences
EW-83075-00
Call 1-888-358-4705 for availability.
$6,800.00 USD/EACH
Qty:
Representative photo only Thermo Scientific Quantech Wide Band Fluorometer 230 VAC
Thermo Scientific Quantech Wide Band Fluorometer, 230 VAC
Product Type Fluorometers,Output RS-232,Applications Various
EW-83075-25
Usually ships in 14 days.
$9,100.00 USD/EACH
Qty:
13 results (showing 1 - 13 )
Per Page 
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title