Subtotal = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
Leasing Available
Financing is avaliable for many of our products.

See Details
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
 
Home  >  Cooling Equipment > Freezers > Thermo Scientific Freezers > Thermo Scientific Freezers > Thermo Scientific Revco Lab Freezers

Thermo Scientific Revco Lab Freezers

Thermo Scientific Revco Lab Freezers

  • Microprocessor driven for precise control and documentation
  • Enzyme freezer offers organized inventory system
  • Interlogic microprocessor control system maintains temperature
Check operating status at a glance with enlarged digital display and graphic thermometer. Set temperature and alarm set points in 1°C increments. IntrLogic™ microprocessor control provides better accuracy than other freezers. Set point security system protects your samples with a key-operated switch for main power and alarm locks in temperature and alarm set points. Alarm system audibly and visually identifies any temperature deviation beyond alarm set points and archives deviated temperatures. Extreme ambient alert warns if room temperature may have affected performance.

Gravity convection air system provides maximum temperature stability without blowers or fans. Manual defrost avoids intermittent coil warming during auto defrost cycles. Each model is constructed of heavy gauge steel and features hermetically sealed compressors and a single self-closing solid door. The full length door handle has keyed lock for security.
 Product Detail
Sort
 Availability
 Price
Representative photo only Thermo Scientific Revco Lab freezer 20C 115 VAC 60 Hz

Thermo Scientific Revco Lab freezer, -20°C, 115 VAC, 60 Hz

Product Type Enzyme Freezers,Capacity cu ft 23.3,Capacity Liters 659
EW-44164-10
Usually ships in 4 days.
$7,030.00 USD/EACH

Qty:

Representative photo only Thermo Scientific Revco Enzyme freezer 20C 208 230 VAC 60 Hz

Thermo Scientific Revco Enzyme freezer, -20°C, 208/230 VAC, 60 Hz

Product Type Enzyme Freezers,Capacity cu ft 23.3,Capacity Liters 659
EW-44164-05
Call 1-888-358-4705 for availability.
$8,140.00 USD/EACH

Qty:

Representative photo only Thermo Scientific Revco Enzyme freezer 20C 115 VAC 60 Hz

Thermo Scientific Revco Enzyme freezer, -20°C, 115 VAC, 60 Hz

Product Type Enzyme Freezers,Capacity cu ft 23.3,Capacity Liters 659
EW-44164-00
Call 1-888-358-4705 for availability.
$8,140.00 USD/EACH

Qty:

Representative photo only Thermo Scientific Revco Lab freezer 20C 220 VAC 50 HZ

Thermo Scientific Revco Lab freezer, -20°C, 220 VAC, 50 HZ

Product Type Enzyme Freezers,Capacity cu ft 23.3,Capacity Liters 659
EW-44164-15
Call 1-888-358-4705 for availability.
$7,030.00 USD/EACH

Qty:

4 results (showing 1 - 4 )
Per Page 
Home  >  Cooling Equipment > Freezers > Thermo Scientific Freezers > Thermo Scientific Freezers > Thermo Scientific Revco Lab Freezers

View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title