United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Home
>
Material Testing & Physical Testing
>
Spectroscopy and Spectrophotometers
>
Spectrometer Accessories
>
Calibration Products and Standards
> Thermo Scientific SPECTRONIC™ Standards Set for Visible Light Spectroscopy
Thermo Scientific SPECTRONIC™ Standards Set for Visible Light Spectroscopy
Easily and accurately perform certified spectrophotometer evaluations in your own laboratory
Designed for use in visible light ranges
ISO17025 accredited for testing and calibration
Check the performance and accuracy of your spectrophotometer right in your own lab. Simply insert standards into your spectrophotometer's cell holder. Standards include:
0% transmittance: the opaque 0%T standard is used to test for light leaks in your spectrophotometer.
Wavelength 590 nm: the wavelength accuracy standard has a transmittance peak near 515 nm which is certified as a wavelength accuracy standard and is NIST traceable. The symmetry of the peak means that the tests of wavelength accuracy are independent of the spectral bandwidth (up to 20 nm) of the instrument being tested. In addition, the testing peak of 515 nm is located at that point when the tungsten lamp shows the flattest energy measurement in response to wavelength change.
Stray radiant energy: each of these three standards is essentially opaque at its test wavelength and highly transmitting at longer wavelengths. The test wavelength is chosen (220 nm, 340 nm, and 400 nm) to correspond to points of low energy for both tungsten and deuterium lamps, readily revealing any stray energy for both the visible and UV regions.
Photometric performance: these 10% T and 50% T neutral density filters test spectrophotometric accuracy, precision, and linearity. No spectral bandwidth is required. The control standard verifies that the holder itself does not affect the 100%T reading.
Optical alignment: optical alignment in the sample compartment can be evaluated by using the control standard and photosensitive paper. The paper records the position of the light beam in the cuvette holder.
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Compare
Product Detail
Sort
Most Popular
Name
Name Descending
Price (Lowest First)
Price (Highest First)
Availability
Price
Thermo Scientific SPECTRONIC
standards, set of 10
Product Type Spectroscopy Calibration Standards ,Applications Visible Light Spectroscopy ,Brand Thermo Scientific
EW-02650-57
Usually ships in 14 days.
$943.00 USD/SET of 1
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Thermo Scientific SPECTRONIC™ Standards Set for Visible Light Spectroscopy
1
results (showing
1
-
1
)
Per Page
5
10
15
25
50
Home
>
Material Testing & Physical Testing
>
Spectroscopy and Spectrophotometers
>
Spectrometer Accessories
>
Calibration Products and Standards
> Thermo Scientific SPECTRONIC™ Standards Set for Visible Light Spectroscopy
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title