log in or register
Price Range:
Availability:
Membrane MWCO (Daltons):
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
Home  >  Life Science > Dialysis > Ready-to-use Dialysis Devices > Thermo Scientific Slide-A-Lyzer Dialysis Cassettes
Thermo Scientific Slide-A-Lyzer Dialysis Cassettes
Thermo Scientific Slide-A-Lyzer Dialysis Cassettes
  • High Performance Dialysis require just half the time of dialysis tubing for sample size from 100µl to 30mL
  • Reduces preparation time
  • The innovative cassette design does not require the use of knots or clips that can lead to leaking sample loss.
  • Color coded translucent frame permits smooth sample withdrawal of sub-filter volumes ¨C removing every last drop is easy.
  • High surface area/sample volume ratio will dialyze twice as fast as dialysis via conventional tubing.
  • Available in a variety of sizes
Slide-A-Lyzer® dialysis cassette is ideal for low molecular weight contaminant removal, buffer exchange, desalting and concentration. The cassette membrane is composed of low-binding regenerated cellulose and features a hermetically sealed sample chamber to maintain the highest possible sample retention. Sample introduction and removal are easily accomplished by penetrating the gasket with a hypodermic needle attached to a syringe. The gasket imparts resealability to the needle puncture, ensuring that no sample is lost from the cassette during dialysis.
Ready-to-use dialysis devices offer conveniences with preparing, loading and retrieving samples. These devices also increase recovery and decrease sample loss, while increasing sampling efficiency. Choose the ready-to-use device that fits your application. The Micro Float-A-Lyzer is ideal for small sample volumes, below 500µL. The Tube-A-Lyzer uses a flow-through buffer chamber to increase transfer rate and drastically reduce run times. The Float-A-Lyzer G2 makes sample loading and retrieval easy, and includes color coded cap colors for different Molecular Weight Cut-Offs (MWCO). The Thermo Scientific Slide-A-Lyzer has a high surface area to sample volume ratio to increase run times.
 Product Detail
Sort
 Availability
 Price
Representative photo only Thermo Scientific Slide A Lyzer Dialysis Cassettes 10 000 MWCO 0 5 3mL 10 cassettes
Thermo Scientific Slide-A-Lyzer Dialysis Cassettes, 10,000 MWCO, 0.5-3mL, 10 cassettes
Product Type Ready-to-use Dialysis Device,Material Regenerated Cellulose,Membrane MWCO (Daltons) 10000
EW-02905-42
In stock.
$86.00 USD/EACH
Qty:
Representative photo only Thermo Scientific Slide A Lyzer Dialysis Cassettes 10 000 MWCO 3 12mL 8 cassettes
Thermo Scientific Slide-A-Lyzer Dialysis Cassettes, 10,000 MWCO, 3-12mL, 8 cassettes
Product Type Ready-to-use Dialysis Device,Material Regenerated Cellulose,Membrane MWCO (Daltons) 10000
EW-02905-44
In stock.
$113.00 USD/EACH
Qty:
Representative photo only Thermo Scientific Slide A Lyzer Dialysis Cassettes 3500 MWCO 0 5 3mL 10 cassettes
Thermo Scientific Slide-A-Lyzer Dialysis Cassettes, 3500 MWCO, 0.5-3mL, 10 cassettes
Product Type Ready-to-use Dialysis Device,Material Regenerated Cellulose,Membrane MWCO (Daltons) 3500
EW-02905-03
Usually ships in 9 days or sooner.
$86.00 USD/EACH
Qty:
Representative photo only Thermo Scientific Slide A Lyzer Dialysis Cassettes 3500 MWCO 3 12mL 8 cassettes
Thermo Scientific Slide-A-Lyzer Dialysis Cassettes, 3500 MWCO, 3-12mL, 8 cassettes
Product Type Ready-to-use Dialysis Device,Material Regenerated Cellulose,Membrane MWCO (Daltons) 3500
EW-02905-05
In stock.
$113.00 USD/EACH
Qty:
Representative photo only Thermo Scientific Slide A Lyzer Dialysis Kit 10 000 MWCO 0 5 3mL 10 cassettes 10 buoys 10 syringes
Thermo Scientific Slide-A-Lyzer Dialysis Kit, 10,000 MWCO, 0.5-3mL, 10 cassettes, 10 buoys, 10 syringes
Product Type Ready-to-use Dialysis Device,Material Regenerated Cellulose,Membrane MWCO (Daltons) 10000
EW-02905-43
Call 1-888-358-4705 for availability.
$122.00 USD/EACH
Qty:
Representative photo only Thermo Scientific Slide A Lyzer Dialysis Kit 7000 MWCO 0 5 3mL 10 cassettes 10 buoys 10 syringes
Thermo Scientific Slide-A-Lyzer Dialysis Kit, 7000 MWCO, 0.5-3mL, 10 cassettes, 10 buoys, 10 syringes
Product Type Ready-to-use Dialysis Device,Material Regenerated Cellulose,Membrane MWCO (Daltons) 7000
EW-02905-33
Usually ships in 10 days.
$122.00 USD/EACH
Qty:
Representative photo only Thermo Scientific Slide A Lyzer Dialysis Kit 3500 MWCO 0 5 3mL 10 cassettes 10 buoys 10 syringes
Thermo Scientific Slide-A-Lyzer Dialysis Kit, 3500 MWCO, 0.5-3mL, 10 cassettes, 10 buoys, 10 syringes
Product Type Ready-to-use Dialysis Device,Material Regenerated Cellulose,Membrane MWCO (Daltons) 3500
EW-02905-04
Usually ships in 4 days.
$122.00 USD/EACH
Qty:
Representative photo only Thermo Scientific Slide A Lyzer Dialysis Cassettes 10 000 MWCO 12 30mL 6 cassettes
Thermo Scientific Slide-A-Lyzer Dialysis Cassettes, 10,000 MWCO, 12-30mL, 6 cassettes
Product Type Ready-to-use Dialysis Device,Material Regenerated Cellulose,Membrane MWCO (Daltons) 10000
EW-02905-46
In stock.
$91.00 USD/PACKAGE
Qty:
EW-02905-30 Thermo Scientific Slide-A-Lyzer<small><sup>&reg;</sup></small> Dialysis Cassettes, 7000 MWCO, 0.1-0.5mL, 10 cassettes
Thermo Scientific Slide-A-Lyzer Dialysis Cassettes, 7000 MWCO, 0.1-0.5mL, 10 cassettes
Product Type Ready-to-use Dialysis Device,Material Regenerated Cellulose,Membrane MWCO (Daltons) 7000
EW-02905-30
Usually ships in 5 days.
$86.00 USD/EACH
Qty:
EW-02905-40 Thermo Scientific Slide-A-Lyzer<small><sup>&reg;</sup></small> Dialysis Cassettes, 10,000 MWCO, 0.1-0.5mL, 10 cassettes
Thermo Scientific Slide-A-Lyzer Dialysis Cassettes, 10,000 MWCO, 0.1-0.5mL, 10 cassettes
Product Type Ready-to-use Dialysis Device,Material Regenerated Cellulose,Membrane MWCO (Daltons) 10000
EW-02905-40
In stock.
$86.00 USD/EACH
Qty:
EW-02905-01 Thermo Scientific Slide-A-Lyzer<small><sup>&reg;</sup></small> Dialysis Cassettes, 3500 MWCO, 0.1-0.5mL, 10 cassettes
Thermo Scientific Slide-A-Lyzer Dialysis Cassettes, 3500 MWCO, 0.1-0.5mL, 10 cassettes
Product Type Ready-to-use Dialysis Device,Material Regenerated Cellulose,Membrane MWCO (Daltons) 3500
EW-02905-01
In stock.
$86.00 USD/EACH
Qty:
Representative photo only Thermo Scientific Slide A Lyzer Dialysis Cassettes 7000 MWCO 3 12mL 8 cassettes
Thermo Scientific Slide-A-Lyzer Dialysis Cassettes, 7000 MWCO, 3-12mL, 8 cassettes
Product Type Ready-to-use Dialysis Device,Material Regenerated Cellulose,Membrane MWCO (Daltons) 7000
EW-02905-34
Usually ships in 5 days.
$113.00 USD/EACH
Qty:
Representative photo only Thermo Scientific Slide A Lyzer Dialysis Cassettes 7000 MWCO 0 5 3mL 10 cassettes
Thermo Scientific Slide-A-Lyzer Dialysis Cassettes, 7000 MWCO, 0.5-3mL, 10 cassettes
Product Type Ready-to-use Dialysis Device,Material Regenerated Cellulose,Membrane MWCO (Daltons) 7000
EW-02905-32
Usually ships in 2 days.
$86.00 USD/EACH
Qty:
Representative photo only Thermo Scientific Slide A Lyzer Dialysis Cassettes 3500 MWCO 12 30mL 6 cassettes
Thermo Scientific Slide-A-Lyzer Dialysis Cassettes, 3500 MWCO, 12-30mL, 6 cassettes
Product Type Ready-to-use Dialysis Device,Material Regenerated Cellulose,Membrane MWCO (Daltons) 3500
EW-02905-07
In stock.
$91.00 USD/PACKAGE
Qty:
Representative photo only Thermo Scientific Slide A Lyzer Dialysis Kit 3500 MWCO 3 12mL 8 cassettes 8 buoys 8 syringes
Thermo Scientific Slide-A-Lyzer Dialysis Kit, 3500 MWCO, 3-12mL, 8 cassettes, 8 buoys, 8 syringes
Product Type Ready-to-use Dialysis Device,Material Regenerated Cellulose,Membrane MWCO (Daltons) 3500
EW-02905-06
Usually ships in 4 days.
$139.00 USD/EACH
Qty:
19 results (showing 1 - 15 )
Per Page 
Home  >  Life Science > Dialysis > Ready-to-use Dialysis Devices > Thermo Scientific Slide-A-Lyzer Dialysis Cassettes
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title