United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
CE Compliance:
Yes
(1)
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Home
>
Heating Equipment
>
Hot Plates
>
Digital Hot Plates
>
Digital Aluminum Top Hot Plates
> Thermo Scientific Super-Nuova Hot Plates, Aluminum Top
Thermo Scientific Super-Nuova Hot Plates, Aluminum Top
Reliable, repeatable performance with maximum precision, safety, and convenience
Precise adjustment of settings in 1° increments displayed on bright LED display
Flashing, red ''hot top'' light provides visual warning to prevent accidental burns
Super-Nuova units feature a convenient timer to shut off the unit when the timed cycle is complete, allows for unattended use and prevents accidental overheating. Digitally adjust temperature in 1° increments up to 698°F (370°C). External probe (included) provides for added monitoring and control of liquid temperature. RS-232 port allows for output of time count, temperature, and set point directly to your PC.
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Compare
Product Detail
Sort
Most Popular
Name
Name Descending
Price (Lowest First)
Price (Highest First)
Availability
Price
Thermo Scientific Super-Nuova
aluminum hot plate, 7 x 7" 120vac
Top plate material Aluminum,Power VAC 120,Brand Super-Nuova
EW-86576-30
Usually ships in 7 days.
$1,015.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Thermo Scientific Super-Nuova Hot Plates, Aluminum Top
Thermo Scientific Super-Nuova
aluminum hot plate, 7 x 7" 220-240v
Top plate material Aluminum,Power VAC 220-240,CE Compliance Yes
EW-86576-35
Usually ships in 13 days.
$1,015.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Thermo Scientific Super-Nuova Hot Plates, Aluminum Top
2
results (showing
1
-
2
)
Per Page
5
10
15
25
50
Home
>
Heating Equipment
>
Hot Plates
>
Digital Hot Plates
>
Digital Aluminum Top Hot Plates
> Thermo Scientific Super-Nuova Hot Plates, Aluminum Top
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title