Item Total = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
Price Range:
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 8 ext. 138-8456
 

Tin Standard

Tin Standard
Choose from our comprehensive line of Tin Standard, 1 mL = 1 mg Sn, 1000 ppm Sn and other Chemicals T products.
 Product Detail
Sort
 Availability
 Price

Tin Standard, 1 mL = 1 mg Sn, 1000 ppm Sn (120 mL)

  • Product Type: Inorganic Chemical
  • Purity grade: Components all ACS or Reagent grade
  • Form: Liquid
EW-86992-58
Usually ships in 7 days.
$26.25 USD/EACH

Qty:

Tin Standard, 1 mL = 1 mg Sn, 1000 ppm Sn (500 mL)

  • Product Type: Inorganic Chemical
  • Purity grade: Components all ACS or Reagent grade
  • Form: Liquid
EW-86976-91
Usually ships in 14 days.
$61.75 USD/EACH

Qty:

2 results (showing 1 - 2 )
Per Page 
#PRODUCTTITLE#

#PRODUCTHEADER#

#IAMGESTICKER#
#CROSSOUTPRICE#
#PRICE#
#PRICEWITHVAT#
#AVAILABILITY#
Qty
Quantity Discounts
Qty: #QTY10COUNT# #QTY15COUNT# #QTY20COUNT#
Price: #QTY10PRICE# #QTY15PRICE# #QTY20PRICE#
#PRODUCTIMAGE#
#POWERREVIEW#
#MARKETINGTEXT#
#ICONS#
#SPECS#