log in or register
Price Range:
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
Home  >  Safety Products > Gas Detectors > Single Gas Detection > ToxiPro Personal Single Gas Detectors
ToxiPro Personal Single Gas Detectors
ToxiPro Personal Single Gas Detectors
  • Fully serviceable and reusable detector with real-time readings
  • IP65/IP67-rated protection against water and dust ingress
This fully serviceable and reusable personal gas detector features real-time Peak, STEL, and TWA readings. Detector also has internal "black-box" data recorder. Event logger stores the last 20 events and sensor type, max reading, average reading, time, and duration. Calibration is made easy by use of a single button. Regular maintenance, bump testing, and calibration can be performed using the IQ Express docking station (sold separately). Detectors also feature audible, visual, and vibrating alarms which are set off for Low, High STEL, and TWA readings. Readings are displayed on a large easy-to-see backlit display. Compact, rugged molded-over boot design is IP65/IP67 rated for protection against dust and water ingress. Unit also exceeds IEC 6100-4-3 for level 3 RFI immunity specifications.
 Product Detail
Sort
 Availability
 Price
Representative photo only ToxiPro Single Gas Detector CO
ToxiPro Single Gas Detector, CO
Product Type Single Gas Monitors/Detectors,Display LCD,Alarm 92dB audible, LED visual, vibrating
EW-62864-02
Usually ships in 9 days.
$274.00 USD/EACH
Qty:
Representative photo only ToxiPro Single Gas Detector O2
ToxiPro Single Gas Detector, O2
Product Type Single Gas Monitors/Detectors,Display LCD,Alarm 92dB audible, LED visual, vibrating
EW-62864-00
Usually ships in 3 days.
$264.00 USD/EACH
Qty:
Representative photo only ToxiPro Single Gas Detector SO2
ToxiPro Single Gas Detector, SO2
Product Type Single Gas Monitors/Detectors,Display LCD,Alarm 92dB audible, LED visual, vibrating
EW-62864-06
Usually ships in 2 days.
$385.00 USD/EACH
Qty:
Representative photo only ToxiPro Single Gas Detector CO Minus H
ToxiPro Single Gas Detector, CO Minus H
Product Type Single Gas Monitors/Detectors,Display LCD,Alarm 92dB audible, LED visual, vibrating
EW-62864-10
Usually ships in 5 days.
$334.00 USD/EACH
Qty:
EW-62864-04 ToxiPro Single Gas Detector, H<sub>2</sub>S
ToxiPro Single Gas Detector, H2S
Product Type Single Gas Monitors/Detectors,Display LCD,Alarm 92dB audible, LED visual, vibrating
EW-62864-04
Usually ships in 11 days.
$274.00 USD/EACH
Qty:
5 results (showing 1 - 5 )
Per Page 
Home  >  Safety Products > Gas Detectors > Single Gas Detection > ToxiPro Personal Single Gas Detectors
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title